Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 57362671-57363182 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ST8SIA3
Entrez Gene ID
51046
Gene Name
ST8 alpha-N-acetyl-neuraminide alpha-2,8-sialyltransferase 3
Gene Aliases
SIAT8C, ST8SiaIII
UniGene
Hs.23172
Gene Chromosome Location
Chr.18: 57352490-57368931 on Build GRCh38
Transcripts
NM_015879.2

SNP Detail

SNP ID Chromosome Location
rs34393724 Chr.18: 57363045-57363046 on Build GRCh38
rs1022898387 Chr.18: 57363004-57363004 on Build GRCh38
rs77792611 Chr.18: 57362820-57362820 on Build GRCh38
rs770786211 Chr.18: 57362814-57362817 on Build GRCh38
rs931994115 Chr.18: 57363096-57363096 on Build GRCh38
rs966952611 Chr.18: 57363016-57363016 on Build GRCh38
rs974672668 Chr.18: 57362981-57362981 on Build GRCh38
rs568010701 Chr.18: 57362891-57362891 on Build GRCh38
rs1011051002 Chr.18: 57363002-57363002 on Build GRCh38
rs1054916945 Chr.18: 57362766-57362766 on Build GRCh38
rs138770889 Chr.18: 57363064-57363064 on Build GRCh38
rs1017670798 Chr.18: 57362938-57362938 on Build GRCh38
rs940503743 Chr.18: 57362982-57362982 on Build GRCh38
rs899170049 Chr.18: 57362717-57362717 on Build GRCh38
rs964828281 Chr.18: 57362975-57362975 on Build GRCh38
rs939042099 Chr.18: 57362742-57362742 on Build GRCh38
rs916309868 Chr.18: 57363115-57363115 on Build GRCh38
rs114036826 Chr.18: 57362783-57362783 on Build GRCh38
rs535352603 Chr.18: 57363023-57363023 on Build GRCh38
rs536738914 Chr.18: 57362805-57362805 on Build GRCh38
rs990551040 Chr.18: 57363107-57363107 on Build GRCh38
rs890495204 Chr.18: 57362880-57362880 on Build GRCh38
rs897044076 Chr.18: 57362827-57362827 on Build GRCh38
rs927235666 Chr.18: 57362921-57362921 on Build GRCh38
rs115661589 Chr.18: 57362733-57362733 on Build GRCh38
rs1050402972 Chr.18: 57363104-57363104 on Build GRCh38
rs960390872 Chr.18: 57363068-57363068 on Build GRCh38
rs1047682622 Chr.18: 57362871-57362871 on Build GRCh38
rs1021338227 Chr.18: 57362825-57362825 on Build GRCh38
rs771756341 Chr.18: 57363144-57363144 on Build GRCh38
rs907706092 Chr.18: 57362978-57362978 on Build GRCh38
rs973901500 Chr.18: 57363145-57363145 on Build GRCh38
rs1010403203 Chr.18: 57362825-57362826 on Build GRCh38
rs890693451 Chr.18: 57362755-57362755 on Build GRCh38
rs994501499 Chr.18: 57362856-57362856 on Build GRCh38
rs770819614 Chr.18: 57362789-57362789 on Build GRCh38
rs759320200 Chr.18: 57363033-57363033 on Build GRCh38
rs1009150927 Chr.18: 57363133-57363133 on Build GRCh38
rs996150924 Chr.18: 57362728-57362728 on Build GRCh38
rs1008927679 Chr.18: 57362894-57362894 on Build GRCh38

View Full Product Details