Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 7697114-7697616 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FCER2
Entrez Gene ID
2208
Gene Name
Fc fragment of IgE receptor II
Gene Aliases
BLAST-2, CD23, CD23A, CLEC4J, FCE2, IGEBF
UniGene
Hs.465778
Gene Chromosome Location
Chr.19: 7688757-7702755 on Build GRCh38
Transcripts
NM_001207019.2 NM_001220500.1 NM_002002.4 XM_005272462.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3693220 c.226G>A p.G76S Chr.19: 7697554-7697554 on Build GRCh38
4534352 c.207G>A p.K69K Chr.19: 7697573-7697573 on Build GRCh38
2729443 c.233A>G p.Q78R Chr.19: 7697547-7697547 on Build GRCh38
6876862 c.243G>A p.Q81Q Chr.19: 7697537-7697537 on Build GRCh38
713890 c.258G>T p.T86T Chr.19: 7697294-7697294 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767687089 Chr.19: 7697265-7697265 on Build GRCh38
rs926107334 Chr.19: 7697360-7697360 on Build GRCh38
rs747562783 Chr.19: 7697515-7697515 on Build GRCh38
rs771565535 Chr.19: 7697205-7697205 on Build GRCh38
rs756249131 Chr.19: 7697269-7697269 on Build GRCh38
rs567385173 Chr.19: 7697288-7697288 on Build GRCh38
rs780566367 Chr.19: 7697323-7697323 on Build GRCh38
rs200021981 Chr.19: 7697576-7697576 on Build GRCh38
rs145568326 Chr.19: 7697541-7697541 on Build GRCh38
rs777793578 Chr.19: 7697281-7697281 on Build GRCh38
rs964854730 Chr.19: 7697431-7697431 on Build GRCh38
rs201038165 Chr.19: 7697540-7697540 on Build GRCh38
rs746460429 Chr.19: 7697302-7697302 on Build GRCh38
rs765140667 Chr.19: 7697229-7697229 on Build GRCh38
rs760527619 Chr.19: 7697477-7697477 on Build GRCh38
rs894639849 Chr.19: 7697588-7697588 on Build GRCh38
rs761817576 Chr.19: 7697567-7697567 on Build GRCh38
rs1024687746 Chr.19: 7697545-7697545 on Build GRCh38
rs913497913 Chr.19: 7697415-7697415 on Build GRCh38
rs555886538 Chr.19: 7697394-7697394 on Build GRCh38
rs74980343 Chr.19: 7697133-7697133 on Build GRCh38
rs996511866 Chr.19: 7697436-7697436 on Build GRCh38
rs780797483 Chr.19: 7697529-7697529 on Build GRCh38
rs1047318984 Chr.19: 7697518-7697518 on Build GRCh38
rs757538767 Chr.19: 7697479-7697479 on Build GRCh38
rs774140522 Chr.19: 7697551-7697551 on Build GRCh38
rs1057220341 Chr.19: 7697154-7697154 on Build GRCh38
rs887293379 Chr.19: 7697134-7697134 on Build GRCh38
rs763027862 Chr.19: 7697261-7697261 on Build GRCh38
rs1003427637 Chr.19: 7697148-7697148 on Build GRCh38
rs72558007 Chr.19: 7697554-7697554 on Build GRCh38
rs752775281 Chr.19: 7697266-7697266 on Build GRCh38
rs776374918 Chr.19: 7697220-7697220 on Build GRCh38
rs751548674 Chr.19: 7697329-7697329 on Build GRCh38
rs149706196 Chr.19: 7697532-7697532 on Build GRCh38
rs779315614 Chr.19: 7697290-7697290 on Build GRCh38
rs746343109 Chr.19: 7697294-7697294 on Build GRCh38
rs770864181 Chr.19: 7697549-7697549 on Build GRCh38
rs779227385 Chr.19: 7697481-7697481 on Build GRCh38
rs528355833 Chr.19: 7697173-7697173 on Build GRCh38
rs765399497 Chr.19: 7697572-7697572 on Build GRCh38
rs375829712 Chr.19: 7697560-7697560 on Build GRCh38
rs1015446614 Chr.19: 7697482-7697482 on Build GRCh38
rs973106418 Chr.19: 7697445-7697445 on Build GRCh38
rs377619010 Chr.19: 7697574-7697574 on Build GRCh38
rs761596541 Chr.19: 7697313-7697313 on Build GRCh38
rs771647021 Chr.19: 7697427-7697427 on Build GRCh38
rs558068649 Chr.19: 7697421-7697421 on Build GRCh38
rs894515778 Chr.19: 7697172-7697172 on Build GRCh38
rs773195059 Chr.19: 7697316-7697316 on Build GRCh38
rs760616294 Chr.19: 7697335-7697335 on Build GRCh38
rs200169018 Chr.19: 7697240-7697240 on Build GRCh38
rs750733191 Chr.19: 7697487-7697487 on Build GRCh38
rs771870116 Chr.19: 7697494-7697495 on Build GRCh38
rs866074528 Chr.19: 7697530-7697530 on Build GRCh38
rs567980420 Chr.19: 7697428-7697428 on Build GRCh38
rs747631191 Chr.19: 7697383-7697383 on Build GRCh38
rs1028077878 Chr.19: 7697159-7697159 on Build GRCh38
rs71179125 Chr.19: 7697499-7697507 on Build GRCh38
rs980176219 Chr.19: 7697555-7697555 on Build GRCh38
rs772612971 Chr.19: 7697295-7697295 on Build GRCh38
rs1031564800 Chr.19: 7697393-7697393 on Build GRCh38
rs768050213 Chr.19: 7697525-7697525 on Build GRCh38
rs936188137 Chr.19: 7697370-7697370 on Build GRCh38
rs113716290 Chr.19: 7697495-7697503 on Build GRCh38
rs186188865 Chr.19: 7697404-7697404 on Build GRCh38
rs769684627 Chr.19: 7697315-7697315 on Build GRCh38
rs928944498 Chr.19: 7697495-7697495 on Build GRCh38
rs780558442 Chr.19: 7697300-7697300 on Build GRCh38
rs768213654 Chr.19: 7697308-7697308 on Build GRCh38
rs868740945 Chr.19: 7697569-7697569 on Build GRCh38
rs776268077 Chr.19: 7697587-7697587 on Build GRCh38
rs572304844 Chr.19: 7697397-7697397 on Build GRCh38
rs571262008 Chr.19: 7697216-7697216 on Build GRCh38
rs754001038 Chr.19: 7697344-7697344 on Build GRCh38
rs780470543 Chr.19: 7697512-7697512 on Build GRCh38
rs961384969 Chr.19: 7697247-7697247 on Build GRCh38
rs535989649 Chr.19: 7697311-7697311 on Build GRCh38
rs1005420586 Chr.19: 7697210-7697210 on Build GRCh38
rs750645161 Chr.19: 7697584-7697584 on Build GRCh38
rs543826378 Chr.19: 7697449-7697449 on Build GRCh38
rs542037732 Chr.19: 7697257-7697257 on Build GRCh38
rs201017112 Chr.19: 7697501-7697501 on Build GRCh38
rs1049503634 Chr.19: 7697440-7697440 on Build GRCh38
rs117434896 Chr.19: 7697219-7697219 on Build GRCh38
rs764291342 Chr.19: 7697340-7697340 on Build GRCh38
rs952547310 Chr.19: 7697203-7697203 on Build GRCh38
rs113240986 Chr.19: 7697407-7697407 on Build GRCh38
rs866167964 Chr.19: 7697236-7697236 on Build GRCh38
rs551705532 Chr.19: 7697195-7697195 on Build GRCh38
rs888270080 Chr.19: 7697455-7697455 on Build GRCh38
rs766379194 Chr.19: 7697325-7697325 on Build GRCh38
rs754102526 Chr.19: 7697282-7697282 on Build GRCh38
rs376024411 Chr.19: 7697226-7697226 on Build GRCh38
rs998801664 Chr.19: 7697336-7697336 on Build GRCh38
rs764048367 Chr.19: 7697558-7697558 on Build GRCh38

View Full Product Details