Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 7901371-7901914 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LRRC8E
Entrez Gene ID
80131
Gene Name
leucine rich repeat containing 8 family member E
Gene Aliases
-
UniGene
Hs.501511
Gene Chromosome Location
Chr.19: 7888505-7902023 on Build GRCh38
Transcripts
NM_001268284.2 NM_001268285.2 NM_025061.5 XM_011528319.2

SNP Detail

SNP ID Chromosome Location
rs78102769 Chr.19: 7901728-7901728 on Build GRCh38
rs985990617 Chr.19: 7901556-7901556 on Build GRCh38
rs545589606 Chr.19: 7901882-7901882 on Build GRCh38
rs902867662 Chr.19: 7901697-7901697 on Build GRCh38
rs896771636 Chr.19: 7901403-7901403 on Build GRCh38
rs908825142 Chr.19: 7901604-7901604 on Build GRCh38
rs1014335066 Chr.19: 7901415-7901415 on Build GRCh38
rs952815776 Chr.19: 7901799-7901799 on Build GRCh38
rs780361726 Chr.19: 7901623-7901626 on Build GRCh38
rs1057376697 Chr.19: 7901634-7901634 on Build GRCh38
rs193048897 Chr.19: 7901625-7901625 on Build GRCh38
rs752487631 Chr.19: 7901495-7901495 on Build GRCh38
rs902897967 Chr.19: 7901493-7901493 on Build GRCh38
rs1032748777 Chr.19: 7901731-7901731 on Build GRCh38
rs551274526 Chr.19: 7901502-7901502 on Build GRCh38
rs1000700426 Chr.19: 7901431-7901431 on Build GRCh38
rs988124379 Chr.19: 7901465-7901465 on Build GRCh38
rs908731715 Chr.19: 7901814-7901814 on Build GRCh38
rs539167913 Chr.19: 7901698-7901698 on Build GRCh38
rs185355470 Chr.19: 7901645-7901645 on Build GRCh38
rs949040613 Chr.19: 7901674-7901674 on Build GRCh38
rs1045782389 Chr.19: 7901694-7901694 on Build GRCh38
rs753617084 Chr.19: 7901519-7901519 on Build GRCh38
rs56688769 Chr.19: 7901862-7901862 on Build GRCh38
rs966025652 Chr.19: 7901598-7901598 on Build GRCh38
rs940238680 Chr.19: 7901628-7901628 on Build GRCh38
rs904931095 Chr.19: 7901690-7901690 on Build GRCh38
rs975911038 Chr.19: 7901557-7901557 on Build GRCh38
rs1032252378 Chr.19: 7901436-7901436 on Build GRCh38
rs74710490 Chr.19: 7901715-7901715 on Build GRCh38
rs921318828 Chr.19: 7901567-7901567 on Build GRCh38
rs566860859 Chr.19: 7901678-7901678 on Build GRCh38
rs957172684 Chr.19: 7901549-7901549 on Build GRCh38
rs1033851476 Chr.19: 7901706-7901706 on Build GRCh38
rs113001695 Chr.19: 7901788-7901788 on Build GRCh38
rs1019456321 Chr.19: 7901759-7901759 on Build GRCh38
rs965761684 Chr.19: 7901781-7901781 on Build GRCh38
rs984823378 Chr.19: 7901637-7901637 on Build GRCh38
rs896691007 Chr.19: 7901675-7901675 on Build GRCh38
rs984281568 Chr.19: 7901599-7901599 on Build GRCh38
rs368424098 Chr.19: 7901741-7901741 on Build GRCh38
rs976447203 Chr.19: 7901603-7901603 on Build GRCh38
rs758184711 Chr.19: 7901786-7901786 on Build GRCh38
rs5826989 Chr.19: 7901714-7901715 on Build GRCh38
rs1023976994 Chr.19: 7901467-7901467 on Build GRCh38
rs965378272 Chr.19: 7901504-7901504 on Build GRCh38
rs931513934 Chr.19: 7901583-7901583 on Build GRCh38
rs527332780 Chr.19: 7901715-7901716 on Build GRCh38
rs1009452429 Chr.19: 7901748-7901748 on Build GRCh38
rs896159865 Chr.19: 7901638-7901638 on Build GRCh38
rs749876265 Chr.19: 7901704-7901704 on Build GRCh38
rs397860135 Chr.19: 7901728-7901728 on Build GRCh38
rs397810755 Chr.19: 7901728-7901729 on Build GRCh38
rs536651853 Chr.19: 7901639-7901639 on Build GRCh38
rs938207568 Chr.19: 7901396-7901396 on Build GRCh38
rs778271596 Chr.19: 7901413-7901413 on Build GRCh38
rs572413961 Chr.19: 7901891-7901891 on Build GRCh38
rs1032820813 Chr.19: 7901513-7901513 on Build GRCh38
rs147563019 Chr.19: 7901649-7901651 on Build GRCh38
rs78725194 Chr.19: 7901729-7901729 on Build GRCh38
rs1017525687 Chr.19: 7901582-7901582 on Build GRCh38
rs34385743 Chr.19: 7901715-7901715 on Build GRCh38
rs956608933 Chr.19: 7901439-7901439 on Build GRCh38
rs575428230 Chr.19: 7901867-7901867 on Build GRCh38
rs1028861142 Chr.19: 7901787-7901787 on Build GRCh38
rs984314068 Chr.19: 7901808-7901808 on Build GRCh38
rs537609540 Chr.19: 7901877-7901877 on Build GRCh38
rs993080916 Chr.19: 7901869-7901869 on Build GRCh38
rs567393265 Chr.19: 7901570-7901570 on Build GRCh38
rs1055292890 Chr.19: 7901400-7901400 on Build GRCh38
rs1001135369 Chr.19: 7901693-7901693 on Build GRCh38
rs576533368 Chr.19: 7901703-7901703 on Build GRCh38
rs969627709 Chr.19: 7901399-7901399 on Build GRCh38
rs917502170 Chr.19: 7901889-7901889 on Build GRCh38

View Full Product Details