Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 8316406-8316914 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NDUFA7
Entrez Gene ID
4701
Gene Name
NADH:ubiquinone oxidoreductase subunit A7
Gene Aliases
B14.5a, CI-B14.5a
UniGene
Hs.333427
Gene Chromosome Location
Chr.19: 8308603-8321413 on Build GRCh38
Transcripts
NM_005001.4 NR_135539.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3797711 c.102-1G>T p.? Chr.19: 8316646-8316646 on Build GRCh38
3363086 c.166A>G p.T56A Chr.19: 8316581-8316581 on Build GRCh38
7161451 c.148T>C p.S50P Chr.19: 8316599-8316599 on Build GRCh38
3836190 c.227T>C p.L76P Chr.19: 8316520-8316520 on Build GRCh38
4581594 c.164G>A p.C55Y Chr.19: 8316583-8316583 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs373274849 Chr.19: 8316640-8316640 on Build GRCh38
rs572388548 Chr.19: 8316768-8316768 on Build GRCh38
rs756774473 Chr.19: 8316583-8316583 on Build GRCh38
rs146105751 Chr.19: 8316595-8316595 on Build GRCh38
rs914291455 Chr.19: 8316826-8316826 on Build GRCh38
rs754365850 Chr.19: 8316648-8316648 on Build GRCh38
rs200664279 Chr.19: 8316485-8316485 on Build GRCh38
rs748877322 Chr.19: 8316532-8316532 on Build GRCh38
rs768386495 Chr.19: 8316776-8316776 on Build GRCh38
rs1054096928 Chr.19: 8316460-8316460 on Build GRCh38
rs751823118 Chr.19: 8316505-8316505 on Build GRCh38
rs760613363 Chr.19: 8316462-8316462 on Build GRCh38
rs745859027 Chr.19: 8316453-8316453 on Build GRCh38
rs180752715 Chr.19: 8316847-8316847 on Build GRCh38
rs746580634 Chr.19: 8316688-8316688 on Build GRCh38
rs564263982 Chr.19: 8316480-8316480 on Build GRCh38
rs761781334 Chr.19: 8316613-8316613 on Build GRCh38
rs534499409 Chr.19: 8316578-8316578 on Build GRCh38
rs924319736 Chr.19: 8316450-8316450 on Build GRCh38
rs1031570989 Chr.19: 8316812-8316812 on Build GRCh38
rs768933004 Chr.19: 8316628-8316628 on Build GRCh38
rs547697707 Chr.19: 8316582-8316582 on Build GRCh38
rs567865675 Chr.19: 8316609-8316609 on Build GRCh38
rs765704343 Chr.19: 8316666-8316666 on Build GRCh38
rs2288415 Chr.19: 8316551-8316551 on Build GRCh38
rs372157683 Chr.19: 8316461-8316461 on Build GRCh38
rs934427634 Chr.19: 8316454-8316454 on Build GRCh38
rs778542183 Chr.19: 8316534-8316534 on Build GRCh38
rs755990875 Chr.19: 8316713-8316713 on Build GRCh38
rs749354422 Chr.19: 8316531-8316533 on Build GRCh38
rs774369260 Chr.19: 8316630-8316631 on Build GRCh38
rs966535443 Chr.19: 8316441-8316441 on Build GRCh38
rs3214554 Chr.19: 8316712-8316713 on Build GRCh38
rs749987452 Chr.19: 8316682-8316682 on Build GRCh38
rs897390941 Chr.19: 8316555-8316555 on Build GRCh38
rs781082313 Chr.19: 8316696-8316696 on Build GRCh38
rs372389586 Chr.19: 8316500-8316500 on Build GRCh38
rs147485841 Chr.19: 8316821-8316821 on Build GRCh38
rs367964698 Chr.19: 8316456-8316456 on Build GRCh38
rs375031844 Chr.19: 8316591-8316591 on Build GRCh38
rs748124558 Chr.19: 8316697-8316697 on Build GRCh38
rs764308196 Chr.19: 8316646-8316646 on Build GRCh38
rs768691484 Chr.19: 8316553-8316553 on Build GRCh38
rs866957932 Chr.19: 8316586-8316586 on Build GRCh38
rs769685392 Chr.19: 8316496-8316497 on Build GRCh38
rs753164049 Chr.19: 8316523-8316523 on Build GRCh38
rs774530389 Chr.19: 8316622-8316622 on Build GRCh38
rs368171790 Chr.19: 8316493-8316493 on Build GRCh38
rs115023062 Chr.19: 8316834-8316834 on Build GRCh38
rs548944919 Chr.19: 8316569-8316569 on Build GRCh38
rs12986323 Chr.19: 8316605-8316605 on Build GRCh38
rs767828157 Chr.19: 8316629-8316629 on Build GRCh38
rs368535993 Chr.19: 8316522-8316522 on Build GRCh38
rs371747571 Chr.19: 8316566-8316566 on Build GRCh38
rs764256926 Chr.19: 8316654-8316654 on Build GRCh38
rs1045914969 Chr.19: 8316701-8316701 on Build GRCh38
rs375994897 Chr.19: 8316628-8316628 on Build GRCh38
rs561540825 Chr.19: 8316436-8316437 on Build GRCh38
rs532995788 Chr.19: 8316574-8316574 on Build GRCh38
rs778109662 Chr.19: 8316531-8316531 on Build GRCh38
rs976553559 Chr.19: 8316449-8316449 on Build GRCh38
rs780171164 Chr.19: 8316507-8316511 on Build GRCh38
rs767232911 Chr.19: 8316459-8316459 on Build GRCh38
rs763195618 Chr.19: 8316558-8316558 on Build GRCh38
rs756602427 Chr.19: 8316530-8316530 on Build GRCh38
rs555730635 Chr.19: 8316711-8316711 on Build GRCh38
rs751841437 Chr.19: 8316568-8316568 on Build GRCh38
rs771154226 Chr.19: 8316616-8316616 on Build GRCh38
rs778346181 Chr.19: 8316584-8316584 on Build GRCh38
rs541013543 Chr.19: 8316452-8316452 on Build GRCh38
rs762268210 Chr.19: 8316685-8316686 on Build GRCh38
rs950838980 Chr.19: 8316539-8316539 on Build GRCh38
rs199760005 Chr.19: 8316685-8316685 on Build GRCh38
rs189447891 Chr.19: 8316690-8316690 on Build GRCh38
rs771878274 Chr.19: 8316545-8316545 on Build GRCh38
rs745465916 Chr.19: 8316535-8316535 on Build GRCh38
rs971011250 Chr.19: 8316580-8316580 on Build GRCh38
rs893523076 Chr.19: 8316747-8316747 on Build GRCh38
rs746884067 Chr.19: 8316600-8316600 on Build GRCh38
rs748176616 Chr.19: 8316473-8316473 on Build GRCh38
rs544116318 Chr.19: 8316700-8316700 on Build GRCh38
rs978377977 Chr.19: 8316803-8316803 on Build GRCh38
rs757723027 Chr.19: 8316653-8316653 on Build GRCh38
rs971361591 Chr.19: 8316788-8316788 on Build GRCh38
rs1024932141 Chr.19: 8316786-8316786 on Build GRCh38
rs943051527 Chr.19: 8316642-8316642 on Build GRCh38
rs375339162 Chr.19: 8316472-8316472 on Build GRCh38
rs773969783 Chr.19: 8316455-8316455 on Build GRCh38
rs937470712 Chr.19: 8316724-8316724 on Build GRCh38
rs886303436 Chr.19: 8316720-8316720 on Build GRCh38
rs755394650 Chr.19: 8316508-8316508 on Build GRCh38
rs909878226 Chr.19: 8316481-8316481 on Build GRCh38
rs1017034325 Chr.19: 8316868-8316868 on Build GRCh38
rs371568456 Chr.19: 8316825-8316825 on Build GRCh38
rs745552595 Chr.19: 8316589-8316589 on Build GRCh38
rs759986417 Chr.19: 8316733-8316733 on Build GRCh38
rs985572336 Chr.19: 8316644-8316644 on Build GRCh38
rs959357059 Chr.19: 8316749-8316749 on Build GRCh38
rs1001956315 Chr.19: 8316577-8316577 on Build GRCh38
rs776631139 Chr.19: 8316610-8316610 on Build GRCh38
rs902471773 Chr.19: 8316785-8316785 on Build GRCh38
rs142961225 Chr.19: 8316635-8316635 on Build GRCh38
rs200409749 Chr.19: 8316557-8316557 on Build GRCh38

View Full Product Details