Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 9160794-9161287 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF317
Entrez Gene ID
57693
Gene Name
zinc finger protein 317
Gene Aliases
-
UniGene
Hs.465829
Gene Chromosome Location
Chr.19: 9140380-9163419 on Build GRCh38
Transcripts
NM_001190791.1 NM_020933.4 NR_102435.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7331518 c.1239C>A p.I413I Chr.19: 9160884-9160884 on Build GRCh38
6466123 c.1611G>A p.K537K Chr.19: 9161256-9161256 on Build GRCh38
5614742 c.1225A>T p.M409L Chr.19: 9160870-9160870 on Build GRCh38
1004205 c.1311G>T p.M437I Chr.19: 9160956-9160956 on Build GRCh38
5542154 c.1348C>T p.L450F Chr.19: 9160993-9160993 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs189336858 Chr.19: 9161223-9161223 on Build GRCh38
rs759272450 Chr.19: 9161220-9161220 on Build GRCh38
rs766844062 Chr.19: 9160851-9160851 on Build GRCh38
rs1001629552 Chr.19: 9161209-9161209 on Build GRCh38
rs759596223 Chr.19: 9160829-9160829 on Build GRCh38
rs778603488 Chr.19: 9160877-9160877 on Build GRCh38
rs773588755 Chr.19: 9161102-9161102 on Build GRCh38
rs766123277 Chr.19: 9161063-9161063 on Build GRCh38
rs866481787 Chr.19: 9160895-9160895 on Build GRCh38
rs767155522 Chr.19: 9161116-9161116 on Build GRCh38
rs774767292 Chr.19: 9160911-9160911 on Build GRCh38
rs753182381 Chr.19: 9160839-9160839 on Build GRCh38
rs774533817 Chr.19: 9161207-9161207 on Build GRCh38
rs756121555 Chr.19: 9161127-9161127 on Build GRCh38
rs186074655 Chr.19: 9161128-9161128 on Build GRCh38
rs757893621 Chr.19: 9160883-9160883 on Build GRCh38
rs767615889 Chr.19: 9160831-9160831 on Build GRCh38
rs1037002438 Chr.19: 9161101-9161101 on Build GRCh38
rs761191518 Chr.19: 9160846-9160846 on Build GRCh38
rs765623451 Chr.19: 9160889-9160889 on Build GRCh38
rs539384909 Chr.19: 9161150-9161150 on Build GRCh38
rs750743763 Chr.19: 9161052-9161052 on Build GRCh38
rs769753252 Chr.19: 9160998-9160998 on Build GRCh38
rs369391399 Chr.19: 9160959-9160959 on Build GRCh38
rs368955133 Chr.19: 9160907-9160907 on Build GRCh38
rs367743743 Chr.19: 9160884-9160884 on Build GRCh38
rs758187060 Chr.19: 9161242-9161242 on Build GRCh38
rs774714322 Chr.19: 9160827-9160827 on Build GRCh38
rs747262264 Chr.19: 9161147-9161147 on Build GRCh38
rs781139661 Chr.19: 9160897-9160897 on Build GRCh38
rs199778120 Chr.19: 9160964-9160964 on Build GRCh38
rs777170174 Chr.19: 9161032-9161032 on Build GRCh38
rs753854143 Chr.19: 9161233-9161233 on Build GRCh38
rs747209318 Chr.19: 9161078-9161078 on Build GRCh38
rs774095221 Chr.19: 9161111-9161111 on Build GRCh38
rs754535035 Chr.19: 9160989-9160989 on Build GRCh38
rs368386967 Chr.19: 9160830-9160830 on Build GRCh38
rs777668084 Chr.19: 9160999-9160999 on Build GRCh38
rs781451545 Chr.19: 9161250-9161250 on Build GRCh38
rs957403959 Chr.19: 9161192-9161192 on Build GRCh38
rs201883092 Chr.19: 9160927-9160927 on Build GRCh38
rs201446229 Chr.19: 9161067-9161067 on Build GRCh38
rs993703150 Chr.19: 9160935-9160935 on Build GRCh38
rs376791164 Chr.19: 9161083-9161083 on Build GRCh38
rs533693593 Chr.19: 9160858-9160858 on Build GRCh38
rs970144862 Chr.19: 9161176-9161176 on Build GRCh38
rs992191406 Chr.19: 9161082-9161082 on Build GRCh38
rs746098165 Chr.19: 9161014-9161014 on Build GRCh38
rs138057564 Chr.19: 9160885-9160885 on Build GRCh38
rs766634578 Chr.19: 9161110-9161110 on Build GRCh38
rs371860809 Chr.19: 9161072-9161072 on Build GRCh38
rs750075061 Chr.19: 9160881-9160881 on Build GRCh38
rs759775377 Chr.19: 9160912-9160912 on Build GRCh38
rs778538919 Chr.19: 9161144-9161144 on Build GRCh38
rs756994764 Chr.19: 9161077-9161077 on Build GRCh38
rs371181730 Chr.19: 9160852-9160852 on Build GRCh38
rs534702883 Chr.19: 9160920-9160920 on Build GRCh38
rs751186575 Chr.19: 9160972-9160972 on Build GRCh38
rs568121926 Chr.19: 9161053-9161053 on Build GRCh38
rs775432032 Chr.19: 9161016-9161016 on Build GRCh38
rs540055574 Chr.19: 9160859-9160859 on Build GRCh38
rs199708182 Chr.19: 9161161-9161161 on Build GRCh38
rs143496194 Chr.19: 9161076-9161076 on Build GRCh38
rs760875855 Chr.19: 9161017-9161017 on Build GRCh38
rs763488817 Chr.19: 9161232-9161232 on Build GRCh38
rs750081571 Chr.19: 9160976-9160976 on Build GRCh38
rs372679809 Chr.19: 9160938-9160938 on Build GRCh38
rs145091857 Chr.19: 9160981-9160981 on Build GRCh38
rs750153589 Chr.19: 9161241-9161241 on Build GRCh38
rs374974069 Chr.19: 9161114-9161114 on Build GRCh38
rs749731597 Chr.19: 9161193-9161193 on Build GRCh38
rs748669143 Chr.19: 9161094-9161094 on Build GRCh38
rs748026616 Chr.19: 9160901-9160901 on Build GRCh38
rs763437095 Chr.19: 9161105-9161105 on Build GRCh38
rs373290260 Chr.19: 9161036-9161036 on Build GRCh38
rs776650509 Chr.19: 9161088-9161088 on Build GRCh38
rs756065175 Chr.19: 9161074-9161074 on Build GRCh38
rs887672726 Chr.19: 9161091-9161091 on Build GRCh38
rs980263384 Chr.19: 9160906-9160906 on Build GRCh38
rs748079444 Chr.19: 9160993-9160993 on Build GRCh38
rs756906379 Chr.19: 9160983-9160983 on Build GRCh38
rs769750079 Chr.19: 9161164-9161164 on Build GRCh38
rs754588125 Chr.19: 9161068-9161068 on Build GRCh38
rs931782952 Chr.19: 9161061-9161061 on Build GRCh38
rs752194620 Chr.19: 9161126-9161126 on Build GRCh38
rs772275829 Chr.19: 9161015-9161015 on Build GRCh38
rs781339999 Chr.19: 9160990-9160990 on Build GRCh38
rs925817653 Chr.19: 9161168-9161168 on Build GRCh38
rs765101807 Chr.19: 9161236-9161236 on Build GRCh38
rs761096249 Chr.19: 9160928-9160928 on Build GRCh38
rs758021114 Chr.19: 9160980-9160980 on Build GRCh38
rs182231417 Chr.19: 9160896-9160896 on Build GRCh38
rs887902240 Chr.19: 9161047-9161047 on Build GRCh38
rs759227515 Chr.19: 9161113-9161113 on Build GRCh38
rs773822055 Chr.19: 9161186-9161186 on Build GRCh38
rs200506167 Chr.19: 9161073-9161073 on Build GRCh38
rs143272164 Chr.19: 9161133-9161133 on Build GRCh38
rs760315528 Chr.19: 9161231-9161231 on Build GRCh38
rs375676116 Chr.19: 9160956-9160956 on Build GRCh38
rs140353330 Chr.19: 9161095-9161095 on Build GRCh38
rs367620887 Chr.19: 9161154-9161154 on Build GRCh38
rs764513496 Chr.19: 9160949-9160949 on Build GRCh38
rs771360298 Chr.19: 9161201-9161201 on Build GRCh38
rs753705072 Chr.19: 9161132-9161132 on Build GRCh38
rs761613430 Chr.19: 9161235-9161235 on Build GRCh38
rs765614441 Chr.19: 9161051-9161051 on Build GRCh38
rs149506507 Chr.19: 9161031-9161031 on Build GRCh38
rs375442218 Chr.19: 9160857-9160857 on Build GRCh38

View Full Product Details