Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 9251721-9252222 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OR7E24
Entrez Gene ID
26648
Gene Name
olfactory receptor family 7 subfamily E member 24
Gene Aliases
HSHT2, OR19-8, OR7E24P, OR7E24Q
UniGene
Hs.129832
Gene Chromosome Location
Chr.19: 9251044-9252063 on Build GRCh38
Transcripts
NM_001079935.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5899765 c.725C>T p.P242L Chr.19: 9251768-9251768 on Build GRCh38
3960841 c.768C>A p.F256L Chr.19: 9251811-9251811 on Build GRCh38
5013363 c.841G>A p.A281T Chr.19: 9251884-9251884 on Build GRCh38
6892845 c.696C>T p.I232I Chr.19: 9251739-9251739 on Build GRCh38
5057133 c.764C>T p.A255V Chr.19: 9251807-9251807 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779242801 Chr.19: 9252022-9252022 on Build GRCh38
rs906706479 Chr.19: 9252152-9252152 on Build GRCh38
rs765820764 Chr.19: 9252077-9252077 on Build GRCh38
rs548186005 Chr.19: 9251859-9251859 on Build GRCh38
rs755850115 Chr.19: 9251753-9251753 on Build GRCh38
rs780002195 Chr.19: 9251984-9251984 on Build GRCh38
rs750040898 Chr.19: 9252060-9252060 on Build GRCh38
rs370482036 Chr.19: 9251860-9251860 on Build GRCh38
rs371305730 Chr.19: 9251807-9251807 on Build GRCh38
rs535452627 Chr.19: 9252105-9252105 on Build GRCh38
rs761124124 Chr.19: 9251932-9251932 on Build GRCh38
rs749985517 Chr.19: 9251978-9251978 on Build GRCh38
rs376341038 Chr.19: 9251966-9251966 on Build GRCh38
rs34791200 Chr.19: 9252073-9252074 on Build GRCh38
rs748342716 Chr.19: 9251814-9251826 on Build GRCh38
rs768175906 Chr.19: 9251913-9251913 on Build GRCh38
rs146858978 Chr.19: 9251769-9251769 on Build GRCh38
rs888942705 Chr.19: 9251866-9251866 on Build GRCh38
rs78240061 Chr.19: 9252129-9252129 on Build GRCh38
rs184473277 Chr.19: 9251869-9251869 on Build GRCh38
rs752472988 Chr.19: 9252095-9252095 on Build GRCh38
rs373369167 Chr.19: 9251927-9251927 on Build GRCh38
rs971374011 Chr.19: 9251959-9251959 on Build GRCh38
rs776919523 Chr.19: 9252056-9252056 on Build GRCh38
rs754665571 Chr.19: 9251799-9251799 on Build GRCh38
rs754925692 Chr.19: 9252089-9252089 on Build GRCh38
rs188631684 Chr.19: 9252094-9252094 on Build GRCh38
rs369434041 Chr.19: 9251986-9251986 on Build GRCh38
rs779946757 Chr.19: 9251863-9251863 on Build GRCh38
rs764306219 Chr.19: 9251938-9251938 on Build GRCh38
rs762144118 Chr.19: 9252059-9252059 on Build GRCh38
rs200671636 Chr.19: 9251907-9251907 on Build GRCh38
rs140655036 Chr.19: 9252033-9252033 on Build GRCh38
rs779298221 Chr.19: 9252111-9252111 on Build GRCh38
rs983621167 Chr.19: 9252028-9252028 on Build GRCh38
rs761441522 Chr.19: 9251956-9251956 on Build GRCh38
rs777264993 Chr.19: 9251754-9251754 on Build GRCh38
rs745746675 Chr.19: 9251778-9251778 on Build GRCh38
rs768120795 Chr.19: 9251833-9251833 on Build GRCh38
rs753380759 Chr.19: 9251848-9251848 on Build GRCh38
rs773509225 Chr.19: 9251888-9251888 on Build GRCh38
rs374983080 Chr.19: 9251861-9251861 on Build GRCh38
rs1031049529 Chr.19: 9251864-9251864 on Build GRCh38
rs374768729 Chr.19: 9251812-9251812 on Build GRCh38
rs757876440 Chr.19: 9251979-9251979 on Build GRCh38
rs753157431 Chr.19: 9251835-9251835 on Build GRCh38
rs768862407 Chr.19: 9251765-9251765 on Build GRCh38
rs553025686 Chr.19: 9252043-9252043 on Build GRCh38
rs58039687 Chr.19: 9252160-9252160 on Build GRCh38
rs537150980 Chr.19: 9251895-9251895 on Build GRCh38
rs201442314 Chr.19: 9252002-9252002 on Build GRCh38
rs771323788 Chr.19: 9252013-9252013 on Build GRCh38
rs770107221 Chr.19: 9251791-9251791 on Build GRCh38
rs773273915 Chr.19: 9251874-9251874 on Build GRCh38
rs780913120 Chr.19: 9251989-9251989 on Build GRCh38
rs761018732 Chr.19: 9252041-9252041 on Build GRCh38
rs771270861 Chr.19: 9251897-9251897 on Build GRCh38
rs774628570 Chr.19: 9251902-9251902 on Build GRCh38
rs746768648 Chr.19: 9251785-9251785 on Build GRCh38
rs751523150 Chr.19: 9252087-9252087 on Build GRCh38
rs770757986 Chr.19: 9252172-9252172 on Build GRCh38
rs753532329 Chr.19: 9251940-9251940 on Build GRCh38
rs917707 Chr.19: 9252123-9252123 on Build GRCh38
rs2240928 Chr.19: 9251767-9251767 on Build GRCh38
rs774516218 Chr.19: 9251811-9251813 on Build GRCh38
rs557329194 Chr.19: 9252117-9252117 on Build GRCh38
rs775330769 Chr.19: 9251782-9251782 on Build GRCh38
rs771860039 Chr.19: 9251779-9251779 on Build GRCh38
rs756402536 Chr.19: 9251840-9251840 on Build GRCh38
rs372301703 Chr.19: 9252031-9252031 on Build GRCh38
rs199799172 Chr.19: 9251825-9251825 on Build GRCh38
rs776075382 Chr.19: 9252035-9252035 on Build GRCh38
rs763838707 Chr.19: 9251847-9251847 on Build GRCh38
rs756803730 Chr.19: 9251857-9251857 on Build GRCh38
rs772296148 Chr.19: 9251839-9251839 on Build GRCh38
rs768299897 Chr.19: 9251883-9251883 on Build GRCh38
rs1037358915 Chr.19: 9252090-9252090 on Build GRCh38
rs773279915 Chr.19: 9251792-9251792 on Build GRCh38
rs762477321 Chr.19: 9252069-9252069 on Build GRCh38
rs559282731 Chr.19: 9251796-9251796 on Build GRCh38

View Full Product Details