Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 9538173-9538682 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC101928238
Entrez Gene ID
101928238
Gene Name
uncharacterized LOC101928238
Gene Aliases
-
UniGene
Hs.324978
Gene Chromosome Location
Chr.19: 9538666-9539730 on Build GRCh38
Transcripts
NR_134920.1 NR_134921.1 NM_001300883.2 NM_001318055.1 NM_001318056.1 NM_024106.2 XM_017027293.1 XM_017027294.1

Gene Symbol
ZNF426
Entrez Gene ID
79088
Gene Name
zinc finger protein 426
Gene Aliases
K-RBP
UniGene
Hs.386476
Gene Chromosome Location
Chr.19: 9523224-9538673 on Build GRCh38
Transcripts
NR_134920.1 NR_134921.1 NM_001300883.2 NM_001318055.1 NM_001318056.1 NM_024106.2 XM_017027293.1 XM_017027294.1

SNP Detail

SNP ID Chromosome Location
rs887614167 Chr.19: 9538560-9538560 on Build GRCh38
rs773299916 Chr.19: 9538595-9538595 on Build GRCh38
rs998494337 Chr.19: 9538515-9538515 on Build GRCh38
rs964125371 Chr.19: 9538351-9538351 on Build GRCh38
rs902966018 Chr.19: 9538494-9538494 on Build GRCh38
rs985960802 Chr.19: 9538317-9538317 on Build GRCh38
rs936151121 Chr.19: 9538455-9538455 on Build GRCh38
rs894186471 Chr.19: 9538251-9538251 on Build GRCh38
rs138572947 Chr.19: 9538582-9538582 on Build GRCh38
rs1042265359 Chr.19: 9538249-9538249 on Build GRCh38
rs910831459 Chr.19: 9538619-9538619 on Build GRCh38
rs944938921 Chr.19: 9538631-9538631 on Build GRCh38
rs73509624 Chr.19: 9538259-9538259 on Build GRCh38
rs1027012954 Chr.19: 9538599-9538599 on Build GRCh38
rs371037869 Chr.19: 9538632-9538632 on Build GRCh38
rs1027882529 Chr.19: 9538319-9538319 on Build GRCh38
rs1048774212 Chr.19: 9538545-9538545 on Build GRCh38
rs568483383 Chr.19: 9538445-9538445 on Build GRCh38
rs150566741 Chr.19: 9538566-9538566 on Build GRCh38
rs986253106 Chr.19: 9538616-9538616 on Build GRCh38
rs890340611 Chr.19: 9538559-9538559 on Build GRCh38
rs967377995 Chr.19: 9538576-9538576 on Build GRCh38
rs977512494 Chr.19: 9538446-9538446 on Build GRCh38
rs7250898 Chr.19: 9538402-9538402 on Build GRCh38
rs1021410018 Chr.19: 9538260-9538260 on Build GRCh38
rs776830601 Chr.19: 9538291-9538291 on Build GRCh38
rs1040136216 Chr.19: 9538297-9538297 on Build GRCh38
rs562018470 Chr.19: 9538611-9538611 on Build GRCh38
rs535876690 Chr.19: 9538448-9538448 on Build GRCh38
rs917367512 Chr.19: 9538424-9538424 on Build GRCh38
rs973282968 Chr.19: 9538587-9538587 on Build GRCh38
rs575686465 Chr.19: 9538502-9538502 on Build GRCh38
rs1033630976 Chr.19: 9538562-9538562 on Build GRCh38
rs114114920 Chr.19: 9538649-9538649 on Build GRCh38
rs1024332132 Chr.19: 9538574-9538574 on Build GRCh38
rs766199061 Chr.19: 9538635-9538635 on Build GRCh38
rs558932063 Chr.19: 9538246-9538246 on Build GRCh38
rs552128360 Chr.19: 9538369-9538369 on Build GRCh38
rs192327773 Chr.19: 9538201-9538201 on Build GRCh38
rs926021595 Chr.19: 9538584-9538584 on Build GRCh38
rs1030242637 Chr.19: 9538303-9538303 on Build GRCh38
rs894627141 Chr.19: 9538450-9538450 on Build GRCh38
rs947531148 Chr.19: 9538651-9538651 on Build GRCh38
rs897597398 Chr.19: 9538493-9538493 on Build GRCh38
rs1050637727 Chr.19: 9538457-9538457 on Build GRCh38
rs73509625 Chr.19: 9538648-9538648 on Build GRCh38
rs558071226 Chr.19: 9538564-9538564 on Build GRCh38
rs924336574 Chr.19: 9538655-9538655 on Build GRCh38
rs77558191 Chr.19: 9538393-9538393 on Build GRCh38
rs974554144 Chr.19: 9538360-9538360 on Build GRCh38
rs569679807 Chr.19: 9538308-9538308 on Build GRCh38
rs187490972 Chr.19: 9538200-9538200 on Build GRCh38
rs951477646 Chr.19: 9538621-9538621 on Build GRCh38
rs1030023457 Chr.19: 9538519-9538519 on Build GRCh38
rs530613225 Chr.19: 9538332-9538332 on Build GRCh38
rs1056379984 Chr.19: 9538247-9538247 on Build GRCh38
rs576967169 Chr.19: 9538270-9538270 on Build GRCh38
rs62105684 Chr.19: 9538368-9538368 on Build GRCh38
rs951888802 Chr.19: 9538350-9538350 on Build GRCh38
rs764775572 Chr.19: 9538491-9538491 on Build GRCh38
rs557306642 Chr.19: 9538452-9538452 on Build GRCh38
rs916189469 Chr.19: 9538650-9538650 on Build GRCh38
rs539916545 Chr.19: 9538506-9538506 on Build GRCh38
rs1007895678 Chr.19: 9538565-9538565 on Build GRCh38
rs183209574 Chr.19: 9538403-9538403 on Build GRCh38
rs992915984 Chr.19: 9538244-9538244 on Build GRCh38
rs753541147 Chr.19: 9538653-9538653 on Build GRCh38
rs1042809899 Chr.19: 9538492-9538492 on Build GRCh38
rs1017855549 Chr.19: 9538348-9538348 on Build GRCh38
rs764663107 Chr.19: 9538514-9538514 on Build GRCh38

View Full Product Details