Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 9847951-9848453 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PIN1
Entrez Gene ID
5300
Gene Name
peptidylprolyl cis/trans isomerase, NIMA-interacting 1
Gene Aliases
DOD, UBL5
UniGene
Hs.465849
Gene Chromosome Location
Chr.19: 9835207-9849689 on Build GRCh38
Transcripts
NM_006221.3 NR_038422.2 NR_038830.1 XM_011528068.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6746695 c.382+4C>T p.? Chr.19: 9848144-9848144 on Build GRCh38
4083679 c.371C>T p.A124V Chr.19: 9848129-9848129 on Build GRCh38
291606 c.300G>T p.E100D Chr.19: 9848058-9848058 on Build GRCh38
5190421 c.334G>A p.D112N Chr.19: 9848092-9848092 on Build GRCh38
6884107 c.370G>A p.A124T Chr.19: 9848128-9848128 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs574368577 Chr.19: 9848245-9848245 on Build GRCh38
rs775616154 Chr.19: 9848000-9848000 on Build GRCh38
rs556443316 Chr.19: 9848329-9848329 on Build GRCh38
rs879180932 Chr.19: 9848189-9848189 on Build GRCh38
rs760603404 Chr.19: 9847981-9847981 on Build GRCh38
rs774621623 Chr.19: 9848030-9848030 on Build GRCh38
rs779993734 Chr.19: 9848145-9848145 on Build GRCh38
rs768670476 Chr.19: 9848012-9848012 on Build GRCh38
rs752935955 Chr.19: 9847981-9847981 on Build GRCh38
rs2010457 Chr.19: 9848202-9848202 on Build GRCh38
rs751185034 Chr.19: 9848238-9848238 on Build GRCh38
rs1009954048 Chr.19: 9848308-9848308 on Build GRCh38
rs776631037 Chr.19: 9848129-9848129 on Build GRCh38
rs754158170 Chr.19: 9848063-9848063 on Build GRCh38
rs776309023 Chr.19: 9848013-9848013 on Build GRCh38
rs1017432052 Chr.19: 9848295-9848295 on Build GRCh38
rs762132291 Chr.19: 9848208-9848208 on Build GRCh38
rs757649316 Chr.19: 9848085-9848085 on Build GRCh38
rs772234231 Chr.19: 9848176-9848176 on Build GRCh38
rs149978400 Chr.19: 9848276-9848276 on Build GRCh38
rs779792017 Chr.19: 9848225-9848225 on Build GRCh38
rs963537786 Chr.19: 9848326-9848326 on Build GRCh38
rs754356118 Chr.19: 9848207-9848207 on Build GRCh38
rs753183358 Chr.19: 9847988-9847988 on Build GRCh38
rs756874277 Chr.19: 9847994-9847994 on Build GRCh38
rs374690400 Chr.19: 9848014-9848014 on Build GRCh38
rs759664255 Chr.19: 9848035-9848035 on Build GRCh38
rs577766712 Chr.19: 9847980-9847980 on Build GRCh38
rs766579491 Chr.19: 9848015-9848015 on Build GRCh38
rs907464294 Chr.19: 9848092-9848092 on Build GRCh38
rs931307292 Chr.19: 9848229-9848229 on Build GRCh38
rs138056875 Chr.19: 9848128-9848128 on Build GRCh38
rs918606746 Chr.19: 9848222-9848222 on Build GRCh38
rs368178295 Chr.19: 9847982-9847982 on Build GRCh38
rs184605323 Chr.19: 9848114-9848114 on Build GRCh38
rs547618347 Chr.19: 9848332-9848332 on Build GRCh38
rs765604593 Chr.19: 9848212-9848212 on Build GRCh38
rs750773650 Chr.19: 9848213-9848213 on Build GRCh38
rs373349508 Chr.19: 9848146-9848146 on Build GRCh38
rs961847306 Chr.19: 9848406-9848406 on Build GRCh38
rs778487364 Chr.19: 9848076-9848076 on Build GRCh38
rs937586587 Chr.19: 9848265-9848265 on Build GRCh38
rs920010365 Chr.19: 9848336-9848336 on Build GRCh38
rs368788515 Chr.19: 9848009-9848009 on Build GRCh38
rs767555744 Chr.19: 9848044-9848044 on Build GRCh38
rs902892251 Chr.19: 9848228-9848228 on Build GRCh38
rs571699083 Chr.19: 9847987-9847987 on Build GRCh38
rs775561444 Chr.19: 9848191-9848191 on Build GRCh38
rs2287838 Chr.19: 9848338-9848338 on Build GRCh38
rs765299977 Chr.19: 9847980-9847981 on Build GRCh38
rs1041422511 Chr.19: 9848318-9848318 on Build GRCh38
rs758142627 Chr.19: 9848224-9848224 on Build GRCh38
rs1057302882 Chr.19: 9848272-9848272 on Build GRCh38
rs752813 Chr.19: 9848420-9848420 on Build GRCh38
rs745510518 Chr.19: 9848110-9848110 on Build GRCh38
rs758576823 Chr.19: 9848169-9848169 on Build GRCh38
rs374933894 Chr.19: 9848072-9848072 on Build GRCh38
rs777938691 Chr.19: 9848237-9848237 on Build GRCh38
rs1029924709 Chr.19: 9848247-9848247 on Build GRCh38
rs34336739 Chr.19: 9848275-9848275 on Build GRCh38
rs371511137 Chr.19: 9848055-9848055 on Build GRCh38
rs764574279 Chr.19: 9848204-9848204 on Build GRCh38
rs778561537 Chr.19: 9847996-9847996 on Build GRCh38
rs373567733 Chr.19: 9848121-9848121 on Build GRCh38
rs866107708 Chr.19: 9848156-9848156 on Build GRCh38
rs928600399 Chr.19: 9848416-9848416 on Build GRCh38
rs145293152 Chr.19: 9848328-9848328 on Build GRCh38
rs35394313 Chr.19: 9848377-9848377 on Build GRCh38
rs369804664 Chr.19: 9848144-9848144 on Build GRCh38
rs369247974 Chr.19: 9848051-9848051 on Build GRCh38
rs747956175 Chr.19: 9848139-9848139 on Build GRCh38
rs370507014 Chr.19: 9848401-9848401 on Build GRCh38
rs1007414353 Chr.19: 9848282-9848282 on Build GRCh38
rs751852487 Chr.19: 9848052-9848052 on Build GRCh38
rs756513450 Chr.19: 9847989-9847989 on Build GRCh38
rs972781234 Chr.19: 9848214-9848214 on Build GRCh38
rs754501467 Chr.19: 9848239-9848239 on Build GRCh38
rs199701761 Chr.19: 9847986-9847986 on Build GRCh38
rs972831699 Chr.19: 9848421-9848421 on Build GRCh38
rs767964996 Chr.19: 9848125-9848125 on Build GRCh38
rs374690127 Chr.19: 9847998-9847998 on Build GRCh38
rs201472487 Chr.19: 9847992-9847992 on Build GRCh38
rs1009193075 Chr.19: 9848366-9848366 on Build GRCh38
rs776605214 Chr.19: 9848373-9848373 on Build GRCh38

View Full Product Details