Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 10118776-10119277 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EIF3G
Entrez Gene ID
8666
Gene Name
eukaryotic translation initiation factor 3 subunit G
Gene Aliases
EIF3-P42, EIF3S4, eIF3-delta, eIF3-p44
UniGene
Hs.529059
Gene Chromosome Location
Chr.19: 10115014-10119923 on Build GRCh38
Transcripts

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
438454 c.133C>T p.P45S Chr.19: 10119106-10119106 on Build GRCh38
6652850 c.159G>A p.L53L Chr.19: 10118949-10118949 on Build GRCh38
6348486 c.122C>A p.T41N Chr.19: 10119117-10119117 on Build GRCh38
3362625 c.78C>A p.V26V Chr.19: 10119161-10119161 on Build GRCh38
6308754 c.105T>C p.P35P Chr.19: 10119134-10119134 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746990697 Chr.19: 10119090-10119090 on Build GRCh38
rs896964779 Chr.19: 10119029-10119029 on Build GRCh38
rs763048873 Chr.19: 10119044-10119045 on Build GRCh38
rs774908312 Chr.19: 10119055-10119060 on Build GRCh38
rs11558273 Chr.19: 10118943-10118943 on Build GRCh38
rs199771066 Chr.19: 10118861-10118861 on Build GRCh38
rs781026678 Chr.19: 10119098-10119098 on Build GRCh38
rs772032611 Chr.19: 10118854-10118854 on Build GRCh38
rs760467792 Chr.19: 10118859-10118859 on Build GRCh38
rs759671379 Chr.19: 10119021-10119021 on Build GRCh38
rs768139425 Chr.19: 10119069-10119071 on Build GRCh38
rs766400160 Chr.19: 10118823-10118823 on Build GRCh38
rs869110209 Chr.19: 10119050-10119071 on Build GRCh38
rs751058651 Chr.19: 10119218-10119218 on Build GRCh38
rs764599879 Chr.19: 10119039-10119039 on Build GRCh38
rs769413677 Chr.19: 10119114-10119114 on Build GRCh38
rs372087131 Chr.19: 10118877-10118877 on Build GRCh38
rs754978543 Chr.19: 10119221-10119221 on Build GRCh38
rs755089151 Chr.19: 10118888-10118888 on Build GRCh38
rs745674033 Chr.19: 10118818-10118818 on Build GRCh38
rs34658893 Chr.19: 10118904-10118904 on Build GRCh38
rs879295784 Chr.19: 10118858-10118858 on Build GRCh38
rs754240648 Chr.19: 10119041-10119041 on Build GRCh38
rs561775868 Chr.19: 10118929-10118929 on Build GRCh38
rs1002809416 Chr.19: 10119121-10119121 on Build GRCh38
rs779590787 Chr.19: 10119213-10119213 on Build GRCh38
rs371539908 Chr.19: 10118840-10118840 on Build GRCh38
rs765470348 Chr.19: 10119194-10119194 on Build GRCh38
rs774365246 Chr.19: 10119018-10119018 on Build GRCh38
rs942924436 Chr.19: 10119020-10119020 on Build GRCh38
rs199860167 Chr.19: 10118885-10118885 on Build GRCh38
rs777468519 Chr.19: 10118903-10118908 on Build GRCh38
rs1008182740 Chr.19: 10118954-10118954 on Build GRCh38
rs1021837719 Chr.19: 10119211-10119211 on Build GRCh38
rs758486267 Chr.19: 10118820-10118820 on Build GRCh38
rs771174175 Chr.19: 10119014-10119014 on Build GRCh38
rs758243797 Chr.19: 10118853-10118854 on Build GRCh38
rs745378386 Chr.19: 10118928-10118928 on Build GRCh38
rs775214803 Chr.19: 10118855-10118855 on Build GRCh38
rs755499541 Chr.19: 10118893-10118893 on Build GRCh38
rs778631406 Chr.19: 10119044-10119044 on Build GRCh38
rs867633866 Chr.19: 10119154-10119154 on Build GRCh38
rs2290687 Chr.19: 10118845-10118845 on Build GRCh38
rs767442961 Chr.19: 10119026-10119026 on Build GRCh38
rs745668671 Chr.19: 10119231-10119231 on Build GRCh38
rs746782621 Chr.19: 10118916-10118917 on Build GRCh38
rs768562211 Chr.19: 10119093-10119093 on Build GRCh38
rs954385108 Chr.19: 10119247-10119247 on Build GRCh38
rs201202253 Chr.19: 10118889-10118889 on Build GRCh38
rs545097687 Chr.19: 10118854-10118854 on Build GRCh38
rs368688647 Chr.19: 10119184-10119184 on Build GRCh38
rs745841203 Chr.19: 10118852-10118852 on Build GRCh38
rs764693407 Chr.19: 10119168-10119168 on Build GRCh38
rs773446668 Chr.19: 10118863-10118863 on Build GRCh38
rs559673267 Chr.19: 10118930-10118930 on Build GRCh38
rs769704768 Chr.19: 10119037-10119037 on Build GRCh38
rs745952257 Chr.19: 10119022-10119023 on Build GRCh38
rs954457328 Chr.19: 10118990-10118990 on Build GRCh38
rs781104486 Chr.19: 10118976-10118977 on Build GRCh38
rs374614386 Chr.19: 10119076-10119076 on Build GRCh38
rs753284253 Chr.19: 10119028-10119028 on Build GRCh38
rs772230875 Chr.19: 10119128-10119128 on Build GRCh38
rs527406747 Chr.19: 10119032-10119032 on Build GRCh38
rs752039700 Chr.19: 10118829-10118829 on Build GRCh38
rs777396639 Chr.19: 10119253-10119253 on Build GRCh38
rs763786787 Chr.19: 10119209-10119209 on Build GRCh38
rs750578328 Chr.19: 10118813-10118813 on Build GRCh38
rs537407672 Chr.19: 10119136-10119136 on Build GRCh38
rs1038937692 Chr.19: 10119027-10119027 on Build GRCh38
rs756945511 Chr.19: 10118924-10118924 on Build GRCh38
rs762252054 Chr.19: 10119187-10119187 on Build GRCh38
rs11558269 Chr.19: 10118905-10118905 on Build GRCh38
rs369348499 Chr.19: 10118847-10118847 on Build GRCh38
rs778556605 Chr.19: 10118996-10118996 on Build GRCh38
rs755964370 Chr.19: 10119235-10119235 on Build GRCh38
rs1021575003 Chr.19: 10118871-10118871 on Build GRCh38
rs780054301 Chr.19: 10118982-10118982 on Build GRCh38
rs756925651 Chr.19: 10119043-10119043 on Build GRCh38
rs759633424 Chr.19: 10119123-10119123 on Build GRCh38
rs375010356 Chr.19: 10118999-10118999 on Build GRCh38
rs200842732 Chr.19: 10119199-10119199 on Build GRCh38
rs754892821 Chr.19: 10119192-10119194 on Build GRCh38
rs1055923365 Chr.19: 10119180-10119180 on Build GRCh38
rs149991692 Chr.19: 10119051-10119071 on Build GRCh38
rs3217685 Chr.19: 10119078-10119098 on Build GRCh38
rs746901060 Chr.19: 10118991-10118991 on Build GRCh38
rs776380871 Chr.19: 10118994-10118994 on Build GRCh38
rs548775589 Chr.19: 10119047-10119047 on Build GRCh38
rs116747780 Chr.19: 10118831-10118831 on Build GRCh38
rs766160253 Chr.19: 10119123-10119140 on Build GRCh38
rs200314636 Chr.19: 10118833-10118833 on Build GRCh38
rs111350427 Chr.19: 10119174-10119174 on Build GRCh38
rs985907242 Chr.19: 10118998-10118998 on Build GRCh38
rs546753386 Chr.19: 10119225-10119225 on Build GRCh38
rs757949445 Chr.19: 10119048-10119048 on Build GRCh38
rs376680911 Chr.19: 10119212-10119212 on Build GRCh38
rs765440288 Chr.19: 10118812-10118812 on Build GRCh38
rs1011415694 Chr.19: 10119186-10119186 on Build GRCh38
rs777577825 Chr.19: 10118844-10118844 on Build GRCh38
rs768472081 Chr.19: 10118993-10118993 on Build GRCh38
rs778344307 Chr.19: 10118925-10118925 on Build GRCh38
rs922910061 Chr.19: 10118907-10118907 on Build GRCh38
rs140589595 Chr.19: 10119133-10119133 on Build GRCh38
rs113291887 Chr.19: 10119173-10119173 on Build GRCh38
rs748986834 Chr.19: 10118911-10118911 on Build GRCh38
rs759104642 Chr.19: 10119078-10119095 on Build GRCh38
rs770915530 Chr.19: 10118969-10118970 on Build GRCh38
rs538548572 Chr.19: 10119226-10119226 on Build GRCh38
rs11558270 Chr.19: 10119116-10119116 on Build GRCh38
rs764612754 Chr.19: 10119099-10119101 on Build GRCh38
rs201403238 Chr.19: 10119072-10119072 on Build GRCh38
rs775807490 Chr.19: 10119037-10119038 on Build GRCh38
rs907314163 Chr.19: 10118838-10118838 on Build GRCh38
rs148715249 Chr.19: 10118895-10118895 on Build GRCh38
rs778239419 Chr.19: 10118906-10118906 on Build GRCh38
rs772211774 Chr.19: 10118947-10118947 on Build GRCh38
rs747937250 Chr.19: 10119112-10119112 on Build GRCh38
rs762261701 Chr.19: 10119062-10119076 on Build GRCh38
rs773794368 Chr.19: 10119073-10119090 on Build GRCh38

View Full Product Details