Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 10393137-10393640 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDC37
Entrez Gene ID
11140
Gene Name
cell division cycle 37
Gene Aliases
P50CDC37
UniGene
Hs.160958
Gene Chromosome Location
Chr.19: 10391128-10403595 on Build GRCh38
Transcripts
NM_007065.3 XM_011527652.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6624237 c.796C>T p.R266C Chr.19: 10393372-10393372 on Build GRCh38
4073823 c.790C>T p.R264W Chr.19: 10393378-10393378 on Build GRCh38
4073822 c.847C>T p.R283C Chr.19: 10393321-10393321 on Build GRCh38
5854558 c.820G>A p.A274T Chr.19: 10393348-10393348 on Build GRCh38
6624233 c.857G>A p.R286Q Chr.19: 10393311-10393311 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs369472850 Chr.19: 10393237-10393237 on Build GRCh38
rs1008098745 Chr.19: 10393350-10393350 on Build GRCh38
rs776363009 Chr.19: 10393181-10393181 on Build GRCh38
rs145608201 Chr.19: 10393285-10393285 on Build GRCh38
rs779989898 Chr.19: 10393232-10393232 on Build GRCh38
rs749399194 Chr.19: 10393463-10393463 on Build GRCh38
rs760166438 Chr.19: 10393194-10393194 on Build GRCh38
rs199570945 Chr.19: 10393231-10393231 on Build GRCh38
rs760227229 Chr.19: 10393422-10393422 on Build GRCh38
rs767141563 Chr.19: 10393575-10393575 on Build GRCh38
rs770001005 Chr.19: 10393327-10393327 on Build GRCh38
rs542591991 Chr.19: 10393602-10393602 on Build GRCh38
rs892284704 Chr.19: 10393322-10393322 on Build GRCh38
rs775293042 Chr.19: 10393418-10393418 on Build GRCh38
rs373270944 Chr.19: 10393300-10393300 on Build GRCh38
rs376420796 Chr.19: 10393307-10393307 on Build GRCh38
rs779580546 Chr.19: 10393162-10393162 on Build GRCh38
rs780706740 Chr.19: 10393238-10393238 on Build GRCh38
rs529788527 Chr.19: 10393583-10393583 on Build GRCh38
rs774253316 Chr.19: 10393338-10393338 on Build GRCh38
rs747889580 Chr.19: 10393239-10393239 on Build GRCh38
rs149857125 Chr.19: 10393571-10393572 on Build GRCh38
rs776153962 Chr.19: 10393251-10393251 on Build GRCh38
rs774703821 Chr.19: 10393189-10393189 on Build GRCh38
rs28382792 Chr.19: 10393460-10393460 on Build GRCh38
rs538226857 Chr.19: 10393554-10393555 on Build GRCh38
rs774090033 Chr.19: 10393409-10393409 on Build GRCh38
rs28382791 Chr.19: 10393554-10393554 on Build GRCh38
rs987730407 Chr.19: 10393439-10393439 on Build GRCh38
rs547194469 Chr.19: 10393206-10393206 on Build GRCh38
rs781564306 Chr.19: 10393382-10393382 on Build GRCh38
rs767871067 Chr.19: 10393597-10393597 on Build GRCh38
rs753272669 Chr.19: 10393247-10393247 on Build GRCh38
rs771396889 Chr.19: 10393188-10393188 on Build GRCh38
rs2231485 Chr.19: 10393210-10393210 on Build GRCh38
rs781722492 Chr.19: 10393447-10393447 on Build GRCh38
rs1045596195 Chr.19: 10393542-10393542 on Build GRCh38
rs558384153 Chr.19: 10393262-10393262 on Build GRCh38
rs373932777 Chr.19: 10393333-10393333 on Build GRCh38
rs762754328 Chr.19: 10393283-10393283 on Build GRCh38
rs920769899 Chr.19: 10393497-10393497 on Build GRCh38
rs912154284 Chr.19: 10393444-10393444 on Build GRCh38
rs780606412 Chr.19: 10393372-10393372 on Build GRCh38
rs750298355 Chr.19: 10393216-10393216 on Build GRCh38
rs369946518 Chr.19: 10393298-10393298 on Build GRCh38
rs959150302 Chr.19: 10393477-10393477 on Build GRCh38
rs143134175 Chr.19: 10393321-10393321 on Build GRCh38
rs768846872 Chr.19: 10393269-10393269 on Build GRCh38
rs745608862 Chr.19: 10393472-10393472 on Build GRCh38
rs111366742 Chr.19: 10393168-10393168 on Build GRCh38
rs772569186 Chr.19: 10393306-10393306 on Build GRCh38
rs372791909 Chr.19: 10393202-10393202 on Build GRCh38
rs201346787 Chr.19: 10393200-10393200 on Build GRCh38
rs1056362484 Chr.19: 10393592-10393592 on Build GRCh38
rs767381772 Chr.19: 10393354-10393354 on Build GRCh38
rs1007736151 Chr.19: 10393585-10393585 on Build GRCh38
rs763773157 Chr.19: 10393360-10393360 on Build GRCh38
rs751893796 Chr.19: 10393437-10393437 on Build GRCh38
rs140381502 Chr.19: 10393271-10393271 on Build GRCh38
rs771089090 Chr.19: 10393466-10393466 on Build GRCh38
rs946325327 Chr.19: 10393500-10393500 on Build GRCh38
rs148106370 Chr.19: 10393357-10393357 on Build GRCh38
rs200470197 Chr.19: 10393482-10393482 on Build GRCh38
rs769835901 Chr.19: 10393384-10393384 on Build GRCh38
rs542282895 Chr.19: 10393361-10393361 on Build GRCh38
rs924844749 Chr.19: 10393552-10393552 on Build GRCh38
rs138014063 Chr.19: 10393286-10393286 on Build GRCh38
rs149499190 Chr.19: 10393320-10393320 on Build GRCh38
rs761507118 Chr.19: 10393214-10393214 on Build GRCh38
rs748196343 Chr.19: 10393383-10393383 on Build GRCh38
rs771796103 Chr.19: 10393232-10393232 on Build GRCh38
rs748199547 Chr.19: 10393326-10393326 on Build GRCh38
rs764354187 Chr.19: 10393208-10393208 on Build GRCh38
rs528716828 Chr.19: 10393533-10393533 on Build GRCh38
rs377764332 Chr.19: 10393248-10393248 on Build GRCh38
rs374121595 Chr.19: 10393246-10393246 on Build GRCh38
rs573570654 Chr.19: 10393408-10393408 on Build GRCh38
rs117880392 Chr.19: 10393415-10393415 on Build GRCh38
rs879074939 Chr.19: 10393511-10393511 on Build GRCh38
rs372409066 Chr.19: 10393164-10393164 on Build GRCh38
rs890527220 Chr.19: 10393319-10393319 on Build GRCh38
rs1802887 Chr.19: 10393273-10393273 on Build GRCh38
rs755464378 Chr.19: 10393378-10393378 on Build GRCh38
rs926636406 Chr.19: 10393546-10393546 on Build GRCh38
rs755303977 Chr.19: 10393445-10393445 on Build GRCh38
rs955251838 Chr.19: 10393436-10393436 on Build GRCh38
rs751016656 Chr.19: 10393235-10393235 on Build GRCh38
rs746176061 Chr.19: 10393304-10393304 on Build GRCh38
rs781528421 Chr.19: 10393569-10393569 on Build GRCh38
rs749551623 Chr.19: 10393187-10393187 on Build GRCh38
rs771175668 Chr.19: 10393243-10393243 on Build GRCh38
rs748665653 Chr.19: 10393452-10393452 on Build GRCh38
rs1022101507 Chr.19: 10393334-10393334 on Build GRCh38
rs765006356 Chr.19: 10393435-10393435 on Build GRCh38
rs369260677 Chr.19: 10393312-10393312 on Build GRCh38
rs765409570 Chr.19: 10393274-10393274 on Build GRCh38
rs753516808 Chr.19: 10393428-10393428 on Build GRCh38
rs141900325 Chr.19: 10393410-10393410 on Build GRCh38
rs370952004 Chr.19: 10393377-10393377 on Build GRCh38
rs758820884 Chr.19: 10393371-10393371 on Build GRCh38
rs756931215 Chr.19: 10393431-10393431 on Build GRCh38
rs756199715 Chr.19: 10393456-10393456 on Build GRCh38
rs377545975 Chr.19: 10393355-10393355 on Build GRCh38
rs773411962 Chr.19: 10393406-10393406 on Build GRCh38
rs777719156 Chr.19: 10393321-10393322 on Build GRCh38
rs777309634 Chr.19: 10393302-10393302 on Build GRCh38
rs754474320 Chr.19: 10393297-10393297 on Build GRCh38
rs200417274 Chr.19: 10393215-10393215 on Build GRCh38
rs763752940 Chr.19: 10393427-10393427 on Build GRCh38
rs1033460162 Chr.19: 10393170-10393182 on Build GRCh38
rs547152260 Chr.19: 10393534-10393534 on Build GRCh38
rs771829538 Chr.19: 10393480-10393480 on Build GRCh38
rs186570101 Chr.19: 10393540-10393540 on Build GRCh38
rs758291512 Chr.19: 10393230-10393230 on Build GRCh38
rs759555262 Chr.19: 10393351-10393351 on Build GRCh38
rs762736154 Chr.19: 10393329-10393329 on Build GRCh38
rs74316913 Chr.19: 10393249-10393249 on Build GRCh38
rs773430428 Chr.19: 10393328-10393328 on Build GRCh38
rs779116774 Chr.19: 10393468-10393468 on Build GRCh38
rs750170274 Chr.19: 10393282-10393282 on Build GRCh38
rs759077511 Chr.19: 10393475-10393475 on Build GRCh38

View Full Product Details