Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 10559330-10559830 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KRI1
Entrez Gene ID
65095
Gene Name
KRI1 homolog
Gene Aliases
-
UniGene
Hs.709967
Gene Chromosome Location
Chr.19: 10553085-10566031 on Build GRCh38
Transcripts
NM_023008.3 XM_011528190.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1189876 c.1198G>A p.D400N Chr.19: 10559373-10559373 on Build GRCh38
7108887 c.1086G>C p.L362L Chr.19: 10559485-10559485 on Build GRCh38
4073893 c.925G>T p.E309* Chr.19: 10559830-10559830 on Build GRCh38
2812759 c.997C>T p.R333C Chr.19: 10559657-10559657 on Build GRCh38
438493 c.971C>T p.A324V Chr.19: 10559683-10559683 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs578077050 Chr.19: 10559364-10559364 on Build GRCh38
rs766592228 Chr.19: 10559597-10559597 on Build GRCh38
rs771940429 Chr.19: 10559355-10559355 on Build GRCh38
rs754899266 Chr.19: 10559418-10559418 on Build GRCh38
rs11672122 Chr.19: 10559775-10559775 on Build GRCh38
rs765767772 Chr.19: 10559580-10559580 on Build GRCh38
rs746936591 Chr.19: 10559409-10559411 on Build GRCh38
rs751271218 Chr.19: 10559779-10559779 on Build GRCh38
rs376136673 Chr.19: 10559735-10559735 on Build GRCh38
rs945031612 Chr.19: 10559477-10559477 on Build GRCh38
rs113565782 Chr.19: 10559415-10559415 on Build GRCh38
rs537254433 Chr.19: 10559395-10559395 on Build GRCh38
rs903500695 Chr.19: 10559489-10559489 on Build GRCh38
rs556885756 Chr.19: 10559717-10559717 on Build GRCh38
rs753366784 Chr.19: 10559625-10559625 on Build GRCh38
rs769073355 Chr.19: 10559574-10559574 on Build GRCh38
rs757654611 Chr.19: 10559793-10559793 on Build GRCh38
rs770493033 Chr.19: 10559455-10559455 on Build GRCh38
rs374985846 Chr.19: 10559514-10559514 on Build GRCh38
rs773154370 Chr.19: 10559416-10559416 on Build GRCh38
rs367563078 Chr.19: 10559549-10559549 on Build GRCh38
rs781327917 Chr.19: 10559430-10559430 on Build GRCh38
rs369754382 Chr.19: 10559406-10559406 on Build GRCh38
rs748778099 Chr.19: 10559550-10559550 on Build GRCh38
rs769960848 Chr.19: 10559597-10559598 on Build GRCh38
rs199538855 Chr.19: 10559672-10559672 on Build GRCh38
rs143561491 Chr.19: 10559484-10559484 on Build GRCh38
rs778225985 Chr.19: 10559709-10559709 on Build GRCh38
rs745911687 Chr.19: 10559799-10559799 on Build GRCh38
rs779734416 Chr.19: 10559715-10559715 on Build GRCh38
rs774639349 Chr.19: 10559762-10559775 on Build GRCh38
rs925692572 Chr.19: 10559460-10559460 on Build GRCh38
rs778330814 Chr.19: 10559723-10559723 on Build GRCh38
rs763119439 Chr.19: 10559704-10559705 on Build GRCh38
rs371223448 Chr.19: 10559629-10559629 on Build GRCh38
rs773039845 Chr.19: 10559373-10559373 on Build GRCh38
rs781271184 Chr.19: 10559544-10559544 on Build GRCh38
rs138762789 Chr.19: 10559394-10559394 on Build GRCh38
rs879739734 Chr.19: 10559763-10559763 on Build GRCh38
rs1056265166 Chr.19: 10559469-10559469 on Build GRCh38
rs952180967 Chr.19: 10559736-10559736 on Build GRCh38
rs773923897 Chr.19: 10559560-10559560 on Build GRCh38
rs767984624 Chr.19: 10559379-10559379 on Build GRCh38
rs756075403 Chr.19: 10559623-10559623 on Build GRCh38
rs760969040 Chr.19: 10559457-10559457 on Build GRCh38
rs775659870 Chr.19: 10559599-10559599 on Build GRCh38
rs374175757 Chr.19: 10559657-10559657 on Build GRCh38
rs763849847 Chr.19: 10559751-10559751 on Build GRCh38
rs756775160 Chr.19: 10559708-10559708 on Build GRCh38
rs747363297 Chr.19: 10559562-10559562 on Build GRCh38
rs751796174 Chr.19: 10559600-10559600 on Build GRCh38
rs779848502 Chr.19: 10559358-10559358 on Build GRCh38
rs762230706 Chr.19: 10559766-10559786 on Build GRCh38
rs201807816 Chr.19: 10559604-10559604 on Build GRCh38
rs897977665 Chr.19: 10559695-10559695 on Build GRCh38
rs753016551 Chr.19: 10559382-10559382 on Build GRCh38
rs765356427 Chr.19: 10559496-10559496 on Build GRCh38
rs549280035 Chr.19: 10559682-10559682 on Build GRCh38
rs777106973 Chr.19: 10559578-10559578 on Build GRCh38
rs913667010 Chr.19: 10559656-10559656 on Build GRCh38
rs377626522 Chr.19: 10559498-10559498 on Build GRCh38
rs779830814 Chr.19: 10559770-10559771 on Build GRCh38
rs980756059 Chr.19: 10559737-10559737 on Build GRCh38
rs763084423 Chr.19: 10559669-10559669 on Build GRCh38
rs147645270 Chr.19: 10559538-10559538 on Build GRCh38
rs760538152 Chr.19: 10559741-10559741 on Build GRCh38
rs761175461 Chr.19: 10559383-10559383 on Build GRCh38
rs752543948 Chr.19: 10559613-10559613 on Build GRCh38
rs771722962 Chr.19: 10559632-10559632 on Build GRCh38
rs751351398 Chr.19: 10559536-10559536 on Build GRCh38
rs367956318 Chr.19: 10559740-10559740 on Build GRCh38
rs773297445 Chr.19: 10559583-10559583 on Build GRCh38
rs373366565 Chr.19: 10559431-10559431 on Build GRCh38
rs745776507 Chr.19: 10559354-10559354 on Build GRCh38
rs145978093 Chr.19: 10559690-10559690 on Build GRCh38
rs749701088 Chr.19: 10559728-10559728 on Build GRCh38
rs748911745 Chr.19: 10559448-10559448 on Build GRCh38
rs561935531 Chr.19: 10559464-10559464 on Build GRCh38
rs141486227 Chr.19: 10559668-10559668 on Build GRCh38
rs911976109 Chr.19: 10559381-10559381 on Build GRCh38
rs62128851 Chr.19: 10559516-10559516 on Build GRCh38
rs762995789 Chr.19: 10559584-10559584 on Build GRCh38
rs1028331424 Chr.19: 10559390-10559390 on Build GRCh38
rs145288128 Chr.19: 10559374-10559374 on Build GRCh38
rs139920982 Chr.19: 10559676-10559676 on Build GRCh38
rs781498495 Chr.19: 10559641-10559641 on Build GRCh38
rs777874500 Chr.19: 10559624-10559624 on Build GRCh38
rs758564901 Chr.19: 10559413-10559413 on Build GRCh38
rs372519282 Chr.19: 10559773-10559773 on Build GRCh38
rs144086254 Chr.19: 10559684-10559684 on Build GRCh38
rs759709819 Chr.19: 10559739-10559739 on Build GRCh38
rs762244246 Chr.19: 10559579-10559579 on Build GRCh38
rs765411673 Chr.19: 10559401-10559401 on Build GRCh38
rs753782223 Chr.19: 10559705-10559705 on Build GRCh38
rs34743532 Chr.19: 10559811-10559811 on Build GRCh38
rs749982946 Chr.19: 10559714-10559714 on Build GRCh38
rs578261407 Chr.19: 10559720-10559720 on Build GRCh38
rs776642722 Chr.19: 10559578-10559579 on Build GRCh38
rs749697096 Chr.19: 10559787-10559787 on Build GRCh38
rs764539550 Chr.19: 10559389-10559389 on Build GRCh38
rs769109959 Chr.19: 10559745-10559745 on Build GRCh38
rs754640104 Chr.19: 10559780-10559780 on Build GRCh38
rs765245057 Chr.19: 10559774-10559774 on Build GRCh38
rs756753825 Chr.19: 10559626-10559626 on Build GRCh38
rs775883149 Chr.19: 10559456-10559456 on Build GRCh38
rs769497891 Chr.19: 10559433-10559433 on Build GRCh38
rs33999611 Chr.19: 10559630-10559630 on Build GRCh38
rs141095275 Chr.19: 10559367-10559367 on Build GRCh38
rs757947813 Chr.19: 10559777-10559777 on Build GRCh38
rs745948192 Chr.19: 10559585-10559586 on Build GRCh38
rs572532488 Chr.19: 10559410-10559410 on Build GRCh38
rs961304288 Chr.19: 10559404-10559404 on Build GRCh38
rs770838994 Chr.19: 10559459-10559460 on Build GRCh38
rs772345684 Chr.19: 10559810-10559810 on Build GRCh38
rs3826709 Chr.19: 10559508-10559508 on Build GRCh38
rs115325998 Chr.19: 10559471-10559471 on Build GRCh38
rs779522318 Chr.19: 10559798-10559798 on Build GRCh38
rs777935058 Chr.19: 10559784-10559784 on Build GRCh38
rs752490810 Chr.19: 10559683-10559683 on Build GRCh38
rs776495703 Chr.19: 10559753-10559753 on Build GRCh38
rs766019812 Chr.19: 10559778-10559778 on Build GRCh38
rs746859306 Chr.19: 10559447-10559447 on Build GRCh38
rs996962880 Chr.19: 10559581-10559581 on Build GRCh38
rs769549520 Chr.19: 10559371-10559371 on Build GRCh38
rs766536165 Chr.19: 10559518-10559518 on Build GRCh38
rs772986833 Chr.19: 10559437-10559437 on Build GRCh38
rs770267486 Chr.19: 10559665-10559665 on Build GRCh38
rs45518133 Chr.19: 10559675-10559675 on Build GRCh38
rs767365967 Chr.19: 10559605-10559605 on Build GRCh38
rs768485618 Chr.19: 10559363-10559363 on Build GRCh38
rs771474801 Chr.19: 10559733-10559733 on Build GRCh38
rs1049838938 Chr.19: 10559596-10559596 on Build GRCh38

View Full Product Details