Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 10712389-10712884 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
QTRT1
Entrez Gene ID
81890
Gene Name
queuine tRNA-ribosyltransferase catalytic subunit 1
Gene Aliases
FP3235, TGT, TGUT
UniGene
Hs.631638
Gene Chromosome Location
Chr.19: 10701436-10713367 on Build GRCh38
Transcripts
NM_031209.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6004101 c.824G>T p.C275F Chr.19: 10712591-10712591 on Build GRCh38
6761059 c.841G>A p.V281I Chr.19: 10712608-10712608 on Build GRCh38
3528346 c.870C>T p.G290G Chr.19: 10712766-10712766 on Build GRCh38
990677 c.863G>A p.R288H Chr.19: 10712759-10712759 on Build GRCh38
3891475 c.834C>T p.F278F Chr.19: 10712601-10712601 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs973254234 Chr.19: 10712565-10712565 on Build GRCh38
rs890168770 Chr.19: 10712802-10712802 on Build GRCh38
rs777332800 Chr.19: 10712858-10712858 on Build GRCh38
rs751297818 Chr.19: 10712794-10712794 on Build GRCh38
rs767510956 Chr.19: 10712809-10712809 on Build GRCh38
rs202178047 Chr.19: 10712533-10712533 on Build GRCh38
rs377229003 Chr.19: 10712520-10712520 on Build GRCh38
rs914021687 Chr.19: 10712687-10712693 on Build GRCh38
rs780575417 Chr.19: 10712850-10712850 on Build GRCh38
rs757510609 Chr.19: 10712584-10712584 on Build GRCh38
rs1043978769 Chr.19: 10712701-10712701 on Build GRCh38
rs750704961 Chr.19: 10712790-10712790 on Build GRCh38
rs762047406 Chr.19: 10712760-10712760 on Build GRCh38
rs377022672 Chr.19: 10712854-10712854 on Build GRCh38
rs114696057 Chr.19: 10712664-10712664 on Build GRCh38
rs755089184 Chr.19: 10712524-10712525 on Build GRCh38
rs769134436 Chr.19: 10712855-10712855 on Build GRCh38
rs775681114 Chr.19: 10712737-10712737 on Build GRCh38
rs764251932 Chr.19: 10712581-10712581 on Build GRCh38
rs1034444014 Chr.19: 10712700-10712720 on Build GRCh38
rs748872175 Chr.19: 10712732-10712732 on Build GRCh38
rs373619206 Chr.19: 10712541-10712541 on Build GRCh38
rs746933825 Chr.19: 10712608-10712608 on Build GRCh38
rs769184902 Chr.19: 10712726-10712726 on Build GRCh38
rs779400870 Chr.19: 10712727-10712727 on Build GRCh38
rs765216154 Chr.19: 10712787-10712787 on Build GRCh38
rs200166101 Chr.19: 10712859-10712859 on Build GRCh38
rs772314158 Chr.19: 10712630-10712630 on Build GRCh38
rs568267343 Chr.19: 10712607-10712607 on Build GRCh38
rs758269870 Chr.19: 10712601-10712601 on Build GRCh38
rs764305145 Chr.19: 10712658-10712658 on Build GRCh38
rs778591627 Chr.19: 10712586-10712586 on Build GRCh38
rs760014409 Chr.19: 10712628-10712628 on Build GRCh38
rs568561273 Chr.19: 10712782-10712782 on Build GRCh38
rs1034632048 Chr.19: 10712829-10712829 on Build GRCh38
rs756322137 Chr.19: 10712580-10712580 on Build GRCh38
rs868517300 Chr.19: 10712498-10712498 on Build GRCh38
rs768002841 Chr.19: 10712548-10712548 on Build GRCh38
rs761432113 Chr.19: 10712537-10712537 on Build GRCh38
rs745820750 Chr.19: 10712527-10712527 on Build GRCh38
rs777542749 Chr.19: 10712727-10712727 on Build GRCh38
rs371403160 Chr.19: 10712665-10712665 on Build GRCh38
rs761237640 Chr.19: 10712657-10712657 on Build GRCh38
rs920529427 Chr.19: 10712569-10712569 on Build GRCh38
rs755701974 Chr.19: 10712835-10712835 on Build GRCh38
rs756410095 Chr.19: 10712514-10712514 on Build GRCh38
rs769379843 Chr.19: 10712622-10712622 on Build GRCh38
rs776321082 Chr.19: 10712536-10712536 on Build GRCh38
rs750604128 Chr.19: 10712669-10712669 on Build GRCh38
rs767516579 Chr.19: 10712503-10712503 on Build GRCh38
rs756815079 Chr.19: 10712848-10712848 on Build GRCh38
rs779690760 Chr.19: 10712679-10712679 on Build GRCh38
rs747591841 Chr.19: 10712723-10712723 on Build GRCh38
rs765430660 Chr.19: 10712667-10712667 on Build GRCh38
rs754869222 Chr.19: 10712707-10712707 on Build GRCh38
rs776852465 Chr.19: 10712758-10712758 on Build GRCh38
rs751538807 Chr.19: 10712680-10712680 on Build GRCh38
rs985365371 Chr.19: 10712655-10712655 on Build GRCh38
rs866190162 Chr.19: 10712805-10712805 on Build GRCh38
rs557036370 Chr.19: 10712697-10712697 on Build GRCh38
rs534533772 Chr.19: 10712499-10712499 on Build GRCh38
rs758270378 Chr.19: 10712672-10712672 on Build GRCh38
rs558366017 Chr.19: 10712793-10712793 on Build GRCh38
rs749004153 Chr.19: 10712841-10712841 on Build GRCh38
rs752765951 Chr.19: 10712505-10712505 on Build GRCh38
rs149020533 Chr.19: 10712797-10712797 on Build GRCh38
rs745616769 Chr.19: 10712596-10712596 on Build GRCh38
rs954600178 Chr.19: 10712494-10712494 on Build GRCh38
rs749511134 Chr.19: 10712517-10712517 on Build GRCh38
rs774585166 Chr.19: 10712627-10712627 on Build GRCh38
rs752619320 Chr.19: 10712826-10712826 on Build GRCh38
rs771992004 Chr.19: 10712531-10712531 on Build GRCh38
rs548731555 Chr.19: 10712621-10712621 on Build GRCh38
rs370414301 Chr.19: 10712534-10712534 on Build GRCh38
rs757171647 Chr.19: 10712521-10712521 on Build GRCh38
rs373707104 Chr.19: 10712635-10712635 on Build GRCh38
rs761282735 Chr.19: 10712741-10712741 on Build GRCh38
rs778210678 Chr.19: 10712516-10712516 on Build GRCh38
rs147590804 Chr.19: 10712578-10712578 on Build GRCh38
rs780648676 Chr.19: 10712620-10712620 on Build GRCh38
rs368902930 Chr.19: 10712528-10712528 on Build GRCh38
rs115606321 Chr.19: 10712748-10712748 on Build GRCh38
rs759974610 Chr.19: 10712544-10712544 on Build GRCh38
rs933925437 Chr.19: 10712611-10712611 on Build GRCh38
rs777253645 Chr.19: 10712647-10712647 on Build GRCh38
rs768204014 Chr.19: 10712618-10712618 on Build GRCh38
rs766768936 Chr.19: 10712539-10712539 on Build GRCh38
rs778823699 Chr.19: 10712526-10712526 on Build GRCh38
rs374443138 Chr.19: 10712721-10712721 on Build GRCh38
rs777119621 Chr.19: 10712840-10712840 on Build GRCh38

View Full Product Details