Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 10795983-10796481 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNM2
Entrez Gene ID
1785
Gene Name
dynamin 2
Gene Aliases
CMT2M, CMTDI1, CMTDIB, DI-CMTB, DYN2, DYNII, LCCS5
UniGene
Hs.211463
Gene Chromosome Location
Chr.19: 10718053-10831910 on Build GRCh38
Transcripts
NM_001005360.2 NM_001005361.2 NM_001005362.2 NM_001190716.1 NM_004945.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1563968 c.1214C>T p.P405L Chr.19: 10796078-10796078 on Build GRCh38
7350237 c.1222G>C p.A408P Chr.19: 10796086-10796086 on Build GRCh38
990697 c.1264G>T p.E422* Chr.19: 10796128-10796128 on Build GRCh38
5428497 c.1197-8G>A p.? Chr.19: 10796053-10796053 on Build GRCh38
4790408 c.1215G>T p.P405P Chr.19: 10796079-10796079 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757922565 Chr.19: 10796153-10796153 on Build GRCh38
rs756790859 Chr.19: 10796130-10796130 on Build GRCh38
rs776804450 Chr.19: 10796055-10796055 on Build GRCh38
rs944194685 Chr.19: 10796335-10796335 on Build GRCh38
rs775754391 Chr.19: 10796210-10796210 on Build GRCh38
rs779753709 Chr.19: 10796160-10796160 on Build GRCh38
rs377319162 Chr.19: 10796091-10796091 on Build GRCh38
rs764680800 Chr.19: 10796250-10796250 on Build GRCh38
rs542089703 Chr.19: 10796064-10796064 on Build GRCh38
rs563985581 Chr.19: 10796082-10796082 on Build GRCh38
rs908416851 Chr.19: 10796147-10796147 on Build GRCh38
rs536433645 Chr.19: 10796291-10796291 on Build GRCh38
rs565010494 Chr.19: 10796242-10796242 on Build GRCh38
rs774748942 Chr.19: 10796079-10796079 on Build GRCh38
rs908918670 Chr.19: 10796390-10796390 on Build GRCh38
rs918828316 Chr.19: 10796364-10796364 on Build GRCh38
rs554498470 Chr.19: 10796324-10796324 on Build GRCh38
rs1047385997 Chr.19: 10796379-10796379 on Build GRCh38
rs866965396 Chr.19: 10796108-10796108 on Build GRCh38
rs186762327 Chr.19: 10796150-10796150 on Build GRCh38
rs767631539 Chr.19: 10796426-10796426 on Build GRCh38
rs770176751 Chr.19: 10796173-10796173 on Build GRCh38
rs755921154 Chr.19: 10796109-10796109 on Build GRCh38
rs749775962 Chr.19: 10796175-10796175 on Build GRCh38
rs773756665 Chr.19: 10796065-10796065 on Build GRCh38
rs753794492 Chr.19: 10796218-10796218 on Build GRCh38
rs757912925 Chr.19: 10796249-10796249 on Build GRCh38
rs750003938 Chr.19: 10796246-10796246 on Build GRCh38
rs771160197 Chr.19: 10796181-10796181 on Build GRCh38
rs1047420749 Chr.19: 10796125-10796125 on Build GRCh38
rs772236702 Chr.19: 10796208-10796208 on Build GRCh38
rs954948728 Chr.19: 10796279-10796279 on Build GRCh38
rs975118669 Chr.19: 10796450-10796450 on Build GRCh38
rs568585837 Chr.19: 10796275-10796275 on Build GRCh38
rs879253980 Chr.19: 10796137-10796137 on Build GRCh38
rs569931989 Chr.19: 10796411-10796411 on Build GRCh38
rs894945448 Chr.19: 10796241-10796241 on Build GRCh38
rs374376619 Chr.19: 10796067-10796067 on Build GRCh38
rs369716035 Chr.19: 10796386-10796386 on Build GRCh38
rs748695148 Chr.19: 10796058-10796058 on Build GRCh38
rs931176769 Chr.19: 10796120-10796120 on Build GRCh38
rs977336614 Chr.19: 10796340-10796340 on Build GRCh38
rs1005967170 Chr.19: 10796219-10796219 on Build GRCh38
rs941708273 Chr.19: 10796425-10796425 on Build GRCh38
rs1009575648 Chr.19: 10796400-10796400 on Build GRCh38
rs759313573 Chr.19: 10796013-10796013 on Build GRCh38
rs958100245 Chr.19: 10796314-10796314 on Build GRCh38
rs778187114 Chr.19: 10796174-10796174 on Build GRCh38
rs745481476 Chr.19: 10796042-10796042 on Build GRCh38
rs753743444 Chr.19: 10796128-10796128 on Build GRCh38
rs911296278 Chr.19: 10796334-10796334 on Build GRCh38
rs763986985 Chr.19: 10796116-10796116 on Build GRCh38
rs530195152 Chr.19: 10796052-10796052 on Build GRCh38
rs778575153 Chr.19: 10796038-10796038 on Build GRCh38
rs904686742 Chr.19: 10796258-10796258 on Build GRCh38
rs757729053 Chr.19: 10796424-10796424 on Build GRCh38
rs371241718 Chr.19: 10796187-10796187 on Build GRCh38
rs150975494 Chr.19: 10796212-10796212 on Build GRCh38
rs930253624 Chr.19: 10796368-10796368 on Build GRCh38
rs140208362 Chr.19: 10796149-10796149 on Build GRCh38
rs111845365 Chr.19: 10796018-10796018 on Build GRCh38
rs752497841 Chr.19: 10796094-10796094 on Build GRCh38
rs79068463 Chr.19: 10796440-10796440 on Build GRCh38
rs748520274 Chr.19: 10796162-10796162 on Build GRCh38
rs759681218 Chr.19: 10796203-10796203 on Build GRCh38
rs45478694 Chr.19: 10796274-10796274 on Build GRCh38
rs1032383765 Chr.19: 10796298-10796298 on Build GRCh38
rs763908447 Chr.19: 10796217-10796217 on Build GRCh38
rs199544971 Chr.19: 10796231-10796231 on Build GRCh38
rs115439545 Chr.19: 10796115-10796115 on Build GRCh38
rs867141113 Chr.19: 10796454-10796454 on Build GRCh38
rs1013841748 Chr.19: 10796261-10796261 on Build GRCh38
rs536988136 Chr.19: 10796427-10796427 on Build GRCh38

View Full Product Details