Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 10985182-10985671 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SMARCA4
Entrez Gene ID
6597
Gene Name
SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily a, member 4
Gene Aliases
BAF190, BAF190A, BRG1, CSS4, MRD16, RTPS2, SNF2, SNF2L4, SNF2LB, SWI2, hSNF2b
UniGene
Hs.327527
Gene Chromosome Location
Chr.19: 10960922-11062282 on Build GRCh38
Transcripts
NM_001128844.1 NM_001128845.1 NM_001128846.1 NM_001128847.1 NM_001128848.1 NM_001128849.1 NM_003072.3 XM_006722845.2 XM_006722846.2 XM_011528198.1 XM_017027160.1 XM_017027161.1 XM_017027162.1 XM_017027163.1 XM_017027164.1 XM_017027165.1 XM_017027166.1 XM_017027167.1 XM_017027168.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7325667 c.246delG p.G83fs*11 Chr.19: 10985296-10985296 on Build GRCh38
6918267 c.301delG p.G102fs*201 Chr.19: 10985351-10985351 on Build GRCh38
3735029 c.233_237CCATG>ACC p.S78fs*3 Chr.19: 10985283-10985287 on Build GRCh38
438515 c.254C>T p.S85L Chr.19: 10985304-10985304 on Build GRCh38
6910873 c.350C>T p.S117F Chr.19: 10985400-10985400 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs999267729 Chr.19: 10985410-10985410 on Build GRCh38
rs947610630 Chr.19: 10985624-10985624 on Build GRCh38
rs1043445785 Chr.19: 10985633-10985633 on Build GRCh38
rs376775933 Chr.19: 10985268-10985268 on Build GRCh38
rs771605253 Chr.19: 10985347-10985347 on Build GRCh38
rs566882838 Chr.19: 10985339-10985339 on Build GRCh38
rs768767515 Chr.19: 10985522-10985522 on Build GRCh38
rs898823360 Chr.19: 10985569-10985569 on Build GRCh38
rs878854209 Chr.19: 10985296-10985296 on Build GRCh38
rs551739868 Chr.19: 10985317-10985317 on Build GRCh38
rs535379695 Chr.19: 10985621-10985621 on Build GRCh38
rs1056057044 Chr.19: 10985340-10985340 on Build GRCh38
rs745559947 Chr.19: 10985249-10985249 on Build GRCh38
rs879075470 Chr.19: 10985207-10985207 on Build GRCh38
rs1012041090 Chr.19: 10985587-10985587 on Build GRCh38
rs767638396 Chr.19: 10985285-10985285 on Build GRCh38
rs917024469 Chr.19: 10985521-10985521 on Build GRCh38
rs866561294 Chr.19: 10985212-10985212 on Build GRCh38
rs757856874 Chr.19: 10985432-10985432 on Build GRCh38
rs779627018 Chr.19: 10985354-10985354 on Build GRCh38
rs569296824 Chr.19: 10985264-10985264 on Build GRCh38
rs757214683 Chr.19: 10985344-10985344 on Build GRCh38
rs763471007 Chr.19: 10985376-10985376 on Build GRCh38
rs750442110 Chr.19: 10985426-10985426 on Build GRCh38
rs778529668 Chr.19: 10985243-10985243 on Build GRCh38
rs778779846 Chr.19: 10985430-10985430 on Build GRCh38
rs930098220 Chr.19: 10985535-10985535 on Build GRCh38
rs138209034 Chr.19: 10985471-10985471 on Build GRCh38
rs148835854 Chr.19: 10985224-10985224 on Build GRCh38
rs528166922 Chr.19: 10985211-10985211 on Build GRCh38
rs1005789011 Chr.19: 10985548-10985548 on Build GRCh38
rs754574700 Chr.19: 10985457-10985457 on Build GRCh38
rs918577444 Chr.19: 10985534-10985534 on Build GRCh38
rs183120332 Chr.19: 10985454-10985454 on Build GRCh38
rs745406632 Chr.19: 10985346-10985346 on Build GRCh38
rs763383326 Chr.19: 10985269-10985269 on Build GRCh38
rs543401767 Chr.19: 10985218-10985218 on Build GRCh38
rs766388110 Chr.19: 10985223-10985223 on Build GRCh38
rs901378371 Chr.19: 10985496-10985496 on Build GRCh38
rs370116117 Chr.19: 10985201-10985201 on Build GRCh38
rs778890805 Chr.19: 10985345-10985345 on Build GRCh38
rs767575401 Chr.19: 10985414-10985414 on Build GRCh38
rs746602808 Chr.19: 10985355-10985355 on Build GRCh38
rs752529336 Chr.19: 10985588-10985588 on Build GRCh38
rs770147215 Chr.19: 10985358-10985358 on Build GRCh38
rs759594383 Chr.19: 10985400-10985400 on Build GRCh38
rs371056395 Chr.19: 10985313-10985313 on Build GRCh38
rs766584863 Chr.19: 10985557-10985557 on Build GRCh38
rs78732262 Chr.19: 10985220-10985220 on Build GRCh38
rs966478275 Chr.19: 10985401-10985401 on Build GRCh38
rs977943456 Chr.19: 10985208-10985208 on Build GRCh38
rs781061055 Chr.19: 10985230-10985230 on Build GRCh38
rs10426670 Chr.19: 10985231-10985231 on Build GRCh38
rs751542188 Chr.19: 10985304-10985304 on Build GRCh38
rs777917127 Chr.19: 10985234-10985234 on Build GRCh38
rs780424104 Chr.19: 10985350-10985351 on Build GRCh38
rs998875986 Chr.19: 10985301-10985301 on Build GRCh38
rs915226562 Chr.19: 10985465-10985465 on Build GRCh38
rs764006928 Chr.19: 10985418-10985418 on Build GRCh38
rs989345000 Chr.19: 10985651-10985651 on Build GRCh38
rs1039757191 Chr.19: 10985478-10985478 on Build GRCh38
rs755987663 Chr.19: 10985321-10985321 on Build GRCh38
rs753861666 Chr.19: 10985341-10985341 on Build GRCh38
rs770573833 Chr.19: 10985239-10985239 on Build GRCh38
rs935214074 Chr.19: 10985453-10985453 on Build GRCh38
rs1021947977 Chr.19: 10985377-10985377 on Build GRCh38
rs200506869 Chr.19: 10985650-10985650 on Build GRCh38
rs970848537 Chr.19: 10985601-10985601 on Build GRCh38
rs115992445 Chr.19: 10985308-10985308 on Build GRCh38
rs371740591 Chr.19: 10985428-10985428 on Build GRCh38
rs749179837 Chr.19: 10985238-10985238 on Build GRCh38
rs762146990 Chr.19: 10985266-10985266 on Build GRCh38
rs908964467 Chr.19: 10985214-10985214 on Build GRCh38
rs370834603 Chr.19: 10985558-10985558 on Build GRCh38
rs1053682643 Chr.19: 10985246-10985246 on Build GRCh38
rs369657349 Chr.19: 10985227-10985227 on Build GRCh38
rs766474711 Chr.19: 10985297-10985297 on Build GRCh38
rs756404222 Chr.19: 10985255-10985255 on Build GRCh38
rs878854207 Chr.19: 10985282-10985282 on Build GRCh38
rs878854211 Chr.19: 10985305-10985305 on Build GRCh38
rs948571406 Chr.19: 10985568-10985568 on Build GRCh38
rs367559956 Chr.19: 10985417-10985417 on Build GRCh38
rs373917129 Chr.19: 10985636-10985636 on Build GRCh38
rs368761916 Chr.19: 10985416-10985416 on Build GRCh38
rs928685957 Chr.19: 10985565-10985565 on Build GRCh38
rs755971216 Chr.19: 10985232-10985232 on Build GRCh38
rs374573270 Chr.19: 10985251-10985251 on Build GRCh38
rs773526113 Chr.19: 10985366-10985366 on Build GRCh38
rs777647740 Chr.19: 10985327-10985327 on Build GRCh38
rs374224953 Chr.19: 10985380-10985380 on Build GRCh38
rs1056342044 Chr.19: 10985614-10985614 on Build GRCh38
rs774919609 Chr.19: 10985387-10985387 on Build GRCh38

View Full Product Details