Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 11376648-11377195 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SWSAP1
Entrez Gene ID
126074
Gene Name
SWIM-type zinc finger 7 associated protein 1
Gene Aliases
C19orf39, SWS1AP1, ZSWIM7AP1
UniGene
Hs.631619
Gene Chromosome Location
Chr.19: 11374707-11376951 on Build GRCh38
Transcripts
NM_175871.3

SNP Detail

SNP ID Chromosome Location
rs914585282 Chr.19: 11377143-11377143 on Build GRCh38
rs1036334438 Chr.19: 11377057-11377057 on Build GRCh38
rs867088719 Chr.19: 11376966-11376966 on Build GRCh38
rs982096905 Chr.19: 11376716-11376716 on Build GRCh38
rs940974028 Chr.19: 11377145-11377145 on Build GRCh38
rs1003071534 Chr.19: 11377049-11377049 on Build GRCh38
rs965152984 Chr.19: 11377113-11377113 on Build GRCh38
rs71164194 Chr.19: 11376971-11376972 on Build GRCh38
rs1038086443 Chr.19: 11376938-11376938 on Build GRCh38
rs113239777 Chr.19: 11377102-11377102 on Build GRCh38
rs951621365 Chr.19: 11377038-11377038 on Build GRCh38
rs941024957 Chr.19: 11376917-11376917 on Build GRCh38
rs991510970 Chr.19: 11376760-11376760 on Build GRCh38
rs947466177 Chr.19: 11376780-11376780 on Build GRCh38
rs149544777 Chr.19: 11376784-11376784 on Build GRCh38
rs995217756 Chr.19: 11377032-11377032 on Build GRCh38
rs935219574 Chr.19: 11376875-11376875 on Build GRCh38
rs997974055 Chr.19: 11377088-11377088 on Build GRCh38
rs771085869 Chr.19: 11377045-11377045 on Build GRCh38
rs1003633138 Chr.19: 11377026-11377044 on Build GRCh38
rs764503817 Chr.19: 11376805-11376805 on Build GRCh38
rs957328161 Chr.19: 11377073-11377073 on Build GRCh38
rs958943183 Chr.19: 11376748-11376748 on Build GRCh38
rs955207312 Chr.19: 11377125-11377125 on Build GRCh38
rs180975926 Chr.19: 11377064-11377064 on Build GRCh38
rs977683107 Chr.19: 11376700-11376700 on Build GRCh38
rs1011539443 Chr.19: 11377080-11377080 on Build GRCh38
rs1024891747 Chr.19: 11376984-11376984 on Build GRCh38
rs143035464 Chr.19: 11376695-11376695 on Build GRCh38
rs977502532 Chr.19: 11377165-11377165 on Build GRCh38
rs968751073 Chr.19: 11377147-11377147 on Build GRCh38
rs902303462 Chr.19: 11376986-11376986 on Build GRCh38
rs112805342 Chr.19: 11377101-11377101 on Build GRCh38
rs923773348 Chr.19: 11376873-11376873 on Build GRCh38
rs1025385763 Chr.19: 11377036-11377036 on Build GRCh38
rs951621924 Chr.19: 11376696-11376696 on Build GRCh38
rs976214223 Chr.19: 11377120-11377121 on Build GRCh38
rs922498151 Chr.19: 11376859-11376859 on Build GRCh38
rs1047043737 Chr.19: 11376897-11376897 on Build GRCh38
rs922412967 Chr.19: 11377121-11377123 on Build GRCh38
rs894224616 Chr.19: 11376958-11376959 on Build GRCh38
rs548327293 Chr.19: 11376876-11376876 on Build GRCh38
rs908463243 Chr.19: 11377135-11377135 on Build GRCh38
rs138468247 Chr.19: 11376675-11376675 on Build GRCh38
rs317927 Chr.19: 11377110-11377110 on Build GRCh38
rs891076090 Chr.19: 11376900-11376900 on Build GRCh38
rs398039171 Chr.19: 11376970-11376971 on Build GRCh38
rs749454029 Chr.19: 11376733-11376733 on Build GRCh38
rs1012703905 Chr.19: 11376960-11376961 on Build GRCh38
rs914636129 Chr.19: 11376767-11376767 on Build GRCh38
rs975642613 Chr.19: 11376787-11376787 on Build GRCh38
rs1031902542 Chr.19: 11377065-11377065 on Build GRCh38
rs370502021 Chr.19: 11377046-11377046 on Build GRCh38
rs191025147 Chr.19: 11376939-11376939 on Build GRCh38
rs1043545166 Chr.19: 11376823-11376823 on Build GRCh38
rs35480777 Chr.19: 11376951-11376952 on Build GRCh38
rs982674905 Chr.19: 11377131-11377131 on Build GRCh38
rs186567827 Chr.19: 11376910-11376910 on Build GRCh38

View Full Product Details