Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 33300222-33300720 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CEBPA
Entrez Gene ID
1050
Gene Name
CCAAT/enhancer binding protein alpha
Gene Aliases
C/EBP-alpha, CEBP
UniGene
Hs.76171
Gene Chromosome Location
Chr.19: 33299934-33302564 on Build GRCh38
Transcripts
NM_001285829.1 NM_001287424.1 NM_001287435.1 NM_004364.4

SNP Detail

SNP ID Chromosome Location
rs976861369 Chr.19: 33300472-33300473 on Build GRCh38
rs564155517 Chr.19: 33300570-33300570 on Build GRCh38
rs906813267 Chr.19: 33300330-33300330 on Build GRCh38
rs967287145 Chr.19: 33300457-33300457 on Build GRCh38
rs906743002 Chr.19: 33300616-33300616 on Build GRCh38
rs879131240 Chr.19: 33300335-33300335 on Build GRCh38
rs563625019 Chr.19: 33300606-33300606 on Build GRCh38
rs1001741074 Chr.19: 33300372-33300372 on Build GRCh38
rs533138144 Chr.19: 33300582-33300582 on Build GRCh38
rs945868668 Chr.19: 33300249-33300249 on Build GRCh38
rs755230057 Chr.19: 33300277-33300277 on Build GRCh38
rs528278380 Chr.19: 33300501-33300501 on Build GRCh38
rs933004923 Chr.19: 33300313-33300313 on Build GRCh38
rs952738654 Chr.19: 33300378-33300378 on Build GRCh38
rs566231195 Chr.19: 33300276-33300276 on Build GRCh38
rs1057012390 Chr.19: 33300589-33300589 on Build GRCh38
rs935757154 Chr.19: 33300286-33300286 on Build GRCh38
rs535297087 Chr.19: 33300291-33300291 on Build GRCh38
rs748271362 Chr.19: 33300341-33300341 on Build GRCh38
rs1018374833 Chr.19: 33300677-33300677 on Build GRCh38
rs549322947 Chr.19: 33300630-33300630 on Build GRCh38
rs550273098 Chr.19: 33300598-33300598 on Build GRCh38
rs544875977 Chr.19: 33300553-33300553 on Build GRCh38
rs886090564 Chr.19: 33300331-33300331 on Build GRCh38
rs1020213190 Chr.19: 33300635-33300635 on Build GRCh38
rs707656 Chr.19: 33300623-33300623 on Build GRCh38
rs796685679 Chr.19: 33300450-33300450 on Build GRCh38
rs572803705 Chr.19: 33300531-33300531 on Build GRCh38
rs1000572363 Chr.19: 33300673-33300673 on Build GRCh38
rs964057493 Chr.19: 33300439-33300439 on Build GRCh38
rs964497806 Chr.19: 33300692-33300692 on Build GRCh38
rs1047293885 Chr.19: 33300511-33300511 on Build GRCh38
rs146104564 Chr.19: 33300333-33300333 on Build GRCh38
rs915669651 Chr.19: 33300516-33300516 on Build GRCh38
rs911260475 Chr.19: 33300464-33300470 on Build GRCh38
rs376667577 Chr.19: 33300514-33300514 on Build GRCh38
rs34017519 Chr.19: 33300305-33300305 on Build GRCh38
rs556765660 Chr.19: 33300458-33300458 on Build GRCh38
rs879006824 Chr.19: 33300375-33300375 on Build GRCh38
rs771978125 Chr.19: 33300576-33300576 on Build GRCh38
rs905698271 Chr.19: 33300359-33300359 on Build GRCh38
rs941652275 Chr.19: 33300528-33300529 on Build GRCh38
rs968753228 Chr.19: 33300672-33300672 on Build GRCh38
rs977187209 Chr.19: 33300463-33300463 on Build GRCh38
rs894134181 Chr.19: 33300302-33300302 on Build GRCh38
rs113670631 Chr.19: 33300252-33300252 on Build GRCh38
rs573809108 Chr.19: 33300471-33300471 on Build GRCh38
rs1005626115 Chr.19: 33300393-33300393 on Build GRCh38
rs922492617 Chr.19: 33300449-33300449 on Build GRCh38
rs761766986 Chr.19: 33300288-33300288 on Build GRCh38
rs1001388715 Chr.19: 33300534-33300534 on Build GRCh38
rs935869272 Chr.19: 33300493-33300493 on Build GRCh38
rs925799275 Chr.19: 33300472-33300472 on Build GRCh38
rs867364161 Chr.19: 33300446-33300446 on Build GRCh38
rs1046626496 Chr.19: 33300326-33300326 on Build GRCh38
rs959922601 Chr.19: 33300403-33300403 on Build GRCh38
rs187516157 Chr.19: 33300344-33300344 on Build GRCh38
rs1037463589 Chr.19: 33300529-33300529 on Build GRCh38
rs769951978 Chr.19: 33300369-33300369 on Build GRCh38
rs907570376 Chr.19: 33300524-33300524 on Build GRCh38
rs562038920 Chr.19: 33300435-33300435 on Build GRCh38
rs905579664 Chr.19: 33300532-33300532 on Build GRCh38
rs748249393 Chr.19: 33300681-33300681 on Build GRCh38

View Full Product Details