Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 33301927-33302445 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CEBPA
Entrez Gene ID
1050
Gene Name
CCAAT/enhancer binding protein alpha
Gene Aliases
C/EBP-alpha, CEBP
UniGene
Hs.76171
Gene Chromosome Location
Chr.19: 33299934-33302564 on Build GRCh38
Transcripts
NM_001285829.1 NM_001287424.1 NM_001287435.1 NM_004364.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
29231 c.91_92insGCCGCCT p.F31fs*79 Chr.19: 33302323-33302324 on Build GRCh38
29267 c.418_419insGTTCG p.D140fs*22 Chr.19: 33301996-33301997 on Build GRCh38
27476 c.392_393insA p.Y131fs*1 Chr.19: 33302022-33302023 on Build GRCh38
5944700 c.202_203insCTAC p.I68fs*41 Chr.19: 33302212-33302213 on Build GRCh38
20919 c.111_121del11 p.G38fs*66 Chr.19: 33302294-33302304 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs986682061 Chr.19: 33301964-33301964 on Build GRCh38
rs902667124 Chr.19: 33302214-33302214 on Build GRCh38
rs892805896 Chr.19: 33302273-33302273 on Build GRCh38
rs1001507122 Chr.19: 33302070-33302070 on Build GRCh38
rs961496947 Chr.19: 33302378-33302378 on Build GRCh38
rs1042672907 Chr.19: 33302052-33302052 on Build GRCh38
rs587776849 Chr.19: 33302204-33302205 on Build GRCh38
rs770636941 Chr.19: 33302351-33302351 on Build GRCh38
rs575182822 Chr.19: 33302350-33302350 on Build GRCh38
rs1054442664 Chr.19: 33302262-33302262 on Build GRCh38
rs137852733 Chr.19: 33302197-33302198 on Build GRCh38
rs919904139 Chr.19: 33302027-33302027 on Build GRCh38
rs878854699 Chr.19: 33302241-33302241 on Build GRCh38
rs1038352346 Chr.19: 33302021-33302021 on Build GRCh38
rs1007915253 Chr.19: 33302374-33302374 on Build GRCh38
rs878854700 Chr.19: 33302163-33302163 on Build GRCh38
rs111662895 Chr.19: 33302334-33302334 on Build GRCh38
rs890855027 Chr.19: 33302353-33302353 on Build GRCh38
rs1042601332 Chr.19: 33302198-33302198 on Build GRCh38
rs896656951 Chr.19: 33302042-33302042 on Build GRCh38
rs777090929 Chr.19: 33302255-33302255 on Build GRCh38
rs878854701 Chr.19: 33302137-33302138 on Build GRCh38
rs137852732 Chr.19: 33302095-33302096 on Build GRCh38
rs957253046 Chr.19: 33302081-33302081 on Build GRCh38
rs908433906 Chr.19: 33302002-33302002 on Build GRCh38
rs137852731 Chr.19: 33302213-33302214 on Build GRCh38
rs917977456 Chr.19: 33302169-33302169 on Build GRCh38
rs137852730 Chr.19: 33302274-33302274 on Build GRCh38
rs137852729 Chr.19: 33302346-33302347 on Build GRCh38
rs28931590 Chr.19: 33302164-33302164 on Build GRCh38
rs780345232 Chr.19: 33302101-33302102 on Build GRCh38
rs878854702 Chr.19: 33302032-33302032 on Build GRCh38
rs938139789 Chr.19: 33302031-33302031 on Build GRCh38
rs868242502 Chr.19: 33302154-33302154 on Build GRCh38
rs747773004 Chr.19: 33302148-33302150 on Build GRCh38
rs1032707295 Chr.19: 33302076-33302076 on Build GRCh38
rs746522150 Chr.19: 33302308-33302308 on Build GRCh38
rs121912791 Chr.19: 33302267-33302267 on Build GRCh38
rs137852728 Chr.19: 33302347-33302347 on Build GRCh38
rs936973108 Chr.19: 33302260-33302260 on Build GRCh38
rs996591962 Chr.19: 33302084-33302084 on Build GRCh38
rs996435066 Chr.19: 33302315-33302315 on Build GRCh38
rs867113214 Chr.19: 33302352-33302352 on Build GRCh38
rs961343504 Chr.19: 33302102-33302110 on Build GRCh38
rs974386262 Chr.19: 33302104-33302104 on Build GRCh38
rs921129497 Chr.19: 33301947-33301947 on Build GRCh38
rs1028043587 Chr.19: 33302349-33302349 on Build GRCh38
rs779373951 Chr.19: 33301956-33301956 on Build GRCh38
rs1027991203 Chr.19: 33302090-33302090 on Build GRCh38
rs758092761 Chr.19: 33302138-33302138 on Build GRCh38
rs919975422 Chr.19: 33302127-33302127 on Build GRCh38
rs867268085 Chr.19: 33302404-33302404 on Build GRCh38
rs949548349 Chr.19: 33302190-33302190 on Build GRCh38
rs1013724730 Chr.19: 33302051-33302051 on Build GRCh38
rs759902033 Chr.19: 33302385-33302385 on Build GRCh38
rs902740061 Chr.19: 33302060-33302060 on Build GRCh38
rs1055210725 Chr.19: 33302036-33302036 on Build GRCh38
rs1015459376 Chr.19: 33302376-33302376 on Build GRCh38
rs558462037 Chr.19: 33302231-33302231 on Build GRCh38
rs752254340 Chr.19: 33302013-33302013 on Build GRCh38

View Full Product Details