Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 35034116-35034619 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SCN1B
Entrez Gene ID
6324
Gene Name
sodium voltage-gated channel beta subunit 1
Gene Aliases
ATFB13, BRGDA5, GEFSP1
UniGene
Hs.436646
Gene Chromosome Location
Chr.19: 35030688-35040449 on Build GRCh38
Transcripts
NM_001037.4 NM_001321605.1 NM_199037.4

SNP Detail

SNP ID Chromosome Location
rs928460945 Chr.19: 35034465-35034465 on Build GRCh38
rs528306248 Chr.19: 35034544-35034544 on Build GRCh38
rs966191447 Chr.19: 35034561-35034561 on Build GRCh38
rs551030996 Chr.19: 35034545-35034545 on Build GRCh38
rs898068226 Chr.19: 35034191-35034191 on Build GRCh38
rs908876567 Chr.19: 35034434-35034434 on Build GRCh38
rs907066969 Chr.19: 35034249-35034249 on Build GRCh38
rs991312626 Chr.19: 35034399-35034399 on Build GRCh38
rs777168533 Chr.19: 35034251-35034251 on Build GRCh38
rs1040605501 Chr.19: 35034535-35034535 on Build GRCh38
rs375266320 Chr.19: 35034183-35034183 on Build GRCh38
rs67777826 Chr.19: 35034349-35034349 on Build GRCh38
rs975815640 Chr.19: 35034364-35034364 on Build GRCh38
rs919046606 Chr.19: 35034390-35034390 on Build GRCh38
rs968295248 Chr.19: 35034320-35034320 on Build GRCh38
rs949867079 Chr.19: 35034411-35034411 on Build GRCh38
rs767745946 Chr.19: 35034398-35034398 on Build GRCh38
rs966187816 Chr.19: 35034292-35034292 on Build GRCh38
rs968461461 Chr.19: 35034327-35034327 on Build GRCh38
rs769718318 Chr.19: 35034512-35034512 on Build GRCh38
rs895595793 Chr.19: 35034150-35034150 on Build GRCh38
rs529181068 Chr.19: 35034452-35034452 on Build GRCh38
rs910726120 Chr.19: 35034149-35034149 on Build GRCh38
rs571035118 Chr.19: 35034596-35034596 on Build GRCh38
rs61377791 Chr.19: 35034418-35034418 on Build GRCh38
rs181577717 Chr.19: 35034521-35034521 on Build GRCh38
rs909167747 Chr.19: 35034214-35034214 on Build GRCh38
rs766429780 Chr.19: 35034584-35034584 on Build GRCh38
rs948633052 Chr.19: 35034443-35034443 on Build GRCh38
rs903870552 Chr.19: 35034541-35034541 on Build GRCh38
rs775492616 Chr.19: 35034268-35034268 on Build GRCh38
rs541514361 Chr.19: 35034274-35034274 on Build GRCh38
rs1022721631 Chr.19: 35034309-35034309 on Build GRCh38
rs930317818 Chr.19: 35034486-35034486 on Build GRCh38
rs148521036 Chr.19: 35034344-35034344 on Build GRCh38
rs950430191 Chr.19: 35034400-35034400 on Build GRCh38
rs763850487 Chr.19: 35034277-35034277 on Build GRCh38
rs192082982 Chr.19: 35034298-35034298 on Build GRCh38
rs750416956 Chr.19: 35034140-35034140 on Build GRCh38
rs745394799 Chr.19: 35034182-35034182 on Build GRCh38
rs1044171971 Chr.19: 35034248-35034248 on Build GRCh38
rs771029432 Chr.19: 35034326-35034326 on Build GRCh38
rs769549392 Chr.19: 35034226-35034226 on Build GRCh38
rs997722669 Chr.19: 35034574-35034574 on Build GRCh38
rs948655385 Chr.19: 35034225-35034225 on Build GRCh38
rs1021471359 Chr.19: 35034582-35034582 on Build GRCh38
rs1049318381 Chr.19: 35034491-35034491 on Build GRCh38
rs1003567394 Chr.19: 35034250-35034250 on Build GRCh38
rs897453912 Chr.19: 35034450-35034450 on Build GRCh38
rs918914134 Chr.19: 35034190-35034190 on Build GRCh38
rs572726115 Chr.19: 35034192-35034192 on Build GRCh38
rs375165496 Chr.19: 35034444-35034444 on Build GRCh38
rs796936623 Chr.19: 35034494-35034494 on Build GRCh38
rs1017897756 Chr.19: 35034555-35034555 on Build GRCh38
rs373458145 Chr.19: 35034148-35034148 on Build GRCh38
rs867287333 Chr.19: 35034334-35034334 on Build GRCh38
rs892513929 Chr.19: 35034501-35034501 on Build GRCh38
rs916986753 Chr.19: 35034410-35034410 on Build GRCh38
rs550906947 Chr.19: 35034308-35034308 on Build GRCh38
rs542946764 Chr.19: 35034481-35034481 on Build GRCh38

View Full Product Details