Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 35907361-35907859 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TYROBP
Entrez Gene ID
7305
Gene Name
TYRO protein tyrosine kinase binding protein
Gene Aliases
DAP12, KARAP, PLOSL
UniGene
Hs.515369
Gene Chromosome Location
Chr.19: 35904401-35908309 on Build GRCh38
Transcripts
NM_001173514.1 NM_001173515.1 NM_003332.3 NM_198125.2 NR_033390.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
255585 c.223G>A p.A75T Chr.19: 35907452-35907452 on Build GRCh38
7138145 c.215G>A p.R72Q Chr.19: 35907460-35907460 on Build GRCh38
264509 c.80C>A p.A27D Chr.19: 35907744-35907744 on Build GRCh38
4956920 c.146G>A p.G49E Chr.19: 35907529-35907529 on Build GRCh38
995326 c.225G>A p.A75A Chr.19: 35907450-35907450 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs386833841 Chr.19: 35907530-35907530 on Build GRCh38
rs892808978 Chr.19: 35907613-35907613 on Build GRCh38
rs775339428 Chr.19: 35907390-35907391 on Build GRCh38
rs945841592 Chr.19: 35907426-35907426 on Build GRCh38
rs769117922 Chr.19: 35907623-35907623 on Build GRCh38
rs761288020 Chr.19: 35907792-35907792 on Build GRCh38
rs747153455 Chr.19: 35907399-35907399 on Build GRCh38
rs772952999 Chr.19: 35907453-35907453 on Build GRCh38
rs949963051 Chr.19: 35907747-35907747 on Build GRCh38
rs1049371263 Chr.19: 35907832-35907832 on Build GRCh38
rs767032012 Chr.19: 35907451-35907451 on Build GRCh38
rs996548979 Chr.19: 35907816-35907816 on Build GRCh38
rs770315263 Chr.19: 35907737-35907737 on Build GRCh38
rs765613710 Chr.19: 35907457-35907457 on Build GRCh38
rs762153343 Chr.19: 35907615-35907615 on Build GRCh38
rs771582920 Chr.19: 35907518-35907518 on Build GRCh38
rs940764664 Chr.19: 35907498-35907498 on Build GRCh38
rs773619483 Chr.19: 35907782-35907782 on Build GRCh38
rs759024155 Chr.19: 35907393-35907393 on Build GRCh38
rs771576196 Chr.19: 35907583-35907583 on Build GRCh38
rs546257418 Chr.19: 35907479-35907479 on Build GRCh38
rs771964613 Chr.19: 35907574-35907574 on Build GRCh38
rs767648127 Chr.19: 35907626-35907631 on Build GRCh38
rs745610215 Chr.19: 35907427-35907427 on Build GRCh38
rs756641773 Chr.19: 35907745-35907750 on Build GRCh38
rs774298388 Chr.19: 35907510-35907510 on Build GRCh38
rs116987202 Chr.19: 35907691-35907691 on Build GRCh38
rs866850462 Chr.19: 35907464-35907464 on Build GRCh38
rs865968843 Chr.19: 35907771-35907771 on Build GRCh38
rs751794674 Chr.19: 35907565-35907566 on Build GRCh38
rs375160467 Chr.19: 35907620-35907620 on Build GRCh38
rs895131769 Chr.19: 35907655-35907655 on Build GRCh38
rs1025192583 Chr.19: 35907661-35907661 on Build GRCh38
rs377715073 Chr.19: 35907674-35907674 on Build GRCh38
rs768644319 Chr.19: 35907439-35907439 on Build GRCh38
rs759301317 Chr.19: 35907603-35907603 on Build GRCh38
rs754097137 Chr.19: 35907388-35907388 on Build GRCh38
rs373306748 Chr.19: 35907705-35907705 on Build GRCh38
rs55746266 Chr.19: 35907720-35907720 on Build GRCh38
rs775196278 Chr.19: 35907435-35907435 on Build GRCh38
rs200634946 Chr.19: 35907466-35907466 on Build GRCh38
rs368252168 Chr.19: 35907555-35907555 on Build GRCh38
rs754616302 Chr.19: 35907636-35907636 on Build GRCh38
rs965056847 Chr.19: 35907704-35907704 on Build GRCh38
rs1013778759 Chr.19: 35907501-35907501 on Build GRCh38
rs749207597 Chr.19: 35907436-35907436 on Build GRCh38
rs903905038 Chr.19: 35907458-35907458 on Build GRCh38
rs758290972 Chr.19: 35907730-35907730 on Build GRCh38
rs200694727 Chr.19: 35907564-35907564 on Build GRCh38
rs78458519 Chr.19: 35907551-35907551 on Build GRCh38
rs780898439 Chr.19: 35907745-35907745 on Build GRCh38
rs766014534 Chr.19: 35907538-35907538 on Build GRCh38
rs766428957 Chr.19: 35907805-35907805 on Build GRCh38
rs376287189 Chr.19: 35907433-35907433 on Build GRCh38
rs767157247 Chr.19: 35907387-35907387 on Build GRCh38
rs201680032 Chr.19: 35907556-35907556 on Build GRCh38
rs775930876 Chr.19: 35907545-35907545 on Build GRCh38
rs760287368 Chr.19: 35907455-35907455 on Build GRCh38
rs753219755 Chr.19: 35907392-35907392 on Build GRCh38
rs868657492 Chr.19: 35907634-35907634 on Build GRCh38
rs781498043 Chr.19: 35907422-35907422 on Build GRCh38
rs754801907 Chr.19: 35907562-35907562 on Build GRCh38
rs199707205 Chr.19: 35907600-35907600 on Build GRCh38
rs754032685 Chr.19: 35907707-35907707 on Build GRCh38
rs769635655 Chr.19: 35907757-35907757 on Build GRCh38
rs77782321 Chr.19: 35907512-35907512 on Build GRCh38
rs777988814 Chr.19: 35907394-35907394 on Build GRCh38
rs111477177 Chr.19: 35907552-35907552 on Build GRCh38
rs138901102 Chr.19: 35907579-35907579 on Build GRCh38
rs79272253 Chr.19: 35907756-35907756 on Build GRCh38
rs368476838 Chr.19: 35907450-35907450 on Build GRCh38
rs778668986 Chr.19: 35907713-35907713 on Build GRCh38
rs762708303 Chr.19: 35907523-35907524 on Build GRCh38
rs913126081 Chr.19: 35907591-35907591 on Build GRCh38
rs774787743 Chr.19: 35907764-35907764 on Build GRCh38
rs759752532 Chr.19: 35907808-35907808 on Build GRCh38
rs201813149 Chr.19: 35907391-35907391 on Build GRCh38
rs755244402 Chr.19: 35907712-35907712 on Build GRCh38
rs757443299 Chr.19: 35907472-35907472 on Build GRCh38
rs570283829 Chr.19: 35907504-35907504 on Build GRCh38
rs1042857888 Chr.19: 35907633-35907633 on Build GRCh38
rs1055564821 Chr.19: 35907642-35907642 on Build GRCh38
rs757488165 Chr.19: 35907418-35907418 on Build GRCh38
rs200264632 Chr.19: 35907467-35907467 on Build GRCh38
rs746506929 Chr.19: 35907590-35907590 on Build GRCh38
rs778559672 Chr.19: 35907672-35907672 on Build GRCh38
rs753979118 Chr.19: 35907806-35907806 on Build GRCh38
rs386833839 Chr.19: 35907559-35907559 on Build GRCh38
rs961541224 Chr.19: 35907505-35907505 on Build GRCh38
rs371976856 Chr.19: 35907812-35907812 on Build GRCh38
rs770560742 Chr.19: 35907594-35907594 on Build GRCh38
rs142807700 Chr.19: 35907731-35907731 on Build GRCh38
rs760363848 Chr.19: 35907537-35907537 on Build GRCh38
rs118043958 Chr.19: 35907659-35907659 on Build GRCh38
rs372140827 Chr.19: 35907535-35907535 on Build GRCh38
rs372455462 Chr.19: 35907626-35907626 on Build GRCh38
rs773779731 Chr.19: 35907444-35907444 on Build GRCh38
rs756345678 Chr.19: 35907489-35907489 on Build GRCh38
rs376453383 Chr.19: 35907549-35907549 on Build GRCh38
rs994296322 Chr.19: 35907673-35907673 on Build GRCh38
rs199970556 Chr.19: 35907563-35907563 on Build GRCh38
rs545730787 Chr.19: 35907447-35907447 on Build GRCh38
rs386833840 Chr.19: 35907534-35907534 on Build GRCh38
rs369808065 Chr.19: 35907700-35907700 on Build GRCh38
rs199583341 Chr.19: 35907495-35907495 on Build GRCh38
rs750455140 Chr.19: 35907696-35907696 on Build GRCh38
rs936762148 Chr.19: 35907641-35907641 on Build GRCh38
rs563691426 Chr.19: 35907460-35907460 on Build GRCh38
rs920668026 Chr.19: 35907703-35907703 on Build GRCh38
rs762374639 Chr.19: 35907627-35907627 on Build GRCh38
rs748058074 Chr.19: 35907565-35907565 on Build GRCh38
rs750536324 Chr.19: 35907629-35907630 on Build GRCh38

View Full Product Details