Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 36392569-36393096 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZFP82
Entrez Gene ID
284406
Gene Name
ZFP82 zinc finger protein
Gene Aliases
ZNF545
UniGene
Hs.558734
Gene Chromosome Location
Chr.19: 36383102-36418674 on Build GRCh38
Transcripts
NM_001321917.1 NM_001321918.1 NM_001321919.1 NM_133466.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
995428 c.1493G>T p.R498I Chr.19: 36392847-36392847 on Build GRCh38
6826834 c.1598A>G p.*533* Chr.19: 36392742-36392742 on Build GRCh38
995430 c.1384C>T p.R462C Chr.19: 36392956-36392956 on Build GRCh38
3532915 c.1299A>C p.L433L Chr.19: 36393041-36393041 on Build GRCh38
6826819 c.1502C>A p.S501Y Chr.19: 36392838-36392838 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764849093 Chr.19: 36393052-36393052 on Build GRCh38
rs759046400 Chr.19: 36393044-36393044 on Build GRCh38
rs754661586 Chr.19: 36392785-36392785 on Build GRCh38
rs373478166 Chr.19: 36392678-36392678 on Build GRCh38
rs953332965 Chr.19: 36392974-36392974 on Build GRCh38
rs147225050 Chr.19: 36392735-36392735 on Build GRCh38
rs373561183 Chr.19: 36392955-36392955 on Build GRCh38
rs749438389 Chr.19: 36392934-36392934 on Build GRCh38
rs1014234750 Chr.19: 36392777-36392777 on Build GRCh38
rs1047255903 Chr.19: 36392660-36392660 on Build GRCh38
rs753446973 Chr.19: 36392773-36392773 on Build GRCh38
rs759091841 Chr.19: 36392639-36392639 on Build GRCh38
rs778257626 Chr.19: 36392715-36392715 on Build GRCh38
rs979493514 Chr.19: 36392599-36392599 on Build GRCh38
rs540537748 Chr.19: 36392878-36392878 on Build GRCh38
rs147661313 Chr.19: 36392985-36392985 on Build GRCh38
rs765045185 Chr.19: 36392851-36392851 on Build GRCh38
rs768926188 Chr.19: 36392750-36392750 on Build GRCh38
rs760697739 Chr.19: 36392767-36392767 on Build GRCh38
rs757831741 Chr.19: 36392719-36392719 on Build GRCh38
rs774558090 Chr.19: 36392772-36392775 on Build GRCh38
rs868864203 Chr.19: 36392805-36392805 on Build GRCh38
rs867982467 Chr.19: 36392989-36392989 on Build GRCh38
rs545857530 Chr.19: 36392684-36392684 on Build GRCh38
rs866079058 Chr.19: 36392838-36392838 on Build GRCh38
rs1010423102 Chr.19: 36392653-36392653 on Build GRCh38
rs892182977 Chr.19: 36392642-36392642 on Build GRCh38
rs1052125360 Chr.19: 36392634-36392634 on Build GRCh38
rs781646270 Chr.19: 36392903-36392903 on Build GRCh38
rs1060813 Chr.19: 36392825-36392825 on Build GRCh38
rs765935119 Chr.19: 36392702-36392702 on Build GRCh38
rs767358685 Chr.19: 36392753-36392753 on Build GRCh38
rs962396976 Chr.19: 36392915-36392915 on Build GRCh38
rs749493416 Chr.19: 36392749-36392749 on Build GRCh38
rs756696848 Chr.19: 36392920-36392920 on Build GRCh38
rs1021451717 Chr.19: 36392917-36392917 on Build GRCh38
rs866579464 Chr.19: 36392763-36392763 on Build GRCh38
rs1034780087 Chr.19: 36392740-36392740 on Build GRCh38
rs764708807 Chr.19: 36392656-36392656 on Build GRCh38
rs557054543 Chr.19: 36392647-36392647 on Build GRCh38
rs775682597 Chr.19: 36392747-36392747 on Build GRCh38
rs1040541503 Chr.19: 36392631-36392631 on Build GRCh38
rs561962329 Chr.19: 36393041-36393041 on Build GRCh38
rs757650164 Chr.19: 36392899-36392899 on Build GRCh38
rs911553906 Chr.19: 36392601-36392601 on Build GRCh38
rs781482349 Chr.19: 36392721-36392721 on Build GRCh38
rs943627281 Chr.19: 36392630-36392630 on Build GRCh38
rs764203189 Chr.19: 36392736-36392739 on Build GRCh38
rs1043578798 Chr.19: 36392661-36392661 on Build GRCh38
rs370140226 Chr.19: 36392938-36392938 on Build GRCh38
rs1015027657 Chr.19: 36392897-36392897 on Build GRCh38
rs752360309 Chr.19: 36393056-36393056 on Build GRCh38
rs754716790 Chr.19: 36392711-36392711 on Build GRCh38
rs771723650 Chr.19: 36392968-36392968 on Build GRCh38
rs368863670 Chr.19: 36393059-36393059 on Build GRCh38
rs865951488 Chr.19: 36393000-36393000 on Build GRCh38
rs190880454 Chr.19: 36392679-36392679 on Build GRCh38
rs753390696 Chr.19: 36392706-36392706 on Build GRCh38
rs923396013 Chr.19: 36393047-36393047 on Build GRCh38
rs780295177 Chr.19: 36392623-36392623 on Build GRCh38
rs905094835 Chr.19: 36392704-36392704 on Build GRCh38
rs143115887 Chr.19: 36392995-36392995 on Build GRCh38
rs763282137 Chr.19: 36392730-36392731 on Build GRCh38
rs202056836 Chr.19: 36392918-36392918 on Build GRCh38
rs375799561 Chr.19: 36392926-36392926 on Build GRCh38
rs933695440 Chr.19: 36392642-36392643 on Build GRCh38
rs767755496 Chr.19: 36393069-36393069 on Build GRCh38
rs747433648 Chr.19: 36392718-36392718 on Build GRCh38
rs376689366 Chr.19: 36392757-36392757 on Build GRCh38
rs766242148 Chr.19: 36392994-36392994 on Build GRCh38
rs140263107 Chr.19: 36392770-36392770 on Build GRCh38
rs760506069 Chr.19: 36393008-36393008 on Build GRCh38
rs779050977 Chr.19: 36392956-36392956 on Build GRCh38
rs769727289 Chr.19: 36392738-36392738 on Build GRCh38
rs770955684 Chr.19: 36393035-36393035 on Build GRCh38
rs774130621 Chr.19: 36392752-36392752 on Build GRCh38

View Full Product Details