Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 36513955-36514474 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF260
Entrez Gene ID
339324
Gene Name
zinc finger protein 260
Gene Aliases
OZRF1, PEX1, ZFP260
UniGene
Hs.18103
Gene Chromosome Location
Chr.19: 36510687-36528488 on Build GRCh38
Transcripts
NM_001012756.2 NM_001166036.1 NM_001166037.1 NM_001166038.1 XM_011526897.2 XM_017026741.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
158618 c.1084T>C p.C362R Chr.19: 36514155-36514155 on Build GRCh38
6260953 c.1209C>A p.H403Q Chr.19: 36514030-36514030 on Build GRCh38
4743352 c.845G>A p.G282E Chr.19: 36514394-36514394 on Build GRCh38
4511595 c.873C>T p.L291L Chr.19: 36514366-36514366 on Build GRCh38
3199692 c.1074A>C p.K358N Chr.19: 36514165-36514165 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs899220108 Chr.19: 36514043-36514043 on Build GRCh38
rs375762580 Chr.19: 36514147-36514147 on Build GRCh38
rs534977672 Chr.19: 36514301-36514301 on Build GRCh38
rs747268495 Chr.19: 36514128-36514128 on Build GRCh38
rs778007819 Chr.19: 36514124-36514124 on Build GRCh38
rs773025394 Chr.19: 36514327-36514327 on Build GRCh38
rs774074050 Chr.19: 36514298-36514298 on Build GRCh38
rs371232522 Chr.19: 36514021-36514021 on Build GRCh38
rs371687431 Chr.19: 36514105-36514105 on Build GRCh38
rs754191663 Chr.19: 36514243-36514243 on Build GRCh38
rs763553967 Chr.19: 36514192-36514192 on Build GRCh38
rs866047031 Chr.19: 36514157-36514157 on Build GRCh38
rs757342173 Chr.19: 36514365-36514365 on Build GRCh38
rs1052753114 Chr.19: 36514359-36514359 on Build GRCh38
rs759909978 Chr.19: 36514328-36514328 on Build GRCh38
rs779772633 Chr.19: 36514411-36514411 on Build GRCh38
rs375644215 Chr.19: 36514261-36514261 on Build GRCh38
rs757539375 Chr.19: 36514275-36514275 on Build GRCh38
rs748204771 Chr.19: 36514118-36514118 on Build GRCh38
rs775467971 Chr.19: 36514280-36514280 on Build GRCh38
rs765556335 Chr.19: 36514332-36514332 on Build GRCh38
rs753568274 Chr.19: 36514075-36514075 on Build GRCh38
rs762373643 Chr.19: 36514037-36514037 on Build GRCh38
rs868515399 Chr.19: 36514315-36514315 on Build GRCh38
rs762485183 Chr.19: 36514191-36514191 on Build GRCh38
rs781342932 Chr.19: 36514375-36514375 on Build GRCh38
rs770739958 Chr.19: 36513980-36513980 on Build GRCh38
rs934437720 Chr.19: 36514352-36514352 on Build GRCh38
rs751202661 Chr.19: 36514054-36514054 on Build GRCh38
rs182026084 Chr.19: 36514296-36514296 on Build GRCh38
rs761415920 Chr.19: 36514299-36514299 on Build GRCh38
rs80254552 Chr.19: 36514138-36514138 on Build GRCh38
rs892776872 Chr.19: 36514389-36514389 on Build GRCh38
rs201257718 Chr.19: 36514279-36514279 on Build GRCh38
rs144667068 Chr.19: 36514088-36514088 on Build GRCh38
rs755915441 Chr.19: 36514409-36514409 on Build GRCh38
rs745603821 Chr.19: 36514278-36514278 on Build GRCh38
rs779009925 Chr.19: 36514442-36514442 on Build GRCh38
rs776515747 Chr.19: 36513985-36513985 on Build GRCh38
rs373841868 Chr.19: 36514249-36514249 on Build GRCh38
rs1010669453 Chr.19: 36514219-36514219 on Build GRCh38
rs369966262 Chr.19: 36514217-36514217 on Build GRCh38
rs1030839945 Chr.19: 36514174-36514174 on Build GRCh38
rs765730160 Chr.19: 36514256-36514256 on Build GRCh38
rs548369914 Chr.19: 36514132-36514132 on Build GRCh38
rs763743850 Chr.19: 36514045-36514045 on Build GRCh38
rs774729431 Chr.19: 36514004-36514005 on Build GRCh38
rs755193624 Chr.19: 36514398-36514398 on Build GRCh38
rs764711249 Chr.19: 36514029-36514029 on Build GRCh38
rs769403421 Chr.19: 36513990-36513990 on Build GRCh38
rs758434038 Chr.19: 36514119-36514119 on Build GRCh38
rs752206231 Chr.19: 36514401-36514401 on Build GRCh38
rs747215641 Chr.19: 36514271-36514271 on Build GRCh38
rs753221805 Chr.19: 36514341-36514341 on Build GRCh38
rs768414071 Chr.19: 36514427-36514427 on Build GRCh38
rs751570165 Chr.19: 36514354-36514354 on Build GRCh38
rs759517086 Chr.19: 36514025-36514025 on Build GRCh38
rs754385676 Chr.19: 36514085-36514085 on Build GRCh38
rs890458488 Chr.19: 36514083-36514083 on Build GRCh38
rs779145175 Chr.19: 36514076-36514077 on Build GRCh38
rs775303025 Chr.19: 36514159-36514159 on Build GRCh38
rs866586932 Chr.19: 36514097-36514097 on Build GRCh38
rs528202813 Chr.19: 36514414-36514414 on Build GRCh38
rs530244779 Chr.19: 36514104-36514104 on Build GRCh38
rs552762595 Chr.19: 36514233-36514233 on Build GRCh38
rs932071411 Chr.19: 36514052-36514052 on Build GRCh38
rs774127028 Chr.19: 36514210-36514210 on Build GRCh38
rs367862666 Chr.19: 36514091-36514091 on Build GRCh38
rs764151687 Chr.19: 36514351-36514351 on Build GRCh38
rs756516339 Chr.19: 36514065-36514065 on Build GRCh38
rs749233923 Chr.19: 36514295-36514295 on Build GRCh38
rs745799752 Chr.19: 36514150-36514150 on Build GRCh38
rs866726867 Chr.19: 36514272-36514272 on Build GRCh38
rs76808668 Chr.19: 36514396-36514396 on Build GRCh38
rs749245716 Chr.19: 36514412-36514412 on Build GRCh38
rs2967505 Chr.19: 36514036-36514036 on Build GRCh38
rs758884870 Chr.19: 36514346-36514346 on Build GRCh38

View Full Product Details