Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 36626842-36627355 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF382
Entrez Gene ID
84911
Gene Name
zinc finger protein 382
Gene Aliases
KS1
UniGene
Hs.631591
Gene Chromosome Location
Chr.19: 36605305-36628597 on Build GRCh38
Transcripts
NM_001256838.1 NM_032825.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1233962 c.1129G>A p.E377K Chr.19: 36627026-36627026 on Build GRCh38
6829724 c.1139A>G p.Y380C Chr.19: 36627036-36627036 on Build GRCh38
6017452 c.1341G>A p.K447K Chr.19: 36627241-36627241 on Build GRCh38
5077937 c.1373_1374insG p.E460fs*7 Chr.19: 36627273-36627274 on Build GRCh38
126381 c.1286C>A p.T429N Chr.19: 36627183-36627183 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763709234 Chr.19: 36627261-36627261 on Build GRCh38
rs755387596 Chr.19: 36627307-36627307 on Build GRCh38
rs373673657 Chr.19: 36626962-36626962 on Build GRCh38
rs200339859 Chr.19: 36627226-36627226 on Build GRCh38
rs955316468 Chr.19: 36627106-36627106 on Build GRCh38
rs868687181 Chr.19: 36626870-36626870 on Build GRCh38
rs746443074 Chr.19: 36626877-36626877 on Build GRCh38
rs765710784 Chr.19: 36627166-36627166 on Build GRCh38
rs769162959 Chr.19: 36627137-36627137 on Build GRCh38
rs772376239 Chr.19: 36627163-36627163 on Build GRCh38
rs568283174 Chr.19: 36627123-36627123 on Build GRCh38
rs374650096 Chr.19: 36626897-36626897 on Build GRCh38
rs749359129 Chr.19: 36627005-36627005 on Build GRCh38
rs377727509 Chr.19: 36626924-36626924 on Build GRCh38
rs761973119 Chr.19: 36627256-36627256 on Build GRCh38
rs148103215 Chr.19: 36627022-36627022 on Build GRCh38
rs572109074 Chr.19: 36627189-36627189 on Build GRCh38
rs201684196 Chr.19: 36627079-36627079 on Build GRCh38
rs770689264 Chr.19: 36627202-36627202 on Build GRCh38
rs377155785 Chr.19: 36626891-36626891 on Build GRCh38
rs774933779 Chr.19: 36627241-36627241 on Build GRCh38
rs780590866 Chr.19: 36627118-36627118 on Build GRCh38
rs764921340 Chr.19: 36626971-36626971 on Build GRCh38
rs762483446 Chr.19: 36627244-36627244 on Build GRCh38
rs894031247 Chr.19: 36627033-36627033 on Build GRCh38
rs1032659383 Chr.19: 36626999-36626999 on Build GRCh38
rs757988766 Chr.19: 36627094-36627094 on Build GRCh38
rs762966331 Chr.19: 36626914-36626914 on Build GRCh38
rs369768939 Chr.19: 36626939-36626939 on Build GRCh38
rs766724243 Chr.19: 36627280-36627280 on Build GRCh38
rs201197350 Chr.19: 36627082-36627082 on Build GRCh38
rs185047483 Chr.19: 36626992-36626992 on Build GRCh38
rs758599737 Chr.19: 36627329-36627329 on Build GRCh38
rs779591609 Chr.19: 36627317-36627317 on Build GRCh38
rs878959074 Chr.19: 36627233-36627233 on Build GRCh38
rs867969154 Chr.19: 36627057-36627057 on Build GRCh38
rs975324349 Chr.19: 36627206-36627206 on Build GRCh38
rs756566547 Chr.19: 36626998-36626998 on Build GRCh38
rs553797186 Chr.19: 36627127-36627127 on Build GRCh38
rs967563089 Chr.19: 36626940-36626940 on Build GRCh38
rs767480822 Chr.19: 36626947-36626947 on Build GRCh38
rs377130465 Chr.19: 36627282-36627282 on Build GRCh38
rs1005527700 Chr.19: 36627124-36627124 on Build GRCh38
rs975309679 Chr.19: 36627054-36627054 on Build GRCh38
rs776479508 Chr.19: 36627048-36627048 on Build GRCh38
rs922467917 Chr.19: 36626960-36626960 on Build GRCh38
rs537671705 Chr.19: 36627000-36627000 on Build GRCh38
rs752791248 Chr.19: 36627318-36627318 on Build GRCh38
rs746247001 Chr.19: 36626995-36626995 on Build GRCh38
rs773141953 Chr.19: 36626955-36626955 on Build GRCh38
rs773090722 Chr.19: 36627024-36627024 on Build GRCh38
rs757614823 Chr.19: 36626982-36626982 on Build GRCh38
rs964323861 Chr.19: 36626881-36626881 on Build GRCh38
rs548897746 Chr.19: 36626965-36626965 on Build GRCh38
rs779075034 Chr.19: 36627145-36627145 on Build GRCh38
rs776625234 Chr.19: 36627217-36627217 on Build GRCh38
rs766247460 Chr.19: 36626963-36626963 on Build GRCh38
rs781469678 Chr.19: 36626989-36626989 on Build GRCh38
rs4439880 Chr.19: 36627096-36627096 on Build GRCh38
rs750747548 Chr.19: 36627099-36627099 on Build GRCh38
rs751264916 Chr.19: 36627273-36627273 on Build GRCh38
rs975186479 Chr.19: 36626885-36626885 on Build GRCh38
rs766307873 Chr.19: 36627045-36627045 on Build GRCh38
rs1045241896 Chr.19: 36627073-36627073 on Build GRCh38
rs906739724 Chr.19: 36627084-36627084 on Build GRCh38
rs748257565 Chr.19: 36627155-36627155 on Build GRCh38
rs752404502 Chr.19: 36626981-36626981 on Build GRCh38
rs754485289 Chr.19: 36626970-36626970 on Build GRCh38
rs1025700489 Chr.19: 36627097-36627097 on Build GRCh38
rs200498882 Chr.19: 36627034-36627034 on Build GRCh38
rs577820635 Chr.19: 36627059-36627059 on Build GRCh38
rs761683134 Chr.19: 36626941-36626941 on Build GRCh38
rs1003622333 Chr.19: 36627087-36627087 on Build GRCh38
rs941444046 Chr.19: 36627279-36627279 on Build GRCh38
rs200497123 Chr.19: 36626898-36626898 on Build GRCh38
rs768671768 Chr.19: 36627019-36627019 on Build GRCh38
rs761091621 Chr.19: 36627274-36627274 on Build GRCh38
rs757782750 Chr.19: 36626949-36626949 on Build GRCh38
rs760133178 Chr.19: 36627117-36627117 on Build GRCh38
rs935644765 Chr.19: 36627014-36627014 on Build GRCh38

View Full Product Details