Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 36878303-36878828 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF345
Entrez Gene ID
25850
Gene Name
zinc finger protein 345
Gene Aliases
HZF10
UniGene
Hs.362324
Gene Chromosome Location
Chr.19: 36850350-36893218 on Build GRCh38
Transcripts
NM_001242472.1 NM_001242474.1 NM_001242475.1 NM_001242476.1 NM_003419.4 NR_038362.1 XM_011526704.2 XM_017026568.1 XM_017026569.1 XM_017026570.1 XM_017026571.1 XM_017026572.1 XM_017026573.1

SNP Detail

SNP ID Chromosome Location
rs948245281 Chr.19: 36878375-36878375 on Build GRCh38
rs906666691 Chr.19: 36878405-36878405 on Build GRCh38
rs545711507 Chr.19: 36878756-36878757 on Build GRCh38
rs530114753 Chr.19: 36878397-36878397 on Build GRCh38
rs185864842 Chr.19: 36878469-36878469 on Build GRCh38
rs559536753 Chr.19: 36878381-36878381 on Build GRCh38
rs778293292 Chr.19: 36878346-36878346 on Build GRCh38
rs536076121 Chr.19: 36878706-36878706 on Build GRCh38
rs555023 Chr.19: 36878552-36878552 on Build GRCh38
rs374555020 Chr.19: 36878379-36878379 on Build GRCh38
rs181850010 Chr.19: 36878587-36878587 on Build GRCh38
rs971731394 Chr.19: 36878459-36878459 on Build GRCh38
rs894057127 Chr.19: 36878374-36878374 on Build GRCh38
rs368685872 Chr.19: 36878688-36878688 on Build GRCh38
rs7259042 Chr.19: 36878476-36878476 on Build GRCh38
rs7256044 Chr.19: 36878641-36878641 on Build GRCh38
rs397859584 Chr.19: 36878633-36878633 on Build GRCh38
rs919913917 Chr.19: 36878609-36878609 on Build GRCh38
rs963974219 Chr.19: 36878509-36878509 on Build GRCh38
rs139360979 Chr.19: 36878429-36878429 on Build GRCh38
rs75247927 Chr.19: 36878726-36878726 on Build GRCh38
rs534001871 Chr.19: 36878534-36878535 on Build GRCh38
rs892756529 Chr.19: 36878796-36878796 on Build GRCh38
rs1003648969 Chr.19: 36878415-36878415 on Build GRCh38
rs990305839 Chr.19: 36878506-36878506 on Build GRCh38
rs973038681 Chr.19: 36878593-36878593 on Build GRCh38
rs576810009 Chr.19: 36878579-36878579 on Build GRCh38
rs931369452 Chr.19: 36878624-36878625 on Build GRCh38
rs957067039 Chr.19: 36878696-36878696 on Build GRCh38
rs796958961 Chr.19: 36878623-36878624 on Build GRCh38
rs975678743 Chr.19: 36878550-36878550 on Build GRCh38
rs915509255 Chr.19: 36878764-36878764 on Build GRCh38
rs180708969 Chr.19: 36878351-36878351 on Build GRCh38
rs10709788 Chr.19: 36878624-36878624 on Build GRCh38
rs1054076815 Chr.19: 36878366-36878366 on Build GRCh38
rs895238503 Chr.19: 36878686-36878686 on Build GRCh38
rs921017988 Chr.19: 36878512-36878515 on Build GRCh38
rs186903245 Chr.19: 36878772-36878772 on Build GRCh38
rs1024194386 Chr.19: 36878436-36878436 on Build GRCh38
rs763041402 Chr.19: 36878350-36878350 on Build GRCh38
rs999163823 Chr.19: 36878757-36878757 on Build GRCh38
rs955358086 Chr.19: 36878636-36878636 on Build GRCh38
rs754887294 Chr.19: 36878336-36878336 on Build GRCh38
rs191421113 Chr.19: 36878478-36878478 on Build GRCh38
rs544642262 Chr.19: 36878594-36878594 on Build GRCh38
rs1045607003 Chr.19: 36878723-36878723 on Build GRCh38

View Full Product Details