Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 36916222-36916723 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF568
Entrez Gene ID
374900
Gene Name
zinc finger protein 568
Gene Aliases
-
UniGene
Hs.404220
Gene Chromosome Location
Chr.19: 36916329-36997932 on Build GRCh38
Transcripts
NM_001204835.1 NM_001204836.1 NM_001204837.1 NM_001204838.1 NM_001204839.1 NM_198539.3 XM_017026772.1 XM_017026773.1 XM_017026774.1 XM_017026775.1 XM_017026779.1 XM_017026780.1 NM_001037232.3 XM_011526933.2

Gene Symbol
ZNF829
Entrez Gene ID
374899
Gene Name
zinc finger protein 829
Gene Aliases
-
UniGene
Hs.720312
Gene Chromosome Location
Chr.19: 36888124-36916387 on Build GRCh38
Transcripts
NM_001204835.1 NM_001204836.1 NM_001204837.1 NM_001204838.1 NM_001204839.1 NM_198539.3 XM_017026772.1 XM_017026773.1 XM_017026774.1 XM_017026775.1 XM_017026779.1 XM_017026780.1 NM_001037232.3 XM_011526933.2

SNP Detail

SNP ID Chromosome Location
rs895779852 Chr.19: 36916393-36916393 on Build GRCh38
rs974151911 Chr.19: 36916651-36916651 on Build GRCh38
rs937174943 Chr.19: 36916680-36916680 on Build GRCh38
rs763379110 Chr.19: 36916461-36916461 on Build GRCh38
rs1041409095 Chr.19: 36916439-36916439 on Build GRCh38
rs752100799 Chr.19: 36916494-36916494 on Build GRCh38
rs560260213 Chr.19: 36916284-36916284 on Build GRCh38
rs992439878 Chr.19: 36916511-36916511 on Build GRCh38
rs561223336 Chr.19: 36916387-36916387 on Build GRCh38
rs919647636 Chr.19: 36916554-36916554 on Build GRCh38
rs1037138614 Chr.19: 36916256-36916256 on Build GRCh38
rs1019109559 Chr.19: 36916363-36916363 on Build GRCh38
rs922856586 Chr.19: 36916244-36916244 on Build GRCh38
rs991318922 Chr.19: 36916685-36916685 on Build GRCh38
rs963047441 Chr.19: 36916628-36916628 on Build GRCh38
rs972510587 Chr.19: 36916552-36916552 on Build GRCh38
rs955319193 Chr.19: 36916258-36916258 on Build GRCh38
rs901960709 Chr.19: 36916413-36916413 on Build GRCh38
rs566612910 Chr.19: 36916583-36916583 on Build GRCh38
rs1055599919 Chr.19: 36916376-36916376 on Build GRCh38
rs926704159 Chr.19: 36916652-36916655 on Build GRCh38
rs939783724 Chr.19: 36916245-36916245 on Build GRCh38
rs1008165313 Chr.19: 36916361-36916361 on Build GRCh38
rs531854493 Chr.19: 36916417-36916417 on Build GRCh38
rs930996527 Chr.19: 36916637-36916637 on Build GRCh38
rs4805190 Chr.19: 36916312-36916312 on Build GRCh38
rs917749983 Chr.19: 36916533-36916533 on Build GRCh38
rs565519719 Chr.19: 36916647-36916648 on Build GRCh38
rs982820156 Chr.19: 36916268-36916268 on Build GRCh38
rs184021721 Chr.19: 36916466-36916466 on Build GRCh38
rs1038881224 Chr.19: 36916302-36916302 on Build GRCh38
rs1029917514 Chr.19: 36916519-36916519 on Build GRCh38
rs539469702 Chr.19: 36916674-36916674 on Build GRCh38
rs548997533 Chr.19: 36916342-36916342 on Build GRCh38
rs902911049 Chr.19: 36916462-36916462 on Build GRCh38
rs77000225 Chr.19: 36916537-36916537 on Build GRCh38
rs533755699 Chr.19: 36916647-36916647 on Build GRCh38
rs975632380 Chr.19: 36916243-36916243 on Build GRCh38
rs898170886 Chr.19: 36916275-36916275 on Build GRCh38
rs930925490 Chr.19: 36916296-36916296 on Build GRCh38
rs61249302 Chr.19: 36916483-36916483 on Build GRCh38
rs117744823 Chr.19: 36916484-36916484 on Build GRCh38
rs908608147 Chr.19: 36916272-36916272 on Build GRCh38
rs949710898 Chr.19: 36916410-36916410 on Build GRCh38
rs757850563 Chr.19: 36916567-36916567 on Build GRCh38
rs997019385 Chr.19: 36916493-36916493 on Build GRCh38
rs1033583226 Chr.19: 36916489-36916489 on Build GRCh38
rs150322722 Chr.19: 36916663-36916663 on Build GRCh38
rs545124850 Chr.19: 36916265-36916265 on Build GRCh38
rs138912412 Chr.19: 36916689-36916689 on Build GRCh38

View Full Product Details