Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 37149998-37150502 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF585A
Entrez Gene ID
199704
Gene Name
zinc finger protein 585A
Gene Aliases
-
UniGene
Hs.659236
Gene Chromosome Location
Chr.19: 37147438-37172741 on Build GRCh38
Transcripts
NM_001288800.1 NM_152655.3 NM_199126.2

SNP Detail

SNP ID Chromosome Location
rs745994421 Chr.19: 37150295-37150295 on Build GRCh38
rs1005079812 Chr.19: 37150252-37150252 on Build GRCh38
rs77284275 Chr.19: 37150099-37150101 on Build GRCh38
rs951904395 Chr.19: 37150072-37150072 on Build GRCh38
rs375097672 Chr.19: 37150314-37150314 on Build GRCh38
rs946149880 Chr.19: 37150151-37150151 on Build GRCh38
rs900056225 Chr.19: 37150409-37150409 on Build GRCh38
rs891326727 Chr.19: 37150167-37150167 on Build GRCh38
rs112254603 Chr.19: 37150343-37150343 on Build GRCh38
rs752118703 Chr.19: 37150047-37150047 on Build GRCh38
rs974412949 Chr.19: 37150039-37150039 on Build GRCh38
rs1056413106 Chr.19: 37150081-37150081 on Build GRCh38
rs1009269618 Chr.19: 37150088-37150088 on Build GRCh38
rs73622720 Chr.19: 37150087-37150087 on Build GRCh38
rs544350768 Chr.19: 37150073-37150073 on Build GRCh38
rs576016086 Chr.19: 37150289-37150289 on Build GRCh38
rs893193528 Chr.19: 37150147-37150147 on Build GRCh38
rs1030456668 Chr.19: 37150423-37150423 on Build GRCh38
rs535882415 Chr.19: 37150186-37150186 on Build GRCh38
rs746576622 Chr.19: 37150199-37150199 on Build GRCh38
rs369604860 Chr.19: 37150451-37150451 on Build GRCh38
rs781173565 Chr.19: 37150257-37150257 on Build GRCh38
rs764580793 Chr.19: 37150033-37150033 on Build GRCh38
rs1027746324 Chr.19: 37150228-37150228 on Build GRCh38
rs568552591 Chr.19: 37150108-37150108 on Build GRCh38
rs900087104 Chr.19: 37150193-37150193 on Build GRCh38
rs927191619 Chr.19: 37150298-37150298 on Build GRCh38
rs912654241 Chr.19: 37150383-37150383 on Build GRCh38
rs559031703 Chr.19: 37150077-37150077 on Build GRCh38
rs997676275 Chr.19: 37150407-37150407 on Build GRCh38
rs534816246 Chr.19: 37150090-37150090 on Build GRCh38
rs1014753266 Chr.19: 37150288-37150288 on Build GRCh38
rs1000512090 Chr.19: 37150397-37150397 on Build GRCh38
rs866588478 Chr.19: 37150091-37150091 on Build GRCh38
rs1048719869 Chr.19: 37150452-37150452 on Build GRCh38
rs944147606 Chr.19: 37150401-37150401 on Build GRCh38
rs540804502 Chr.19: 37150364-37150364 on Build GRCh38
rs1054406266 Chr.19: 37150093-37150093 on Build GRCh38
rs1042227191 Chr.19: 37150378-37150378 on Build GRCh38
rs1005534590 Chr.19: 37150464-37150464 on Build GRCh38
rs563025467 Chr.19: 37150078-37150078 on Build GRCh38
rs1007229889 Chr.19: 37150094-37150108 on Build GRCh38
rs903528956 Chr.19: 37150386-37150386 on Build GRCh38
rs951023248 Chr.19: 37150146-37150146 on Build GRCh38
rs995818666 Chr.19: 37150219-37150219 on Build GRCh38
rs111802547 Chr.19: 37150088-37150088 on Build GRCh38
rs1039944339 Chr.19: 37150177-37150177 on Build GRCh38
rs958629918 Chr.19: 37150309-37150309 on Build GRCh38
rs573398165 Chr.19: 37150351-37150351 on Build GRCh38
rs1039913283 Chr.19: 37150402-37150402 on Build GRCh38
rs553234357 Chr.19: 37150092-37150092 on Build GRCh38
rs995782555 Chr.19: 37150413-37150413 on Build GRCh38
rs887475569 Chr.19: 37150477-37150477 on Build GRCh38
rs530026115 Chr.19: 37150170-37150170 on Build GRCh38
rs927339904 Chr.19: 37150297-37150297 on Build GRCh38
rs138277725 Chr.19: 37150399-37150399 on Build GRCh38
rs557355500 Chr.19: 37150237-37150237 on Build GRCh38
rs1041869476 Chr.19: 37150153-37150153 on Build GRCh38
rs1027367531 Chr.19: 37150058-37150058 on Build GRCh38
rs1018618082 Chr.19: 37150032-37150032 on Build GRCh38
rs529714291 Chr.19: 37150460-37150460 on Build GRCh38

View Full Product Details