Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 37426166-37426585 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF569
Entrez Gene ID
148266
Gene Name
zinc finger protein 569
Gene Aliases
ZAP1, ZNF
UniGene
Hs.511848
Gene Chromosome Location
Chr.19: 37411146-37469268 on Build GRCh38
Transcripts
NM_152484.2 XM_006723046.2 XM_006723047.3 XM_006723048.3 XM_011526538.2 XM_011526539.2 XM_017026376.1 XM_017026377.1 XM_017026378.1 XM_017026379.1 XM_017026380.1 XM_017026381.1 XM_017026382.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5811730 c.86A>T p.Q29L Chr.19: 37426308-37426308 on Build GRCh38
418294 c.46G>A p.D16N Chr.19: 37426348-37426348 on Build GRCh38
3200880 c.79C>T p.P27S Chr.19: 37426315-37426315 on Build GRCh38
3892418 c.78T>C p.D26D Chr.19: 37426316-37426316 on Build GRCh38
1600056 c.19A>T p.T7S Chr.19: 37426375-37426375 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775647312 Chr.19: 37426430-37426430 on Build GRCh38
rs550758553 Chr.19: 37426282-37426282 on Build GRCh38
rs886766875 Chr.19: 37426195-37426195 on Build GRCh38
rs772640874 Chr.19: 37426203-37426203 on Build GRCh38
rs758876046 Chr.19: 37426411-37426411 on Build GRCh38
rs771091714 Chr.19: 37426235-37426235 on Build GRCh38
rs371804860 Chr.19: 37426253-37426253 on Build GRCh38
rs981770995 Chr.19: 37426221-37426221 on Build GRCh38
rs534538271 Chr.19: 37426435-37426435 on Build GRCh38
rs182686660 Chr.19: 37426214-37426214 on Build GRCh38
rs187007327 Chr.19: 37426494-37426494 on Build GRCh38
rs760622871 Chr.19: 37426443-37426443 on Build GRCh38
rs755804446 Chr.19: 37426398-37426398 on Build GRCh38
rs200022900 Chr.19: 37426348-37426348 on Build GRCh38
rs768847098 Chr.19: 37426379-37426380 on Build GRCh38
rs771341306 Chr.19: 37426417-37426417 on Build GRCh38
rs757826900 Chr.19: 37426428-37426429 on Build GRCh38
rs752006845 Chr.19: 37426290-37426290 on Build GRCh38
rs555627155 Chr.19: 37426471-37426471 on Build GRCh38
rs368337278 Chr.19: 37426243-37426243 on Build GRCh38
rs374033929 Chr.19: 37426349-37426349 on Build GRCh38
rs760789777 Chr.19: 37426231-37426231 on Build GRCh38
rs755708774 Chr.19: 37426291-37426291 on Build GRCh38
rs929870673 Chr.19: 37426444-37426444 on Build GRCh38
rs764048548 Chr.19: 37426358-37426358 on Build GRCh38
rs781340596 Chr.19: 37426426-37426426 on Build GRCh38
rs201187258 Chr.19: 37426320-37426320 on Build GRCh38
rs779804567 Chr.19: 37426400-37426400 on Build GRCh38
rs761708799 Chr.19: 37426269-37426269 on Build GRCh38
rs751089348 Chr.19: 37426272-37426272 on Build GRCh38
rs529081401 Chr.19: 37426273-37426273 on Build GRCh38
rs767356761 Chr.19: 37426334-37426334 on Build GRCh38
rs761631983 Chr.19: 37426372-37426372 on Build GRCh38
rs566752914 Chr.19: 37426413-37426413 on Build GRCh38
rs753145020 Chr.19: 37426410-37426410 on Build GRCh38
rs759873498 Chr.19: 37426335-37426335 on Build GRCh38
rs770021923 Chr.19: 37426428-37426428 on Build GRCh38
rs143190321 Chr.19: 37426293-37426293 on Build GRCh38
rs199661417 Chr.19: 37426364-37426364 on Build GRCh38
rs1002462550 Chr.19: 37426361-37426361 on Build GRCh38
rs375677375 Chr.19: 37426427-37426427 on Build GRCh38
rs752488454 Chr.19: 37426254-37426254 on Build GRCh38
rs543785057 Chr.19: 37426477-37426477 on Build GRCh38
rs773439041 Chr.19: 37426215-37426215 on Build GRCh38
rs772728740 Chr.19: 37426296-37426296 on Build GRCh38
rs1048381191 Chr.19: 37426470-37426470 on Build GRCh38
rs561891830 Chr.19: 37426242-37426242 on Build GRCh38
rs116491235 Chr.19: 37426294-37426294 on Build GRCh38
rs762879387 Chr.19: 37426268-37426268 on Build GRCh38
rs749896272 Chr.19: 37426390-37426390 on Build GRCh38
rs776955172 Chr.19: 37426236-37426236 on Build GRCh38
rs767112999 Chr.19: 37426379-37426379 on Build GRCh38
rs911976462 Chr.19: 37426539-37426539 on Build GRCh38
rs747612120 Chr.19: 37426414-37426414 on Build GRCh38
rs762684822 Chr.19: 37426350-37426350 on Build GRCh38
rs771094349 Chr.19: 37426326-37426326 on Build GRCh38
rs987603155 Chr.19: 37426521-37426521 on Build GRCh38
rs569393301 Chr.19: 37426309-37426309 on Build GRCh38
rs750138129 Chr.19: 37426285-37426285 on Build GRCh38
rs747490733 Chr.19: 37426323-37426323 on Build GRCh38

View Full Product Details