Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 38303990-38304495 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C19orf33
Entrez Gene ID
64073
Gene Name
chromosome 19 open reading frame 33
Gene Aliases
H2RSP, IMUP, IMUP-1, IMUP-2
UniGene
Hs.631544
Gene Chromosome Location
Chr.19: 38304161-38305006 on Build GRCh38
Transcripts
NM_001317801.1 NM_033520.2 NM_001039671.2 NM_001039672.2 NM_001039673.2 NM_001145461.1 NM_001145462.1 NM_001145463.1

Gene Symbol
YIF1B
Entrez Gene ID
90522
Gene Name
Yip1 interacting factor homolog B, membrane trafficking protein
Gene Aliases
FinGER8
UniGene
Hs.280741
Gene Chromosome Location
Chr.19: 38303560-38315976 on Build GRCh38
Transcripts
NM_001317801.1 NM_033520.2 NM_001039671.2 NM_001039672.2 NM_001039673.2 NM_001145461.1 NM_001145462.1 NM_001145463.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6203876 c.27G>A p.L9L Chr.19: 38304379-38304379 on Build GRCh38
6203877 c.27G>A p.L9L Chr.19: 38304379-38304379 on Build GRCh38
439390 c.17G>A p.G6E Chr.19: 38304278-38304278 on Build GRCh38
6331346 c.39C>A p.S13S Chr.19: 38304391-38304391 on Build GRCh38
4547723 c.42G>A p.Q14Q Chr.19: 38304394-38304394 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780165916 Chr.19: 38304406-38304406 on Build GRCh38
rs1021815947 Chr.19: 38304186-38304186 on Build GRCh38
rs2304176 Chr.19: 38304255-38304255 on Build GRCh38
rs114727482 Chr.19: 38304190-38304190 on Build GRCh38
rs747884813 Chr.19: 38304314-38304314 on Build GRCh38
rs565666445 Chr.19: 38304456-38304456 on Build GRCh38
rs537750223 Chr.19: 38304161-38304161 on Build GRCh38
rs201946524 Chr.19: 38304372-38304372 on Build GRCh38
rs1036310233 Chr.19: 38304383-38304383 on Build GRCh38
rs927849184 Chr.19: 38304231-38304231 on Build GRCh38
rs749681108 Chr.19: 38304194-38304195 on Build GRCh38
rs373145066 Chr.19: 38304462-38304462 on Build GRCh38
rs371716289 Chr.19: 38304401-38304401 on Build GRCh38
rs372124907 Chr.19: 38304215-38304215 on Build GRCh38
rs35282731 Chr.19: 38304418-38304419 on Build GRCh38
rs764429517 Chr.19: 38304226-38304226 on Build GRCh38
rs939150291 Chr.19: 38304063-38304063 on Build GRCh38
rs749494978 Chr.19: 38304305-38304305 on Build GRCh38
rs559247488 Chr.19: 38304178-38304178 on Build GRCh38
rs762128437 Chr.19: 38304473-38304473 on Build GRCh38
rs374419460 Chr.19: 38304047-38304047 on Build GRCh38
rs1025412347 Chr.19: 38304188-38304188 on Build GRCh38
rs536882195 Chr.19: 38304145-38304145 on Build GRCh38
rs921222552 Chr.19: 38304286-38304286 on Build GRCh38
rs780176597 Chr.19: 38304296-38304296 on Build GRCh38
rs35308902 Chr.19: 38304109-38304110 on Build GRCh38
rs367660814 Chr.19: 38304309-38304309 on Build GRCh38
rs977726917 Chr.19: 38304205-38304205 on Build GRCh38
rs747350011 Chr.19: 38304351-38304351 on Build GRCh38
rs1005334176 Chr.19: 38304449-38304449 on Build GRCh38
rs117478214 Chr.19: 38304334-38304334 on Build GRCh38
rs759185913 Chr.19: 38304373-38304373 on Build GRCh38
rs776204949 Chr.19: 38304370-38304370 on Build GRCh38
rs960543450 Chr.19: 38304236-38304236 on Build GRCh38
rs868674331 Chr.19: 38304198-38304198 on Build GRCh38
rs1048433855 Chr.19: 38304321-38304321 on Build GRCh38
rs754829305 Chr.19: 38304369-38304369 on Build GRCh38
rs374256836 Chr.19: 38304194-38304194 on Build GRCh38
rs572168647 Chr.19: 38304400-38304400 on Build GRCh38
rs887575571 Chr.19: 38304107-38304107 on Build GRCh38
rs1015230872 Chr.19: 38304143-38304143 on Build GRCh38
rs531365851 Chr.19: 38304390-38304390 on Build GRCh38
rs541403080 Chr.19: 38304192-38304192 on Build GRCh38
rs758814002 Chr.19: 38304222-38304222 on Build GRCh38
rs372040444 Chr.19: 38304244-38304244 on Build GRCh38
rs770454639 Chr.19: 38304362-38304362 on Build GRCh38
rs570227484 Chr.19: 38304160-38304160 on Build GRCh38
rs371088164 Chr.19: 38304316-38304316 on Build GRCh38
rs781685759 Chr.19: 38304242-38304242 on Build GRCh38
rs771859056 Chr.19: 38304424-38304424 on Build GRCh38
rs758991899 Chr.19: 38304457-38304457 on Build GRCh38
rs369719001 Chr.19: 38304409-38304409 on Build GRCh38
rs773151210 Chr.19: 38304324-38304324 on Build GRCh38
rs3816044 Chr.19: 38304110-38304110 on Build GRCh38
rs375165148 Chr.19: 38304466-38304466 on Build GRCh38
rs747979155 Chr.19: 38304423-38304423 on Build GRCh38
rs981091283 Chr.19: 38304024-38304024 on Build GRCh38
rs375904286 Chr.19: 38304407-38304407 on Build GRCh38
rs753650253 Chr.19: 38304317-38304317 on Build GRCh38
rs779216511 Chr.19: 38304421-38304421 on Build GRCh38
rs1007177708 Chr.19: 38304148-38304148 on Build GRCh38
rs547355038 Chr.19: 38304425-38304425 on Build GRCh38
rs747949178 Chr.19: 38304185-38304185 on Build GRCh38
rs11878474 Chr.19: 38304180-38304180 on Build GRCh38
rs946175550 Chr.19: 38304302-38304302 on Build GRCh38
rs145255633 Chr.19: 38304053-38304053 on Build GRCh38
rs76682100 Chr.19: 38304146-38304146 on Build GRCh38
rs757770006 Chr.19: 38304229-38304229 on Build GRCh38
rs532051981 Chr.19: 38304419-38304419 on Build GRCh38
rs901395702 Chr.19: 38304396-38304396 on Build GRCh38
rs200055717 Chr.19: 38304216-38304216 on Build GRCh38
rs149154861 Chr.19: 38304398-38304398 on Build GRCh38
rs942655454 Chr.19: 38304329-38304329 on Build GRCh38
rs746951877 Chr.19: 38304427-38304427 on Build GRCh38
rs931404517 Chr.19: 38304303-38304303 on Build GRCh38
rs752128564 Chr.19: 38304228-38304228 on Build GRCh38
rs762121560 Chr.19: 38304395-38304395 on Build GRCh38
rs771915401 Chr.19: 38304317-38304317 on Build GRCh38
rs975814498 Chr.19: 38304256-38304256 on Build GRCh38
rs764807460 Chr.19: 38304379-38304379 on Build GRCh38
rs770417120 Chr.19: 38304440-38304440 on Build GRCh38
rs547223647 Chr.19: 38304083-38304083 on Build GRCh38
rs768625548 Chr.19: 38304306-38304306 on Build GRCh38
rs771799308 Chr.19: 38304251-38304251 on Build GRCh38

View Full Product Details