Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 38405240-38405742 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM98C
Entrez Gene ID
147965
Gene Name
family with sequence similarity 98 member C
Gene Aliases
-
UniGene
Hs.355162
Gene Chromosome Location
Chr.19: 38403124-38409088 on Build GRCh38
Transcripts
NM_174905.3 XM_017026354.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5761252 c.665G>T p.R222I Chr.19: 38405550-38405550 on Build GRCh38
7318278 c.575C>T p.S192F Chr.19: 38405363-38405363 on Build GRCh38
5855646 c.580C>T p.P194S Chr.19: 38405368-38405368 on Build GRCh38
7367568 c.679T>G p.C227G Chr.19: 38405564-38405564 on Build GRCh38
328442 c.674A>C p.Y225S Chr.19: 38405559-38405559 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762787816 Chr.19: 38405614-38405614 on Build GRCh38
rs372620728 Chr.19: 38405507-38405507 on Build GRCh38
rs747255664 Chr.19: 38405581-38405581 on Build GRCh38
rs374960953 Chr.19: 38405636-38405636 on Build GRCh38
rs768502589 Chr.19: 38405368-38405368 on Build GRCh38
rs372012533 Chr.19: 38405627-38405627 on Build GRCh38
rs139841886 Chr.19: 38405378-38405378 on Build GRCh38
rs200721594 Chr.19: 38405628-38405628 on Build GRCh38
rs374391516 Chr.19: 38405436-38405436 on Build GRCh38
rs3745962 Chr.19: 38405604-38405604 on Build GRCh38
rs201418056 Chr.19: 38405371-38405371 on Build GRCh38
rs376152986 Chr.19: 38405403-38405403 on Build GRCh38
rs370418898 Chr.19: 38405506-38405506 on Build GRCh38
rs75686841 Chr.19: 38405419-38405419 on Build GRCh38
rs980278685 Chr.19: 38405398-38405398 on Build GRCh38
rs755936002 Chr.19: 38405298-38405298 on Build GRCh38
rs542705240 Chr.19: 38405528-38405528 on Build GRCh38
rs748537969 Chr.19: 38405571-38405571 on Build GRCh38
rs749730784 Chr.19: 38405567-38405567 on Build GRCh38
rs376752317 Chr.19: 38405666-38405666 on Build GRCh38
rs765216485 Chr.19: 38405591-38405591 on Build GRCh38
rs772301573 Chr.19: 38405571-38405572 on Build GRCh38
rs778310972 Chr.19: 38405667-38405667 on Build GRCh38
rs774787024 Chr.19: 38405453-38405453 on Build GRCh38
rs73044953 Chr.19: 38405435-38405435 on Build GRCh38
rs760803944 Chr.19: 38405484-38405484 on Build GRCh38
rs928999588 Chr.19: 38405611-38405611 on Build GRCh38
rs367904601 Chr.19: 38405411-38405411 on Build GRCh38
rs774390061 Chr.19: 38405308-38405308 on Build GRCh38
rs534837618 Chr.19: 38405588-38405588 on Build GRCh38
rs749065738 Chr.19: 38405335-38405336 on Build GRCh38
rs376864098 Chr.19: 38405568-38405568 on Build GRCh38
rs3745963 Chr.19: 38405655-38405655 on Build GRCh38
rs373033689 Chr.19: 38405474-38405474 on Build GRCh38
rs888913037 Chr.19: 38405337-38405337 on Build GRCh38
rs779342574 Chr.19: 38405570-38405570 on Build GRCh38
rs371505017 Chr.19: 38405658-38405658 on Build GRCh38
rs771291087 Chr.19: 38405676-38405676 on Build GRCh38
rs747159817 Chr.19: 38405670-38405670 on Build GRCh38
rs536032697 Chr.19: 38405641-38405641 on Build GRCh38
rs571759281 Chr.19: 38405328-38405330 on Build GRCh38
rs745408880 Chr.19: 38405297-38405298 on Build GRCh38
rs564852306 Chr.19: 38405271-38405271 on Build GRCh38
rs137896913 Chr.19: 38405575-38405577 on Build GRCh38
rs375645286 Chr.19: 38405519-38405519 on Build GRCh38
rs763275463 Chr.19: 38405297-38405297 on Build GRCh38
rs758194490 Chr.19: 38405540-38405540 on Build GRCh38
rs748417734 Chr.19: 38405568-38405569 on Build GRCh38
rs117354953 Chr.19: 38405564-38405564 on Build GRCh38
rs886461762 Chr.19: 38405717-38405717 on Build GRCh38
rs945961967 Chr.19: 38405418-38405418 on Build GRCh38
rs763901480 Chr.19: 38405616-38405616 on Build GRCh38
rs370978130 Chr.19: 38405300-38405300 on Build GRCh38
rs1023227766 Chr.19: 38405523-38405523 on Build GRCh38
rs754796630 Chr.19: 38405383-38405383 on Build GRCh38
rs369832859 Chr.19: 38405460-38405460 on Build GRCh38
rs922663557 Chr.19: 38405273-38405273 on Build GRCh38
rs956117880 Chr.19: 38405557-38405557 on Build GRCh38
rs753863576 Chr.19: 38405374-38405374 on Build GRCh38
rs547434902 Chr.19: 38405370-38405370 on Build GRCh38
rs375777338 Chr.19: 38405459-38405459 on Build GRCh38
rs978884516 Chr.19: 38405530-38405530 on Build GRCh38
rs751263079 Chr.19: 38405561-38405561 on Build GRCh38
rs755643095 Chr.19: 38405650-38405650 on Build GRCh38
rs377183116 Chr.19: 38405492-38405492 on Build GRCh38
rs752252206 Chr.19: 38405388-38405388 on Build GRCh38
rs374364012 Chr.19: 38405319-38405319 on Build GRCh38
rs76199147 Chr.19: 38405559-38405559 on Build GRCh38
rs371827845 Chr.19: 38405413-38405413 on Build GRCh38
rs759334559 Chr.19: 38405590-38405590 on Build GRCh38
rs772321000 Chr.19: 38405473-38405473 on Build GRCh38
rs774524855 Chr.19: 38405429-38405430 on Build GRCh38
rs200340957 Chr.19: 38405576-38405576 on Build GRCh38
rs756709692 Chr.19: 38405562-38405562 on Build GRCh38
rs374261710 Chr.19: 38405589-38405589 on Build GRCh38
rs750540471 Chr.19: 38405348-38405348 on Build GRCh38
rs200150470 Chr.19: 38405695-38405695 on Build GRCh38
rs192772592 Chr.19: 38405525-38405525 on Build GRCh38
rs777374482 Chr.19: 38405555-38405555 on Build GRCh38
rs778788736 Chr.19: 38405384-38405384 on Build GRCh38
rs370061836 Chr.19: 38405524-38405524 on Build GRCh38
rs762131282 Chr.19: 38405313-38405313 on Build GRCh38
rs201635019 Chr.19: 38405402-38405402 on Build GRCh38
rs368773270 Chr.19: 38405287-38405287 on Build GRCh38
rs753779056 Chr.19: 38405503-38405503 on Build GRCh38
rs148104841 Chr.19: 38405704-38405704 on Build GRCh38
rs371195895 Chr.19: 38405580-38405580 on Build GRCh38

View Full Product Details