Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 38717686-38718203 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACTN4
Entrez Gene ID
81
Gene Name
actinin alpha 4
Gene Aliases
ACTININ-4, FSGS, FSGS1
UniGene
Hs.270291
Gene Chromosome Location
Chr.19: 38647616-38730532 on Build GRCh38
Transcripts
NM_001322033.1 NM_004924.5 XM_005259281.4 XM_006723406.2 XM_017027331.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6408565 c.1228C>T p.R410W Chr.19: 38718011-38718011 on Build GRCh38
3797049 c.1206G>A p.L402L Chr.19: 38717989-38717989 on Build GRCh38
4880942 c.1194G>A p.E398E Chr.19: 38717977-38717977 on Build GRCh38
7212870 c.1260G>C p.Q420H Chr.19: 38718043-38718043 on Build GRCh38
1612194 c.1154A>T p.N385I Chr.19: 38717937-38717937 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764895145 Chr.19: 38717885-38717885 on Build GRCh38
rs779211830 Chr.19: 38718058-38718058 on Build GRCh38
rs7248577 Chr.19: 38718155-38718155 on Build GRCh38
rs765746867 Chr.19: 38717791-38717791 on Build GRCh38
rs748358981 Chr.19: 38717949-38717949 on Build GRCh38
rs1035213561 Chr.19: 38718173-38718173 on Build GRCh38
rs774297184 Chr.19: 38717879-38717880 on Build GRCh38
rs763579928 Chr.19: 38717909-38717909 on Build GRCh38
rs745861958 Chr.19: 38718081-38718081 on Build GRCh38
rs762573515 Chr.19: 38718000-38718000 on Build GRCh38
rs578211163 Chr.19: 38718110-38718110 on Build GRCh38
rs879081225 Chr.19: 38717925-38717925 on Build GRCh38
rs113496871 Chr.19: 38717849-38717849 on Build GRCh38
rs879204175 Chr.19: 38717924-38717924 on Build GRCh38
rs749109401 Chr.19: 38718102-38718102 on Build GRCh38
rs773146645 Chr.19: 38718085-38718086 on Build GRCh38
rs776733169 Chr.19: 38717974-38717974 on Build GRCh38
rs996803518 Chr.19: 38717760-38717760 on Build GRCh38
rs201128110 Chr.19: 38718062-38718062 on Build GRCh38
rs761287376 Chr.19: 38718114-38718114 on Build GRCh38
rs944837268 Chr.19: 38717717-38717717 on Build GRCh38
rs151102337 Chr.19: 38718153-38718153 on Build GRCh38
rs767806046 Chr.19: 38717919-38717920 on Build GRCh38
rs951805786 Chr.19: 38717795-38717795 on Build GRCh38
rs1014635236 Chr.19: 38717864-38717864 on Build GRCh38
rs113002708 Chr.19: 38717714-38717714 on Build GRCh38
rs199711892 Chr.19: 38718122-38718122 on Build GRCh38
rs760851285 Chr.19: 38718086-38718095 on Build GRCh38
rs780542075 Chr.19: 38717915-38717915 on Build GRCh38
rs750099000 Chr.19: 38717724-38717724 on Build GRCh38
rs758730164 Chr.19: 38718091-38718091 on Build GRCh38
rs944763331 Chr.19: 38718077-38718077 on Build GRCh38
rs545698094 Chr.19: 38718115-38718115 on Build GRCh38
rs1027917238 Chr.19: 38717784-38717784 on Build GRCh38
rs201704771 Chr.19: 38718078-38718078 on Build GRCh38
rs200541372 Chr.19: 38718116-38718116 on Build GRCh38
rs143174736 Chr.19: 38717957-38717957 on Build GRCh38
rs781248275 Chr.19: 38718079-38718079 on Build GRCh38
rs572602792 Chr.19: 38718121-38718121 on Build GRCh38
rs777619106 Chr.19: 38718046-38718046 on Build GRCh38
rs377082312 Chr.19: 38718084-38718084 on Build GRCh38
rs202151136 Chr.19: 38718024-38718024 on Build GRCh38
rs755318100 Chr.19: 38717932-38717932 on Build GRCh38
rs754390440 Chr.19: 38718019-38718019 on Build GRCh38
rs368725797 Chr.19: 38717999-38717999 on Build GRCh38
rs371340759 Chr.19: 38717887-38717887 on Build GRCh38
rs114750273 Chr.19: 38718109-38718109 on Build GRCh38
rs766819959 Chr.19: 38718018-38718018 on Build GRCh38
rs375645155 Chr.19: 38717956-38717956 on Build GRCh38
rs760458644 Chr.19: 38717771-38717771 on Build GRCh38
rs1006148354 Chr.19: 38717829-38717829 on Build GRCh38
rs1004551127 Chr.19: 38717814-38717814 on Build GRCh38
rs561382762 Chr.19: 38718113-38718113 on Build GRCh38
rs1003001316 Chr.19: 38718159-38718159 on Build GRCh38
rs752949932 Chr.19: 38718059-38718059 on Build GRCh38
rs761905639 Chr.19: 38717899-38717900 on Build GRCh38
rs142111133 Chr.19: 38718073-38718073 on Build GRCh38
rs969616071 Chr.19: 38717848-38717848 on Build GRCh38
rs751000446 Chr.19: 38718011-38718011 on Build GRCh38
rs573020950 Chr.19: 38717916-38717916 on Build GRCh38
rs988264051 Chr.19: 38717978-38717978 on Build GRCh38
rs374458190 Chr.19: 38717960-38717960 on Build GRCh38
rs760019463 Chr.19: 38718119-38718119 on Build GRCh38
rs759384679 Chr.19: 38717975-38717975 on Build GRCh38
rs779173708 Chr.19: 38717937-38717937 on Build GRCh38
rs887402879 Chr.19: 38717802-38717802 on Build GRCh38
rs762497545 Chr.19: 38717892-38717892 on Build GRCh38
rs900688156 Chr.19: 38717729-38717729 on Build GRCh38
rs565299215 Chr.19: 38718168-38718168 on Build GRCh38
rs78679563 Chr.19: 38717987-38717987 on Build GRCh38
rs550226120 Chr.19: 38718162-38718162 on Build GRCh38
rs148259739 Chr.19: 38718001-38718001 on Build GRCh38
rs755337601 Chr.19: 38718052-38718052 on Build GRCh38
rs533607295 Chr.19: 38717913-38717913 on Build GRCh38
rs1043958961 Chr.19: 38718043-38718043 on Build GRCh38
rs761342349 Chr.19: 38718017-38718017 on Build GRCh38
rs1017071657 Chr.19: 38717842-38717842 on Build GRCh38
rs1040924091 Chr.19: 38717726-38717726 on Build GRCh38
rs772186055 Chr.19: 38717950-38717950 on Build GRCh38
rs747039049 Chr.19: 38718074-38718074 on Build GRCh38
rs533052586 Chr.19: 38718172-38718172 on Build GRCh38
rs766868950 Chr.19: 38718109-38718110 on Build GRCh38
rs750919242 Chr.19: 38717912-38717912 on Build GRCh38
rs1026465327 Chr.19: 38717749-38717749 on Build GRCh38

View Full Product Details