Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 38802178-38802683 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LGALS4
Entrez Gene ID
3960
Gene Name
galectin 4
Gene Aliases
GAL4, L36LBP
UniGene
Hs.5302
Gene Chromosome Location
Chr.19: 38801671-38813544 on Build GRCh38
Transcripts
NM_006149.3 XM_011526973.1 XM_011526974.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6700682 c.595C>T p.Q199* Chr.19: 38802380-38802380 on Build GRCh38
3533566 c.585C>T p.F195F Chr.19: 38802390-38802390 on Build GRCh38
996146 c.610G>A p.A204T Chr.19: 38802365-38802365 on Build GRCh38
4432511 c.578C>G p.P193R Chr.19: 38802397-38802397 on Build GRCh38
1256418 c.656A>G p.K219R Chr.19: 38802319-38802319 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs377023305 Chr.19: 38802290-38802290 on Build GRCh38
rs760235320 Chr.19: 38802361-38802361 on Build GRCh38
rs369591090 Chr.19: 38802292-38802292 on Build GRCh38
rs1050311445 Chr.19: 38802639-38802639 on Build GRCh38
rs747262021 Chr.19: 38802304-38802304 on Build GRCh38
rs754906531 Chr.19: 38802316-38802317 on Build GRCh38
rs772928274 Chr.19: 38802439-38802439 on Build GRCh38
rs1007060605 Chr.19: 38802537-38802537 on Build GRCh38
rs1044916911 Chr.19: 38802336-38802336 on Build GRCh38
rs538737243 Chr.19: 38802354-38802354 on Build GRCh38
rs898539078 Chr.19: 38802651-38802651 on Build GRCh38
rs1026039929 Chr.19: 38802561-38802561 on Build GRCh38
rs148155288 Chr.19: 38802640-38802640 on Build GRCh38
rs772662808 Chr.19: 38802355-38802355 on Build GRCh38
rs760052233 Chr.19: 38802441-38802441 on Build GRCh38
rs768461954 Chr.19: 38802331-38802331 on Build GRCh38
rs758800866 Chr.19: 38802299-38802299 on Build GRCh38
rs767046687 Chr.19: 38802283-38802283 on Build GRCh38
rs965491312 Chr.19: 38802257-38802257 on Build GRCh38
rs763670773 Chr.19: 38802199-38802199 on Build GRCh38
rs771645084 Chr.19: 38802425-38802425 on Build GRCh38
rs750406604 Chr.19: 38802408-38802408 on Build GRCh38
rs149018638 Chr.19: 38802536-38802536 on Build GRCh38
rs774760271 Chr.19: 38802390-38802390 on Build GRCh38
rs183332836 Chr.19: 38802367-38802367 on Build GRCh38
rs111291316 Chr.19: 38802489-38802489 on Build GRCh38
rs901640137 Chr.19: 38802620-38802620 on Build GRCh38
rs757385638 Chr.19: 38802308-38802308 on Build GRCh38
rs948008857 Chr.19: 38802615-38802615 on Build GRCh38
rs778088078 Chr.19: 38802300-38802300 on Build GRCh38
rs778736683 Chr.19: 38802423-38802423 on Build GRCh38
rs747791964 Chr.19: 38802424-38802424 on Build GRCh38
rs773953511 Chr.19: 38802334-38802334 on Build GRCh38
rs976855966 Chr.19: 38802555-38802555 on Build GRCh38
rs933181579 Chr.19: 38802638-38802638 on Build GRCh38
rs761501851 Chr.19: 38802337-38802337 on Build GRCh38
rs1018487378 Chr.19: 38802540-38802540 on Build GRCh38
rs774008071 Chr.19: 38802200-38802200 on Build GRCh38
rs577868923 Chr.19: 38802328-38802328 on Build GRCh38
rs751812907 Chr.19: 38802389-38802389 on Build GRCh38
rs767001617 Chr.19: 38802339-38802339 on Build GRCh38
rs924982770 Chr.19: 38802520-38802520 on Build GRCh38
rs201990706 Chr.19: 38802419-38802419 on Build GRCh38
rs187364688 Chr.19: 38802411-38802411 on Build GRCh38
rs769690163 Chr.19: 38802321-38802321 on Build GRCh38
rs984408184 Chr.19: 38802571-38802571 on Build GRCh38
rs367948809 Chr.19: 38802282-38802282 on Build GRCh38
rs749280227 Chr.19: 38802420-38802420 on Build GRCh38
rs12461466 Chr.19: 38802655-38802655 on Build GRCh38
rs951423560 Chr.19: 38802562-38802562 on Build GRCh38
rs375729824 Chr.19: 38802383-38802383 on Build GRCh38
rs1038373650 Chr.19: 38802642-38802642 on Build GRCh38
rs965438707 Chr.19: 38802548-38802548 on Build GRCh38
rs781330354 Chr.19: 38802397-38802397 on Build GRCh38
rs372115489 Chr.19: 38802285-38802285 on Build GRCh38
rs770501629 Chr.19: 38802442-38802442 on Build GRCh38
rs191759909 Chr.19: 38802484-38802484 on Build GRCh38
rs935071846 Chr.19: 38802591-38802591 on Build GRCh38
rs556747233 Chr.19: 38802419-38802420 on Build GRCh38
rs745807083 Chr.19: 38802315-38802315 on Build GRCh38
rs143812403 Chr.19: 38802619-38802619 on Build GRCh38
rs533231798 Chr.19: 38802653-38802653 on Build GRCh38
rs753266881 Chr.19: 38802372-38802372 on Build GRCh38
rs1006173539 Chr.19: 38802641-38802641 on Build GRCh38
rs765629585 Chr.19: 38802289-38802289 on Build GRCh38
rs561257319 Chr.19: 38802527-38802527 on Build GRCh38
rs190785460 Chr.19: 38802309-38802309 on Build GRCh38
rs758791990 Chr.19: 38802380-38802380 on Build GRCh38
rs775502985 Chr.19: 38802324-38802324 on Build GRCh38
rs535825484 Chr.19: 38802573-38802573 on Build GRCh38

View Full Product Details