Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 38930771-38931265 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MRPS12
Entrez Gene ID
6183
Gene Name
mitochondrial ribosomal protein S12
Gene Aliases
MPR-S12, MT-RPS12, RPMS12, RPS12, RPSM12
UniGene
Hs.411125
Gene Chromosome Location
Chr.19: 38930708-38933020 on Build GRCh38
Transcripts
NM_021107.1 NM_033362.3 NM_033363.1 NM_001145901.1 NM_017827.3

Gene Symbol
SARS2
Entrez Gene ID
54938
Gene Name
seryl-tRNA synthetase 2, mitochondrial
Gene Aliases
SARS, SARSM, SERS, SYS, SerRS, SerRSmt, mtSerRS
UniGene
Hs.709416
Gene Chromosome Location
Chr.19: 38915264-38930896 on Build GRCh38
Transcripts
NM_021107.1 NM_033362.3 NM_033363.1 NM_001145901.1 NM_017827.3

SNP Detail

SNP ID Chromosome Location
rs897312343 Chr.19: 38931052-38931052 on Build GRCh38
rs376123983 Chr.19: 38930905-38930905 on Build GRCh38
rs1027570829 Chr.19: 38931079-38931079 on Build GRCh38
rs956842311 Chr.19: 38931118-38931118 on Build GRCh38
rs562390907 Chr.19: 38930986-38930986 on Build GRCh38
rs907057358 Chr.19: 38931002-38931002 on Build GRCh38
rs548918324 Chr.19: 38931102-38931102 on Build GRCh38
rs527313094 Chr.19: 38931092-38931092 on Build GRCh38
rs931735065 Chr.19: 38930978-38930978 on Build GRCh38
rs935219369 Chr.19: 38930940-38930940 on Build GRCh38
rs1049171682 Chr.19: 38930971-38930971 on Build GRCh38
rs149607876 Chr.19: 38930864-38930864 on Build GRCh38
rs911539281 Chr.19: 38931190-38931190 on Build GRCh38
rs555376160 Chr.19: 38930865-38930865 on Build GRCh38
rs931702269 Chr.19: 38931164-38931164 on Build GRCh38
rs966220942 Chr.19: 38931121-38931121 on Build GRCh38
rs978167554 Chr.19: 38930826-38930826 on Build GRCh38
rs2304116 Chr.19: 38931180-38931180 on Build GRCh38
rs931670361 Chr.19: 38930962-38930962 on Build GRCh38
rs550472130 Chr.19: 38931112-38931112 on Build GRCh38
rs183703431 Chr.19: 38931149-38931149 on Build GRCh38
rs887444814 Chr.19: 38930973-38930973 on Build GRCh38
rs767335734 Chr.19: 38930834-38930834 on Build GRCh38
rs143403017 Chr.19: 38930991-38930991 on Build GRCh38
rs1036056354 Chr.19: 38930993-38930993 on Build GRCh38
rs188275221 Chr.19: 38931156-38931156 on Build GRCh38
rs922216853 Chr.19: 38931140-38931140 on Build GRCh38
rs752903214 Chr.19: 38930975-38930975 on Build GRCh38
rs544092057 Chr.19: 38930982-38930982 on Build GRCh38
rs944413914 Chr.19: 38931208-38931208 on Build GRCh38
rs554221329 Chr.19: 38930842-38930842 on Build GRCh38
rs16972885 Chr.19: 38931111-38931111 on Build GRCh38
rs1041303014 Chr.19: 38931037-38931037 on Build GRCh38
rs970607011 Chr.19: 38930850-38930850 on Build GRCh38
rs922156988 Chr.19: 38930936-38930936 on Build GRCh38
rs11541303 Chr.19: 38931143-38931143 on Build GRCh38
rs1053939481 Chr.19: 38930947-38930947 on Build GRCh38
rs112064258 Chr.19: 38931051-38931051 on Build GRCh38
rs1022383626 Chr.19: 38930807-38930807 on Build GRCh38
rs976211767 Chr.19: 38931138-38931138 on Build GRCh38
rs530416008 Chr.19: 38930904-38930904 on Build GRCh38
rs952804268 Chr.19: 38931094-38931094 on Build GRCh38
rs546791254 Chr.19: 38931114-38931114 on Build GRCh38
rs765655307 Chr.19: 38930963-38930963 on Build GRCh38
rs1008419250 Chr.19: 38931022-38931022 on Build GRCh38
rs868829713 Chr.19: 38930857-38930857 on Build GRCh38
rs758146225 Chr.19: 38931068-38931068 on Build GRCh38
rs572364406 Chr.19: 38930847-38930847 on Build GRCh38
rs991187627 Chr.19: 38930791-38930791 on Build GRCh38
rs968570868 Chr.19: 38930815-38930815 on Build GRCh38
rs970744168 Chr.19: 38931107-38931107 on Build GRCh38
rs368918274 Chr.19: 38931072-38931072 on Build GRCh38
rs867137023 Chr.19: 38930837-38930837 on Build GRCh38
rs561435464 Chr.19: 38931103-38931103 on Build GRCh38
rs571901745 Chr.19: 38931126-38931126 on Build GRCh38
rs1002637535 Chr.19: 38931034-38931034 on Build GRCh38
rs763284698 Chr.19: 38930980-38930980 on Build GRCh38
rs2304115 Chr.19: 38931064-38931064 on Build GRCh38

View Full Product Details