Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 39417083-39417624 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PLEKHG2
Entrez Gene ID
64857
Gene Name
pleckstrin homology and RhoGEF domain containing G2
Gene Aliases
ARHGEF42, CLG, LDAMD
UniGene
Hs.631574
Gene Chromosome Location
Chr.19: 39412582-39428415 on Build GRCh38
Transcripts
NM_022835.2 XM_005259163.2 XM_006723334.2 XM_011527232.2 XM_017027150.1 XM_017027151.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3404215 c.807T>C p.A269A Chr.19: 39417617-39417617 on Build GRCh38
4753348 c.790G>A p.E264K Chr.19: 39417600-39417600 on Build GRCh38
233124 c.644G>A p.G215D Chr.19: 39417580-39417580 on Build GRCh38
3404216 c.681T>C p.A227A Chr.19: 39417617-39417617 on Build GRCh38
233125 c.770G>A p.G257D Chr.19: 39417580-39417580 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1032056852 Chr.19: 39417396-39417396 on Build GRCh38
rs991149695 Chr.19: 39417271-39417271 on Build GRCh38
rs1035317286 Chr.19: 39417402-39417402 on Build GRCh38
rs62119733 Chr.19: 39417212-39417212 on Build GRCh38
rs189775046 Chr.19: 39417517-39417517 on Build GRCh38
rs778253026 Chr.19: 39417522-39417522 on Build GRCh38
rs866389625 Chr.19: 39417533-39417533 on Build GRCh38
rs958114134 Chr.19: 39417163-39417163 on Build GRCh38
rs972405519 Chr.19: 39417524-39417524 on Build GRCh38
rs10402009 Chr.19: 39417141-39417141 on Build GRCh38
rs948809592 Chr.19: 39417344-39417344 on Build GRCh38
rs985157629 Chr.19: 39417272-39417272 on Build GRCh38
rs192920790 Chr.19: 39417235-39417235 on Build GRCh38
rs190105085 Chr.19: 39417187-39417187 on Build GRCh38
rs545113383 Chr.19: 39417243-39417243 on Build GRCh38
rs985564875 Chr.19: 39417508-39417508 on Build GRCh38
rs939257075 Chr.19: 39417335-39417335 on Build GRCh38
rs931016099 Chr.19: 39417298-39417298 on Build GRCh38
rs551011343 Chr.19: 39417361-39417361 on Build GRCh38
rs915550761 Chr.19: 39417505-39417505 on Build GRCh38
rs954342214 Chr.19: 39417498-39417498 on Build GRCh38
rs947884659 Chr.19: 39417580-39417580 on Build GRCh38
rs760399453 Chr.19: 39417593-39417593 on Build GRCh38
rs770114667 Chr.19: 39417226-39417226 on Build GRCh38
rs959571936 Chr.19: 39417491-39417491 on Build GRCh38
rs771936993 Chr.19: 39417579-39417579 on Build GRCh38
rs1031945467 Chr.19: 39417186-39417186 on Build GRCh38
rs386809184 Chr.19: 39417211-39417212 on Build GRCh38
rs893902285 Chr.19: 39417342-39417342 on Build GRCh38
rs758801242 Chr.19: 39417521-39417521 on Build GRCh38
rs142309421 Chr.19: 39417575-39417575 on Build GRCh38
rs972350007 Chr.19: 39417316-39417316 on Build GRCh38
rs753270064 Chr.19: 39417597-39417597 on Build GRCh38
rs1000198214 Chr.19: 39417373-39417373 on Build GRCh38
rs535113841 Chr.19: 39417483-39417483 on Build GRCh38
rs1022995965 Chr.19: 39417138-39417138 on Build GRCh38
rs555803584 Chr.19: 39417215-39417215 on Build GRCh38
rs966928176 Chr.19: 39417382-39417382 on Build GRCh38
rs370859038 Chr.19: 39417519-39417519 on Build GRCh38
rs918517535 Chr.19: 39417331-39417331 on Build GRCh38
rs145127766 Chr.19: 39417211-39417211 on Build GRCh38
rs766028297 Chr.19: 39417595-39417595 on Build GRCh38
rs761517117 Chr.19: 39417577-39417577 on Build GRCh38
rs111240744 Chr.19: 39417154-39417154 on Build GRCh38
rs201109394 Chr.19: 39417389-39417389 on Build GRCh38
rs773058426 Chr.19: 39417589-39417589 on Build GRCh38
rs10407472 Chr.19: 39417354-39417354 on Build GRCh38
rs73033371 Chr.19: 39417574-39417574 on Build GRCh38
rs1056337609 Chr.19: 39417336-39417336 on Build GRCh38
rs574414583 Chr.19: 39417192-39417192 on Build GRCh38
rs551887768 Chr.19: 39417444-39417444 on Build GRCh38
rs765976195 Chr.19: 39417510-39417510 on Build GRCh38
rs769924375 Chr.19: 39417541-39417541 on Build GRCh38
rs140586164 Chr.19: 39417297-39417297 on Build GRCh38
rs566945309 Chr.19: 39417445-39417445 on Build GRCh38
rs1029856493 Chr.19: 39417451-39417451 on Build GRCh38
rs757581848 Chr.19: 39417527-39417527 on Build GRCh38
rs555857622 Chr.19: 39417339-39417339 on Build GRCh38
rs892083110 Chr.19: 39417408-39417408 on Build GRCh38
rs749296655 Chr.19: 39417570-39417570 on Build GRCh38
rs534454174 Chr.19: 39417509-39417509 on Build GRCh38
rs199928152 Chr.19: 39417537-39417537 on Build GRCh38
rs779962934 Chr.19: 39417550-39417550 on Build GRCh38
rs147254507 Chr.19: 39417255-39417255 on Build GRCh38
rs764543187 Chr.19: 39417600-39417600 on Build GRCh38
rs1028400176 Chr.19: 39417152-39417152 on Build GRCh38
rs573301051 Chr.19: 39417194-39417194 on Build GRCh38
rs552397391 Chr.19: 39417129-39417129 on Build GRCh38
rs184690615 Chr.19: 39417368-39417368 on Build GRCh38
rs759041640 Chr.19: 39417599-39417599 on Build GRCh38
rs753313783 Chr.19: 39417518-39417518 on Build GRCh38
rs35361532 Chr.19: 39417303-39417304 on Build GRCh38
rs751894646 Chr.19: 39417523-39417523 on Build GRCh38
rs998843227 Chr.19: 39417423-39417423 on Build GRCh38
rs781402554 Chr.19: 39417528-39417528 on Build GRCh38
rs976957003 Chr.19: 39417216-39417216 on Build GRCh38
rs62119734 Chr.19: 39417306-39417306 on Build GRCh38
rs968690488 Chr.19: 39417140-39417140 on Build GRCh38
rs939306486 Chr.19: 39417536-39417536 on Build GRCh38
rs1028243272 Chr.19: 39417341-39417341 on Build GRCh38
rs1048714722 Chr.19: 39417303-39417303 on Build GRCh38

View Full Product Details