Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 46267374-46267876 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RAD54L
Entrez Gene ID
8438
Gene Name
RAD54-like (S. cerevisiae)
Gene Aliases
HR54, RAD54A, hHR54, hRAD54
UniGene
Hs.642042
Gene Chromosome Location
Chr.1: 46247695-46278477 on Build GRCh38
Transcripts
NM_001142548.1 NM_003579.3 XM_006710975.3 XM_011542299.2 XM_011542300.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6761966 c.991G>T p.D331Y Chr.1: 46267558-46267558 on Build GRCh38
5485711 c.1034G>A p.G345D Chr.1: 46267601-46267601 on Build GRCh38
5565570 c.958C>T p.R320W Chr.1: 46267525-46267525 on Build GRCh38
6332230 c.955C>T p.R319W Chr.1: 46267522-46267522 on Build GRCh38
1321018 c.956G>A p.R319Q Chr.1: 46267523-46267523 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761626726 Chr.1: 46267647-46267647 on Build GRCh38
rs369086807 Chr.1: 46267599-46267599 on Build GRCh38
rs754817150 Chr.1: 46267525-46267525 on Build GRCh38
rs760314792 Chr.1: 46267521-46267521 on Build GRCh38
rs1031850002 Chr.1: 46267815-46267815 on Build GRCh38
rs766735139 Chr.1: 46267411-46267411 on Build GRCh38
rs934981237 Chr.1: 46267698-46267698 on Build GRCh38
rs538393510 Chr.1: 46267432-46267432 on Build GRCh38
rs754894596 Chr.1: 46267435-46267435 on Build GRCh38
rs767075563 Chr.1: 46267428-46267428 on Build GRCh38
rs775966419 Chr.1: 46267496-46267496 on Build GRCh38
rs60121285 Chr.1: 46267461-46267461 on Build GRCh38
rs778893165 Chr.1: 46267526-46267526 on Build GRCh38
rs758044000 Chr.1: 46267543-46267543 on Build GRCh38
rs140547125 Chr.1: 46267539-46267539 on Build GRCh38
rs760092954 Chr.1: 46267622-46267622 on Build GRCh38
rs77100721 Chr.1: 46267821-46267821 on Build GRCh38
rs752570658 Chr.1: 46267647-46267647 on Build GRCh38
rs1019686884 Chr.1: 46267502-46267502 on Build GRCh38
rs760041006 Chr.1: 46267413-46267413 on Build GRCh38
rs1012080022 Chr.1: 46267401-46267401 on Build GRCh38
rs763748241 Chr.1: 46267526-46267526 on Build GRCh38
rs763503794 Chr.1: 46267503-46267503 on Build GRCh38
rs372494747 Chr.1: 46267512-46267512 on Build GRCh38
rs777755012 Chr.1: 46267448-46267448 on Build GRCh38
rs772723123 Chr.1: 46267610-46267610 on Build GRCh38
rs753402940 Chr.1: 46267634-46267634 on Build GRCh38
rs776998118 Chr.1: 46267470-46267470 on Build GRCh38
rs758431541 Chr.1: 46267534-46267534 on Build GRCh38
rs766969374 Chr.1: 46267509-46267509 on Build GRCh38
rs756758491 Chr.1: 46267425-46267425 on Build GRCh38
rs777867960 Chr.1: 46267548-46267548 on Build GRCh38
rs759901390 Chr.1: 46267473-46267473 on Build GRCh38
rs1010684864 Chr.1: 46267736-46267736 on Build GRCh38
rs28363230 Chr.1: 46267664-46267664 on Build GRCh38
rs778987555 Chr.1: 46267436-46267436 on Build GRCh38
rs374967930 Chr.1: 46267586-46267586 on Build GRCh38
rs771081819 Chr.1: 46267463-46267463 on Build GRCh38
rs121908690 Chr.1: 46267540-46267540 on Build GRCh38
rs1044907637 Chr.1: 46267792-46267792 on Build GRCh38
rs35561479 Chr.1: 46267601-46267602 on Build GRCh38
rs111469649 Chr.1: 46267809-46267809 on Build GRCh38
rs771125014 Chr.1: 46267562-46267562 on Build GRCh38
rs973635040 Chr.1: 46267554-46267554 on Build GRCh38
rs960282527 Chr.1: 46267838-46267838 on Build GRCh38
rs558185107 Chr.1: 46267712-46267712 on Build GRCh38
rs770214317 Chr.1: 46267484-46267484 on Build GRCh38
rs61457846 Chr.1: 46267687-46267687 on Build GRCh38
rs184907804 Chr.1: 46267438-46267438 on Build GRCh38
rs977680953 Chr.1: 46267666-46267666 on Build GRCh38
rs200157085 Chr.1: 46267415-46267415 on Build GRCh38
rs772255960 Chr.1: 46267443-46267443 on Build GRCh38
rs747062991 Chr.1: 46267549-46267549 on Build GRCh38
rs797044888 Chr.1: 46267600-46267600 on Build GRCh38
rs951373493 Chr.1: 46267452-46267452 on Build GRCh38
rs76138558 Chr.1: 46267826-46267826 on Build GRCh38
rs769051790 Chr.1: 46267609-46267609 on Build GRCh38
rs182039451 Chr.1: 46267412-46267412 on Build GRCh38
rs900244997 Chr.1: 46267707-46267707 on Build GRCh38
rs975533167 Chr.1: 46267561-46267561 on Build GRCh38
rs752660420 Chr.1: 46267533-46267533 on Build GRCh38
rs77851852 Chr.1: 46267807-46267807 on Build GRCh38
rs142375065 Chr.1: 46267522-46267522 on Build GRCh38
rs751006671 Chr.1: 46267416-46267416 on Build GRCh38
rs765982042 Chr.1: 46267625-46267625 on Build GRCh38
rs749047867 Chr.1: 46267434-46267434 on Build GRCh38
rs1056568586 Chr.1: 46267752-46267752 on Build GRCh38
rs781255100 Chr.1: 46267580-46267580 on Build GRCh38
rs1017333291 Chr.1: 46267410-46267410 on Build GRCh38
rs540084016 Chr.1: 46267811-46267811 on Build GRCh38
rs544363923 Chr.1: 46267825-46267825 on Build GRCh38
rs17102091 Chr.1: 46267433-46267433 on Build GRCh38
rs1043600062 Chr.1: 46267798-46267798 on Build GRCh38
rs746152916 Chr.1: 46267583-46267583 on Build GRCh38
rs947809809 Chr.1: 46267791-46267791 on Build GRCh38
rs770108471 Chr.1: 46267584-46267584 on Build GRCh38
rs759311409 Chr.1: 46267638-46267638 on Build GRCh38
rs750056697 Chr.1: 46267431-46267431 on Build GRCh38
rs776360282 Chr.1: 46267592-46267592 on Build GRCh38
rs751637440 Chr.1: 46267646-46267646 on Build GRCh38
rs917673108 Chr.1: 46267786-46267786 on Build GRCh38

View Full Product Details