Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 48385051-48385599 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SPATA6
Entrez Gene ID
54558
Gene Name
spermatogenesis associated 6
Gene Aliases
HASH, SRF-1, SRF1
UniGene
Hs.538103
Gene Chromosome Location
Chr.1: 48258643-48472208 on Build GRCh38
Transcripts
NM_001286238.1 NM_001286239.1 NM_019073.3 XM_006710699.2 XM_006710700.2 XM_006710701.3 XM_011541606.1 XM_011541607.2 XM_011541608.1 XM_011541609.2 XM_017001509.1 XM_017001510.1 XM_017001511.1 XM_017001512.1 XM_017001513.1 XM_017001514.1 XM_017001515.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5638397 c.883G>C p.G295R Chr.1: 48385335-48385335 on Build GRCh38
2191917 c.899A>C p.K300T Chr.1: 48385319-48385319 on Build GRCh38
6574526 c.869-3C>A p.? Chr.1: 48385352-48385352 on Build GRCh38
6413951 c.909G>T p.K303N Chr.1: 48385309-48385309 on Build GRCh38
6063752 c.893G>T p.R298L Chr.1: 48385325-48385325 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs112881124 Chr.1: 48385454-48385454 on Build GRCh38
rs202088180 Chr.1: 48385345-48385345 on Build GRCh38
rs759829617 Chr.1: 48385150-48385153 on Build GRCh38
rs373669831 Chr.1: 48385273-48385273 on Build GRCh38
rs532929847 Chr.1: 48385110-48385110 on Build GRCh38
rs966495344 Chr.1: 48385200-48385200 on Build GRCh38
rs755050886 Chr.1: 48385271-48385271 on Build GRCh38
rs778189058 Chr.1: 48385097-48385097 on Build GRCh38
rs777988281 Chr.1: 48385280-48385280 on Build GRCh38
rs761347576 Chr.1: 48385334-48385334 on Build GRCh38
rs776660332 Chr.1: 48385386-48385386 on Build GRCh38
rs918761945 Chr.1: 48385349-48385349 on Build GRCh38
rs111973193 Chr.1: 48385249-48385249 on Build GRCh38
rs746400286 Chr.1: 48385308-48385308 on Build GRCh38
rs781623819 Chr.1: 48385304-48385304 on Build GRCh38
rs902360870 Chr.1: 48385573-48385573 on Build GRCh38
rs901783735 Chr.1: 48385155-48385155 on Build GRCh38
rs750518838 Chr.1: 48385397-48385397 on Build GRCh38
rs1020759327 Chr.1: 48385177-48385177 on Build GRCh38
rs758408418 Chr.1: 48385399-48385416 on Build GRCh38
rs1048434858 Chr.1: 48385124-48385124 on Build GRCh38
rs759576800 Chr.1: 48385395-48385395 on Build GRCh38
rs200474406 Chr.1: 48385341-48385341 on Build GRCh38
rs766290795 Chr.1: 48385375-48385375 on Build GRCh38
rs771289740 Chr.1: 48385303-48385303 on Build GRCh38
rs138599139 Chr.1: 48385160-48385160 on Build GRCh38
rs1012686457 Chr.1: 48385348-48385348 on Build GRCh38
rs987057942 Chr.1: 48385166-48385166 on Build GRCh38
rs964316522 Chr.1: 48385540-48385540 on Build GRCh38
rs1019858627 Chr.1: 48385530-48385530 on Build GRCh38
rs547056043 Chr.1: 48385495-48385495 on Build GRCh38
rs889792973 Chr.1: 48385128-48385128 on Build GRCh38
rs931436088 Chr.1: 48385354-48385354 on Build GRCh38
rs562916912 Chr.1: 48385325-48385325 on Build GRCh38
rs975601108 Chr.1: 48385241-48385241 on Build GRCh38
rs912850693 Chr.1: 48385203-48385203 on Build GRCh38
rs901154733 Chr.1: 48385451-48385451 on Build GRCh38
rs997409748 Chr.1: 48385494-48385494 on Build GRCh38
rs914313330 Chr.1: 48385247-48385247 on Build GRCh38
rs758698405 Chr.1: 48385274-48385274 on Build GRCh38
rs951448947 Chr.1: 48385431-48385431 on Build GRCh38
rs942840858 Chr.1: 48385213-48385213 on Build GRCh38
rs1042243843 Chr.1: 48385513-48385513 on Build GRCh38
rs771811962 Chr.1: 48385337-48385337 on Build GRCh38
rs773954906 Chr.1: 48385326-48385326 on Build GRCh38
rs770411353 Chr.1: 48385403-48385403 on Build GRCh38
rs765037821 Chr.1: 48385371-48385374 on Build GRCh38
rs990538665 Chr.1: 48385265-48385265 on Build GRCh38
rs879373210 Chr.1: 48385423-48385423 on Build GRCh38
rs1016063199 Chr.1: 48385361-48385361 on Build GRCh38
rs1041361904 Chr.1: 48385275-48385275 on Build GRCh38
rs902853505 Chr.1: 48385298-48385298 on Build GRCh38
rs1041442326 Chr.1: 48385144-48385144 on Build GRCh38
rs1026082669 Chr.1: 48385140-48385140 on Build GRCh38
rs761769311 Chr.1: 48385448-48385449 on Build GRCh38
rs41301259 Chr.1: 48385370-48385370 on Build GRCh38
rs144534669 Chr.1: 48385338-48385338 on Build GRCh38
rs370120461 Chr.1: 48385264-48385264 on Build GRCh38
rs974071516 Chr.1: 48385121-48385121 on Build GRCh38
rs1032055399 Chr.1: 48385221-48385221 on Build GRCh38
rs753830879 Chr.1: 48385261-48385261 on Build GRCh38
rs747474857 Chr.1: 48385294-48385294 on Build GRCh38

View Full Product Details