Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 48533833-48534341 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AGBL4
Entrez Gene ID
84871
Gene Name
ATP/GTP binding protein like 4
Gene Aliases
CCP6
UniGene
Hs.551844
Gene Chromosome Location
Chr.1: 48520157-50024011 on Build GRCh38
Transcripts
NM_001323573.1 NM_001323574.1 NM_001323575.1 NM_032785.3 NR_136623.1 XM_011542308.2 XM_017002595.1 XM_017002597.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5562574 c.1495C>T p.P499S Chr.1: 48534190-48534190 on Build GRCh38
2191951 c.1421C>A p.P474Q Chr.1: 48534264-48534264 on Build GRCh38
6540437 c.1435C>A p.P479T Chr.1: 48534250-48534250 on Build GRCh38
4527419 c.1462G>A p.G488R Chr.1: 48534223-48534223 on Build GRCh38
6460540 c.1413C>T p.Y471Y Chr.1: 48534272-48534272 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765946170 Chr.1: 48534028-48534028 on Build GRCh38
rs558587800 Chr.1: 48533893-48533893 on Build GRCh38
rs774721958 Chr.1: 48534306-48534306 on Build GRCh38
rs748376932 Chr.1: 48534250-48534250 on Build GRCh38
rs912353962 Chr.1: 48534202-48534202 on Build GRCh38
rs906094511 Chr.1: 48534290-48534290 on Build GRCh38
rs937556743 Chr.1: 48534302-48534302 on Build GRCh38
rs577583885 Chr.1: 48533981-48533981 on Build GRCh38
rs1045280598 Chr.1: 48534255-48534255 on Build GRCh38
rs975344380 Chr.1: 48534057-48534057 on Build GRCh38
rs758409874 Chr.1: 48534122-48534122 on Build GRCh38
rs560172694 Chr.1: 48534265-48534265 on Build GRCh38
rs1002089617 Chr.1: 48533970-48533970 on Build GRCh38
rs757274505 Chr.1: 48533947-48533947 on Build GRCh38
rs898713402 Chr.1: 48534279-48534279 on Build GRCh38
rs201412334 Chr.1: 48534220-48534220 on Build GRCh38
rs753500526 Chr.1: 48534037-48534037 on Build GRCh38
rs866654635 Chr.1: 48533953-48533953 on Build GRCh38
rs751591163 Chr.1: 48534182-48534182 on Build GRCh38
rs2798111 Chr.1: 48533883-48533883 on Build GRCh38
rs914527732 Chr.1: 48534072-48534072 on Build GRCh38
rs759276011 Chr.1: 48534061-48534061 on Build GRCh38
rs746913812 Chr.1: 48533911-48533911 on Build GRCh38
rs960390332 Chr.1: 48534041-48534041 on Build GRCh38
rs935400881 Chr.1: 48534167-48534167 on Build GRCh38
rs182998502 Chr.1: 48534092-48534092 on Build GRCh38
rs1043259868 Chr.1: 48533922-48533922 on Build GRCh38
rs1043336788 Chr.1: 48534274-48534274 on Build GRCh38
rs575005147 Chr.1: 48534166-48534166 on Build GRCh38
rs772322423 Chr.1: 48534299-48534299 on Build GRCh38
rs746126835 Chr.1: 48534241-48534241 on Build GRCh38
rs947029939 Chr.1: 48534256-48534256 on Build GRCh38
rs752544723 Chr.1: 48534117-48534117 on Build GRCh38
rs1052717821 Chr.1: 48533876-48533876 on Build GRCh38
rs535734618 Chr.1: 48534108-48534108 on Build GRCh38
rs371932819 Chr.1: 48534229-48534229 on Build GRCh38
rs780618320 Chr.1: 48534259-48534259 on Build GRCh38
rs774502535 Chr.1: 48534246-48534246 on Build GRCh38
rs35734220 Chr.1: 48534057-48534058 on Build GRCh38
rs896465817 Chr.1: 48534235-48534235 on Build GRCh38
rs201944358 Chr.1: 48533882-48533882 on Build GRCh38
rs768750265 Chr.1: 48534236-48534236 on Build GRCh38
rs919820580 Chr.1: 48534151-48534151 on Build GRCh38
rs769268192 Chr.1: 48534305-48534305 on Build GRCh38

View Full Product Details