Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 51077973-51078477 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KLK14
Entrez Gene ID
43847
Gene Name
kallikrein related peptidase 14
Gene Aliases
KLK-L6
UniGene
Hs.283925
Gene Chromosome Location
Chr.19: 51077495-51084210 on Build GRCh38
Transcripts
NM_001311182.1 NM_022046.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4555403 c.653G>A p.G218D Chr.19: 51078158-51078158 on Build GRCh38
6200973 c.614G>A p.G205E Chr.19: 51078149-51078149 on Build GRCh38
6085659 c.697G>A p.V233I Chr.19: 51078066-51078066 on Build GRCh38
3990203 c.608A>G p.D203G Chr.19: 51078155-51078155 on Build GRCh38
1481388 c.659C>G p.S220C Chr.19: 51078152-51078152 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs143444893 Chr.19: 51078008-51078008 on Build GRCh38
rs972605015 Chr.19: 51078305-51078305 on Build GRCh38
rs911058518 Chr.19: 51078330-51078330 on Build GRCh38
rs770282625 Chr.19: 51078165-51078165 on Build GRCh38
rs765719105 Chr.19: 51078196-51078196 on Build GRCh38
rs758361632 Chr.19: 51078132-51078132 on Build GRCh38
rs780194270 Chr.19: 51078134-51078134 on Build GRCh38
rs752977911 Chr.19: 51078076-51078076 on Build GRCh38
rs1052449940 Chr.19: 51078384-51078384 on Build GRCh38
rs191557700 Chr.19: 51078300-51078300 on Build GRCh38
rs760129945 Chr.19: 51078310-51078310 on Build GRCh38
rs1003090618 Chr.19: 51078032-51078032 on Build GRCh38
rs532811876 Chr.19: 51078427-51078427 on Build GRCh38
rs779332929 Chr.19: 51078029-51078029 on Build GRCh38
rs200225995 Chr.19: 51078158-51078158 on Build GRCh38
rs761466695 Chr.19: 51078002-51078002 on Build GRCh38
rs1030653389 Chr.19: 51078387-51078387 on Build GRCh38
rs754050419 Chr.19: 51078024-51078024 on Build GRCh38
rs773910306 Chr.19: 51078170-51078170 on Build GRCh38
rs746854459 Chr.19: 51078142-51078142 on Build GRCh38
rs78227838 Chr.19: 51078227-51078227 on Build GRCh38
rs1016705938 Chr.19: 51078411-51078411 on Build GRCh38
rs769609771 Chr.19: 51078162-51078162 on Build GRCh38
rs752399836 Chr.19: 51078126-51078126 on Build GRCh38
rs894637964 Chr.19: 51078342-51078342 on Build GRCh38
rs750688755 Chr.19: 51078112-51078112 on Build GRCh38
rs955304280 Chr.19: 51078312-51078312 on Build GRCh38
rs201690513 Chr.19: 51078070-51078070 on Build GRCh38
rs928659861 Chr.19: 51078321-51078321 on Build GRCh38
rs1025271980 Chr.19: 51078451-51078451 on Build GRCh38
rs373194700 Chr.19: 51078087-51078087 on Build GRCh38
rs764673364 Chr.19: 51078180-51078180 on Build GRCh38
rs75626146 Chr.19: 51077996-51077996 on Build GRCh38
rs776407764 Chr.19: 51078059-51078059 on Build GRCh38
rs1024453784 Chr.19: 51078022-51078022 on Build GRCh38
rs952976953 Chr.19: 51078369-51078369 on Build GRCh38
rs371041103 Chr.19: 51078107-51078107 on Build GRCh38
rs201296079 Chr.19: 51078155-51078155 on Build GRCh38
rs750551298 Chr.19: 51078209-51078209 on Build GRCh38
rs1023815158 Chr.19: 51078229-51078229 on Build GRCh38
rs764938192 Chr.19: 51078006-51078006 on Build GRCh38
rs200128422 Chr.19: 51078062-51078062 on Build GRCh38
rs1043233413 Chr.19: 51078361-51078361 on Build GRCh38
rs935296942 Chr.19: 51078370-51078370 on Build GRCh38
rs559353638 Chr.19: 51078409-51078409 on Build GRCh38
rs756388582 Chr.19: 51078020-51078020 on Build GRCh38
rs778298496 Chr.19: 51078023-51078023 on Build GRCh38
rs140861882 Chr.19: 51078386-51078386 on Build GRCh38
rs200139723 Chr.19: 51078017-51078017 on Build GRCh38
rs765940858 Chr.19: 51078088-51078088 on Build GRCh38
rs548406316 Chr.19: 51078393-51078393 on Build GRCh38
rs1001680580 Chr.19: 51078410-51078410 on Build GRCh38
rs970173655 Chr.19: 51078247-51078247 on Build GRCh38
rs759023564 Chr.19: 51078176-51078176 on Build GRCh38
rs753090297 Chr.19: 51078092-51078092 on Build GRCh38
rs911267329 Chr.19: 51078149-51078149 on Build GRCh38
rs969849373 Chr.19: 51078039-51078039 on Build GRCh38
rs556767445 Chr.19: 51078240-51078241 on Build GRCh38
rs938919684 Chr.19: 51078327-51078327 on Build GRCh38
rs764761150 Chr.19: 51078101-51078101 on Build GRCh38
rs967704370 Chr.19: 51078193-51078193 on Build GRCh38
rs911043816 Chr.19: 51078243-51078243 on Build GRCh38
rs201803429 Chr.19: 51078046-51078046 on Build GRCh38
rs543605418 Chr.19: 51077994-51077994 on Build GRCh38
rs371317443 Chr.19: 51078040-51078040 on Build GRCh38
rs754897887 Chr.19: 51078148-51078148 on Build GRCh38
rs757006659 Chr.19: 51078146-51078146 on Build GRCh38
rs1052712253 Chr.19: 51078225-51078225 on Build GRCh38
rs753264353 Chr.19: 51078416-51078416 on Build GRCh38
rs1001331207 Chr.19: 51078257-51078257 on Build GRCh38
rs1045638849 Chr.19: 51078400-51078400 on Build GRCh38
rs757545723 Chr.19: 51078026-51078026 on Build GRCh38
rs201317571 Chr.19: 51078111-51078111 on Build GRCh38
rs561910129 Chr.19: 51078332-51078332 on Build GRCh38
rs979573707 Chr.19: 51078354-51078354 on Build GRCh38
rs577964926 Chr.19: 51078184-51078184 on Build GRCh38
rs148061520 Chr.19: 51078259-51078259 on Build GRCh38
rs762227199 Chr.19: 51078189-51078189 on Build GRCh38
rs999769880 Chr.19: 51078212-51078212 on Build GRCh38
rs202109551 Chr.19: 51078164-51078164 on Build GRCh38
rs551789715 Chr.19: 51078050-51078050 on Build GRCh38
rs532898894 Chr.19: 51078018-51078018 on Build GRCh38
rs377295815 Chr.19: 51078093-51078093 on Build GRCh38
rs185904542 Chr.19: 51078228-51078228 on Build GRCh38
rs768277102 Chr.19: 51078058-51078058 on Build GRCh38
rs779355615 Chr.19: 51078068-51078074 on Build GRCh38
rs903572271 Chr.19: 51078365-51078365 on Build GRCh38
rs16988877 Chr.19: 51077997-51077997 on Build GRCh38
rs74700168 Chr.19: 51078448-51078448 on Build GRCh38
rs755396181 Chr.19: 51078062-51078065 on Build GRCh38
rs772892984 Chr.19: 51078073-51078073 on Build GRCh38
rs987001911 Chr.19: 51078314-51078314 on Build GRCh38
rs933848667 Chr.19: 51078136-51078136 on Build GRCh38
rs570300966 Chr.19: 51078069-51078069 on Build GRCh38
rs746987037 Chr.19: 51078052-51078052 on Build GRCh38
rs930067303 Chr.19: 51078306-51078306 on Build GRCh38

View Full Product Details