Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 51239701-51240210 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD33
Entrez Gene ID
945
Gene Name
CD33 molecule
Gene Aliases
SIGLEC-3, SIGLEC3, p67
UniGene
Hs.83731
Gene Chromosome Location
Chr.19: 51211054-51240019 on Build GRCh38
Transcripts
NM_001082618.1 NM_001772.3 XM_011527531.2 XM_011527532.2 XM_017027508.1 XM_017027509.1 XM_017027510.1

SNP Detail

SNP ID Chromosome Location
rs976155041 Chr.19: 51239742-51239742 on Build GRCh38
rs150956639 Chr.19: 51240144-51240144 on Build GRCh38
rs894722495 Chr.19: 51239782-51239782 on Build GRCh38
rs909503609 Chr.19: 51239804-51239804 on Build GRCh38
rs953433815 Chr.19: 51239768-51239768 on Build GRCh38
rs1036580071 Chr.19: 51239827-51239827 on Build GRCh38
rs186064696 Chr.19: 51239949-51239949 on Build GRCh38
rs534890515 Chr.19: 51239935-51239935 on Build GRCh38
rs867118376 Chr.19: 51239855-51239855 on Build GRCh38
rs1034315375 Chr.19: 51239775-51239775 on Build GRCh38
rs938710329 Chr.19: 51240050-51240050 on Build GRCh38
rs909211621 Chr.19: 51240037-51240037 on Build GRCh38
rs746142392 Chr.19: 51239723-51239723 on Build GRCh38
rs984840908 Chr.19: 51240032-51240032 on Build GRCh38
rs1001232835 Chr.19: 51239912-51239912 on Build GRCh38
rs183239363 Chr.19: 51239915-51239915 on Build GRCh38
rs907282876 Chr.19: 51239736-51239736 on Build GRCh38
rs953582696 Chr.19: 51240020-51240020 on Build GRCh38
rs975729797 Chr.19: 51239906-51239906 on Build GRCh38
rs940931037 Chr.19: 51239818-51239818 on Build GRCh38
rs560472329 Chr.19: 51240156-51240156 on Build GRCh38
rs1803254 Chr.19: 51239889-51239889 on Build GRCh38
rs995750626 Chr.19: 51239807-51239807 on Build GRCh38
rs928711694 Chr.19: 51240005-51240005 on Build GRCh38
rs945941260 Chr.19: 51239988-51239988 on Build GRCh38
rs1051202 Chr.19: 51239774-51239774 on Build GRCh38
rs997291945 Chr.19: 51240014-51240014 on Build GRCh38
rs553496706 Chr.19: 51239944-51239944 on Build GRCh38
rs949563072 Chr.19: 51240179-51240179 on Build GRCh38
rs921991535 Chr.19: 51239766-51239766 on Build GRCh38
rs1055823256 Chr.19: 51240062-51240062 on Build GRCh38
rs1041565067 Chr.19: 51240001-51240001 on Build GRCh38
rs898756069 Chr.19: 51239973-51239986 on Build GRCh38
rs547010454 Chr.19: 51240102-51240102 on Build GRCh38
rs1002908347 Chr.19: 51239742-51239742 on Build GRCh38
rs539077408 Chr.19: 51240004-51240004 on Build GRCh38
rs36110147 Chr.19: 51240136-51240137 on Build GRCh38
rs779275316 Chr.19: 51239722-51239722 on Build GRCh38
rs750538371 Chr.19: 51239968-51239968 on Build GRCh38
rs893088654 Chr.19: 51239917-51239917 on Build GRCh38
rs1004322073 Chr.19: 51239883-51239883 on Build GRCh38
rs1004372956 Chr.19: 51240185-51240185 on Build GRCh38
rs965812040 Chr.19: 51239739-51239739 on Build GRCh38
rs142564497 Chr.19: 51239856-51239856 on Build GRCh38
rs965761852 Chr.19: 51239894-51239894 on Build GRCh38
rs200669561 Chr.19: 51239797-51239797 on Build GRCh38
rs914453054 Chr.19: 51239969-51239969 on Build GRCh38
rs894448192 Chr.19: 51240063-51240063 on Build GRCh38
rs1029057778 Chr.19: 51240015-51240015 on Build GRCh38

View Full Product Details