Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 52075953-52076384 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF841
Entrez Gene ID
284371
Gene Name
zinc finger protein 841
Gene Aliases
-
UniGene
Hs.665751
Gene Chromosome Location
Chr.19: 52050028-52095780 on Build GRCh38
Transcripts
NM_001136499.1 NM_001321349.1 XM_011526833.2 XM_017026652.1 XM_017026653.1 XM_017026654.1 XM_017026655.1 XM_017026656.1 XM_017026657.1 XM_017026658.1 XM_017026659.1 XM_017026660.1 XM_017026661.1 XM_017026662.1 XM_017026663.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1481425 c.175A>C p.M59L Chr.19: 52076140-52076140 on Build GRCh38
5524297 c.180G>A p.L60L Chr.19: 52076135-52076135 on Build GRCh38
5519710 c.143-9delT p.? Chr.19: 52076181-52076181 on Build GRCh38
1161296 c.157G>A p.D53N Chr.19: 52076158-52076158 on Build GRCh38
4824496 c.267C>T p.I89I Chr.19: 52076048-52076048 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775599211 Chr.19: 52076023-52076023 on Build GRCh38
rs985658074 Chr.19: 52076119-52076119 on Build GRCh38
rs769864601 Chr.19: 52076234-52076234 on Build GRCh38
rs533692246 Chr.19: 52076329-52076329 on Build GRCh38
rs571913880 Chr.19: 52075985-52075986 on Build GRCh38
rs1005776687 Chr.19: 52076027-52076027 on Build GRCh38
rs757791475 Chr.19: 52076109-52076109 on Build GRCh38
rs144690937 Chr.19: 52076045-52076045 on Build GRCh38
rs112668310 Chr.19: 52076289-52076289 on Build GRCh38
rs7359836 Chr.19: 52076256-52076256 on Build GRCh38
rs375319177 Chr.19: 52076062-52076062 on Build GRCh38
rs776758972 Chr.19: 52076054-52076054 on Build GRCh38
rs924057398 Chr.19: 52076099-52076099 on Build GRCh38
rs764100349 Chr.19: 52076185-52076188 on Build GRCh38
rs565543298 Chr.19: 52075995-52075995 on Build GRCh38
rs911417067 Chr.19: 52076139-52076139 on Build GRCh38
rs954322957 Chr.19: 52076105-52076105 on Build GRCh38
rs1011423241 Chr.19: 52076279-52076279 on Build GRCh38
rs377051208 Chr.19: 52076194-52076196 on Build GRCh38
rs751182294 Chr.19: 52076087-52076087 on Build GRCh38
rs1002599782 Chr.19: 52076258-52076258 on Build GRCh38
rs759157906 Chr.19: 52076088-52076088 on Build GRCh38
rs1015356568 Chr.19: 52076042-52076042 on Build GRCh38
rs1038631332 Chr.19: 52076197-52076197 on Build GRCh38
rs768622797 Chr.19: 52076053-52076053 on Build GRCh38
rs370201812 Chr.19: 52076015-52076015 on Build GRCh38
rs28706645 Chr.19: 52076064-52076064 on Build GRCh38
rs898189915 Chr.19: 52075993-52075993 on Build GRCh38
rs189490844 Chr.19: 52076220-52076220 on Build GRCh38
rs1051262264 Chr.19: 52076110-52076110 on Build GRCh38
rs750914631 Chr.19: 52076176-52076176 on Build GRCh38
rs958543483 Chr.19: 52076273-52076273 on Build GRCh38
rs779597495 Chr.19: 52076151-52076151 on Build GRCh38
rs185161581 Chr.19: 52076205-52076205 on Build GRCh38
rs766145852 Chr.19: 52076080-52076080 on Build GRCh38
rs757901261 Chr.19: 52076008-52076008 on Build GRCh38
rs758705131 Chr.19: 52076193-52076193 on Build GRCh38
rs773074514 Chr.19: 52076198-52076198 on Build GRCh38
rs755658353 Chr.19: 52076117-52076117 on Build GRCh38
rs563254412 Chr.19: 52076344-52076344 on Build GRCh38
rs1034183452 Chr.19: 52076044-52076044 on Build GRCh38
rs767270365 Chr.19: 52076098-52076098 on Build GRCh38
rs181050237 Chr.19: 52075994-52075994 on Build GRCh38
rs971601927 Chr.19: 52076003-52076003 on Build GRCh38
rs372086918 Chr.19: 52076109-52076109 on Build GRCh38
rs968419219 Chr.19: 52076339-52076340 on Build GRCh38
rs200386268 Chr.19: 52076190-52076192 on Build GRCh38
rs780400965 Chr.19: 52076199-52076199 on Build GRCh38
rs7359993 Chr.19: 52076259-52076259 on Build GRCh38
rs772445467 Chr.19: 52076018-52076018 on Build GRCh38
rs765207314 Chr.19: 52076063-52076063 on Build GRCh38

View Full Product Details