Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 52841422-52841959 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF468
Entrez Gene ID
90333
Gene Name
zinc finger protein 468
Gene Aliases
-
UniGene
Hs.467223
Gene Chromosome Location
Chr.19: 52838532-52857649 on Build GRCh38
Transcripts
NM_001008801.1 NM_001277120.1 NR_102299.1 NR_102300.1 NR_102301.1 XM_017027443.1 XM_017027444.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3835608 c.788G>T p.R263I Chr.19: 52841506-52841506 on Build GRCh38
712960 c.371G>T p.G124V Chr.19: 52841923-52841923 on Build GRCh38
4972414 c.863T>G p.I288S Chr.19: 52841431-52841431 on Build GRCh38
2928074 c.777C>A p.A259A Chr.19: 52841517-52841517 on Build GRCh38
4000888 c.628A>C p.M210L Chr.19: 52841666-52841666 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs368468758 Chr.19: 52841686-52841686 on Build GRCh38
rs768242928 Chr.19: 52841575-52841575 on Build GRCh38
rs368987975 Chr.19: 52841519-52841519 on Build GRCh38
rs140725847 Chr.19: 52841837-52841837 on Build GRCh38
rs745725356 Chr.19: 52841565-52841565 on Build GRCh38
rs142695012 Chr.19: 52841809-52841809 on Build GRCh38
rs755267088 Chr.19: 52841543-52841543 on Build GRCh38
rs757262688 Chr.19: 52841620-52841620 on Build GRCh38
rs767405907 Chr.19: 52841926-52841926 on Build GRCh38
rs150813228 Chr.19: 52841599-52841599 on Build GRCh38
rs952752298 Chr.19: 52841456-52841456 on Build GRCh38
rs756360967 Chr.19: 52841553-52841553 on Build GRCh38
rs527889538 Chr.19: 52841505-52841505 on Build GRCh38
rs139909919 Chr.19: 52841701-52841701 on Build GRCh38
rs10420793 Chr.19: 52841448-52841448 on Build GRCh38
rs762600739 Chr.19: 52841814-52841814 on Build GRCh38
rs142536763 Chr.19: 52841843-52841843 on Build GRCh38
rs770213625 Chr.19: 52841899-52841899 on Build GRCh38
rs780041966 Chr.19: 52841640-52841640 on Build GRCh38
rs61732713 Chr.19: 52841923-52841923 on Build GRCh38
rs748652964 Chr.19: 52841597-52841597 on Build GRCh38
rs748508235 Chr.19: 52841893-52841893 on Build GRCh38
rs776991904 Chr.19: 52841761-52841761 on Build GRCh38
rs765486414 Chr.19: 52841764-52841764 on Build GRCh38
rs753181773 Chr.19: 52841672-52841672 on Build GRCh38
rs766034496 Chr.19: 52841447-52841448 on Build GRCh38
rs373781547 Chr.19: 52841826-52841826 on Build GRCh38
rs772609522 Chr.19: 52841737-52841737 on Build GRCh38
rs781704250 Chr.19: 52841891-52841891 on Build GRCh38
rs112078093 Chr.19: 52841634-52841634 on Build GRCh38
rs769967837 Chr.19: 52841470-52841470 on Build GRCh38
rs746843924 Chr.19: 52841721-52841721 on Build GRCh38
rs770588088 Chr.19: 52841812-52841812 on Build GRCh38
rs753279618 Chr.19: 52841527-52841527 on Build GRCh38
rs980049883 Chr.19: 52841596-52841596 on Build GRCh38
rs769364482 Chr.19: 52841649-52841649 on Build GRCh38
rs746796681 Chr.19: 52841639-52841639 on Build GRCh38
rs768024697 Chr.19: 52841610-52841610 on Build GRCh38
rs766349454 Chr.19: 52841770-52841770 on Build GRCh38
rs58239286 Chr.19: 52841666-52841666 on Build GRCh38
rs767243942 Chr.19: 52841515-52841515 on Build GRCh38
rs961492461 Chr.19: 52841789-52841789 on Build GRCh38
rs775229828 Chr.19: 52841568-52841568 on Build GRCh38
rs139254214 Chr.19: 52841624-52841624 on Build GRCh38
rs773959892 Chr.19: 52841815-52841815 on Build GRCh38
rs557421265 Chr.19: 52841776-52841776 on Build GRCh38
rs747527455 Chr.19: 52841816-52841816 on Build GRCh38
rs371386707 Chr.19: 52841541-52841541 on Build GRCh38
rs888727549 Chr.19: 52841849-52841849 on Build GRCh38
rs776119328 Chr.19: 52841751-52841751 on Build GRCh38
rs766781497 Chr.19: 52841532-52841532 on Build GRCh38
rs375437640 Chr.19: 52841767-52841767 on Build GRCh38
rs747786358 Chr.19: 52841726-52841726 on Build GRCh38
rs149580791 Chr.19: 52841651-52841651 on Build GRCh38
rs770969750 Chr.19: 52841557-52841557 on Build GRCh38
rs546360972 Chr.19: 52841852-52841852 on Build GRCh38
rs61729942 Chr.19: 52841857-52841857 on Build GRCh38
rs760039034 Chr.19: 52841607-52841607 on Build GRCh38
rs748980876 Chr.19: 52841858-52841858 on Build GRCh38
rs768414686 Chr.19: 52841722-52841722 on Build GRCh38
rs763625917 Chr.19: 52841524-52841524 on Build GRCh38
rs780912199 Chr.19: 52841724-52841724 on Build GRCh38
rs772737407 Chr.19: 52841654-52841654 on Build GRCh38
rs61729939 Chr.19: 52841842-52841842 on Build GRCh38
rs761052881 Chr.19: 52841633-52841634 on Build GRCh38
rs368126101 Chr.19: 52841649-52841649 on Build GRCh38
rs763917860 Chr.19: 52841766-52841766 on Build GRCh38
rs758331206 Chr.19: 52841629-52841629 on Build GRCh38
rs370732747 Chr.19: 52841655-52841655 on Build GRCh38
rs61729957 Chr.19: 52841890-52841890 on Build GRCh38
rs748277946 Chr.19: 52841457-52841457 on Build GRCh38
rs771790702 Chr.19: 52841567-52841567 on Build GRCh38
rs554907102 Chr.19: 52841827-52841827 on Build GRCh38
rs368886195 Chr.19: 52841799-52841799 on Build GRCh38
rs61747364 Chr.19: 52841807-52841807 on Build GRCh38
rs760489803 Chr.19: 52841935-52841935 on Build GRCh38
rs531943295 Chr.19: 52841528-52841528 on Build GRCh38
rs61747369 Chr.19: 52841823-52841823 on Build GRCh38
rs562023544 Chr.19: 52841930-52841930 on Build GRCh38
rs761215426 Chr.19: 52841760-52841760 on Build GRCh38
rs189864341 Chr.19: 52841591-52841591 on Build GRCh38
rs201739972 Chr.19: 52841840-52841840 on Build GRCh38
rs778139055 Chr.19: 52841618-52841618 on Build GRCh38
rs780852217 Chr.19: 52841787-52841787 on Build GRCh38
rs772567431 Chr.19: 52841597-52841598 on Build GRCh38
rs372670332 Chr.19: 52841522-52841522 on Build GRCh38
rs773464176 Chr.19: 52841618-52841618 on Build GRCh38
rs556758852 Chr.19: 52841841-52841841 on Build GRCh38
rs754869305 Chr.19: 52841783-52841783 on Build GRCh38
rs749368180 Chr.19: 52841556-52841556 on Build GRCh38
rs139255764 Chr.19: 52841887-52841887 on Build GRCh38
rs376660879 Chr.19: 52841828-52841828 on Build GRCh38
rs372370233 Chr.19: 52841879-52841879 on Build GRCh38
rs754803618 Chr.19: 52841715-52841715 on Build GRCh38
rs750463269 Chr.19: 52841706-52841706 on Build GRCh38
rs756904256 Chr.19: 52841875-52841875 on Build GRCh38
rs377446855 Chr.19: 52841523-52841523 on Build GRCh38
rs984166520 Chr.19: 52841469-52841469 on Build GRCh38
rs754013523 Chr.19: 52841619-52841619 on Build GRCh38
rs373657914 Chr.19: 52841907-52841907 on Build GRCh38
rs758439077 Chr.19: 52841768-52841768 on Build GRCh38
rs1024522731 Chr.19: 52841806-52841806 on Build GRCh38
rs748800318 Chr.19: 52841810-52841810 on Build GRCh38
rs758391769 Chr.19: 52841717-52841717 on Build GRCh38
rs778867669 Chr.19: 52841705-52841705 on Build GRCh38
rs764460652 Chr.19: 52841693-52841693 on Build GRCh38
rs143407197 Chr.19: 52841749-52841749 on Build GRCh38
rs201629784 Chr.19: 52841905-52841905 on Build GRCh38
rs747917943 Chr.19: 52841792-52841792 on Build GRCh38
rs755917273 Chr.19: 52841856-52841856 on Build GRCh38
rs759249419 Chr.19: 52841510-52841510 on Build GRCh38
rs150110385 Chr.19: 52841838-52841838 on Build GRCh38
rs753042865 Chr.19: 52841614-52841614 on Build GRCh38
rs779959528 Chr.19: 52841719-52841719 on Build GRCh38
rs202238742 Chr.19: 52841547-52841547 on Build GRCh38
rs758116399 Chr.19: 52841646-52841646 on Build GRCh38
rs146755405 Chr.19: 52841782-52841782 on Build GRCh38
rs781265507 Chr.19: 52841451-52841451 on Build GRCh38
rs761221140 Chr.19: 52841598-52841598 on Build GRCh38
rs761958700 Chr.19: 52841762-52841762 on Build GRCh38
rs748400365 Chr.19: 52841551-52841551 on Build GRCh38
rs781522253 Chr.19: 52841549-52841549 on Build GRCh38
rs769419642 Chr.19: 52841736-52841736 on Build GRCh38
rs773373456 Chr.19: 52841474-52841474 on Build GRCh38
rs778002930 Chr.19: 52841555-52841555 on Build GRCh38
rs774469813 Chr.19: 52841916-52841916 on Build GRCh38
rs185070718 Chr.19: 52841506-52841506 on Build GRCh38
rs760328566 Chr.19: 52841574-52841574 on Build GRCh38
rs372954739 Chr.19: 52841509-52841509 on Build GRCh38
rs568485455 Chr.19: 52841560-52841560 on Build GRCh38
rs756416077 Chr.19: 52841616-52841616 on Build GRCh38
rs749265715 Chr.19: 52841494-52841494 on Build GRCh38
rs201739586 Chr.19: 52841534-52841534 on Build GRCh38
rs771981710 Chr.19: 52841630-52841630 on Build GRCh38
rs752499680 Chr.19: 52841846-52841846 on Build GRCh38

View Full Product Details