Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 52880497-52881027 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF320
Entrez Gene ID
162967
Gene Name
zinc finger protein 320
Gene Aliases
ZFPL
UniGene
Hs.369632
Gene Chromosome Location
Chr.19: 52863790-52897695 on Build GRCh38
Transcripts
NM_207333.2 XM_006723059.3 XM_017026393.1 XM_017026394.1 XM_017026395.1 XM_017026396.1 XM_017026397.1 XM_017026398.1 XM_017026399.1 XM_017026400.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
363852 c.1192G>C p.A398P Chr.19: 52880934-52880934 on Build GRCh38
1396026 c.1265A>G p.Y422C Chr.19: 52880861-52880861 on Build GRCh38
5390602 c.1151C>A p.P384H Chr.19: 52880975-52880975 on Build GRCh38
7130207 c.1438C>T p.L480L Chr.19: 52880688-52880688 on Build GRCh38
1396024 c.1483A>G p.N495D Chr.19: 52880643-52880643 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771304841 Chr.19: 52880793-52880795 on Build GRCh38
rs776730042 Chr.19: 52880854-52880854 on Build GRCh38
rs893266172 Chr.19: 52880538-52880538 on Build GRCh38
rs758038010 Chr.19: 52880711-52880711 on Build GRCh38
rs751302762 Chr.19: 52880873-52880873 on Build GRCh38
rs764040180 Chr.19: 52880623-52880623 on Build GRCh38
rs768969674 Chr.19: 52880914-52880914 on Build GRCh38
rs747373803 Chr.19: 52880743-52880744 on Build GRCh38
rs780533589 Chr.19: 52880898-52880898 on Build GRCh38
rs1033117472 Chr.19: 52880535-52880535 on Build GRCh38
rs1001878850 Chr.19: 52880757-52880757 on Build GRCh38
rs181303436 Chr.19: 52880521-52880521 on Build GRCh38
rs758231454 Chr.19: 52880744-52880744 on Build GRCh38
rs146337252 Chr.19: 52880943-52880943 on Build GRCh38
rs757081122 Chr.19: 52880531-52880531 on Build GRCh38
rs566255769 Chr.19: 52880985-52880985 on Build GRCh38
rs919786546 Chr.19: 52880655-52880655 on Build GRCh38
rs977284479 Chr.19: 52880591-52880591 on Build GRCh38
rs768248205 Chr.19: 52880648-52880648 on Build GRCh38
rs370860662 Chr.19: 52880929-52880929 on Build GRCh38
rs954475999 Chr.19: 52880616-52880616 on Build GRCh38
rs532293032 Chr.19: 52880925-52880925 on Build GRCh38
rs61745485 Chr.19: 52880944-52880944 on Build GRCh38
rs373942525 Chr.19: 52880994-52880994 on Build GRCh38
rs761804318 Chr.19: 52880726-52880726 on Build GRCh38
rs752761166 Chr.19: 52880610-52880610 on Build GRCh38
rs754729607 Chr.19: 52880878-52880878 on Build GRCh38
rs754532447 Chr.19: 52880769-52880769 on Build GRCh38
rs778304049 Chr.19: 52880606-52880607 on Build GRCh38
rs6509701 Chr.19: 52880932-52880932 on Build GRCh38
rs746327975 Chr.19: 52880812-52880812 on Build GRCh38
rs748325945 Chr.19: 52880924-52880924 on Build GRCh38
rs764990633 Chr.19: 52880578-52880578 on Build GRCh38
rs867472984 Chr.19: 52880647-52880647 on Build GRCh38
rs750264235 Chr.19: 52880737-52880737 on Build GRCh38
rs141749614 Chr.19: 52880962-52880962 on Build GRCh38
rs962704617 Chr.19: 52880905-52880905 on Build GRCh38
rs966857102 Chr.19: 52880590-52880590 on Build GRCh38
rs775658665 Chr.19: 52880831-52880831 on Build GRCh38
rs758817691 Chr.19: 52880897-52880897 on Build GRCh38
rs746399920 Chr.19: 52880892-52880892 on Build GRCh38
rs369717991 Chr.19: 52880826-52880826 on Build GRCh38
rs113232243 Chr.19: 52880995-52880995 on Build GRCh38
rs759898284 Chr.19: 52880608-52880608 on Build GRCh38
rs759230675 Chr.19: 52880941-52880941 on Build GRCh38
rs957015760 Chr.19: 52880552-52880552 on Build GRCh38
rs753900632 Chr.19: 52880725-52880725 on Build GRCh38
rs779678566 Chr.19: 52880752-52880752 on Build GRCh38
rs997267579 Chr.19: 52880841-52880841 on Build GRCh38
rs761014009 Chr.19: 52880833-52880833 on Build GRCh38
rs373579320 Chr.19: 52880767-52880767 on Build GRCh38
rs762736981 Chr.19: 52880859-52880859 on Build GRCh38
rs756197597 Chr.19: 52880614-52880614 on Build GRCh38
rs983545923 Chr.19: 52880797-52880797 on Build GRCh38
rs773795610 Chr.19: 52880569-52880569 on Build GRCh38
rs779731223 Chr.19: 52880645-52880645 on Build GRCh38
rs779540500 Chr.19: 52880808-52880808 on Build GRCh38
rs376442224 Chr.19: 52880786-52880786 on Build GRCh38
rs763754730 Chr.19: 52880949-52880949 on Build GRCh38
rs149561496 Chr.19: 52880886-52880886 on Build GRCh38
rs140558068 Chr.19: 52880704-52880704 on Build GRCh38
rs749829226 Chr.19: 52880664-52880664 on Build GRCh38
rs763008743 Chr.19: 52880926-52880926 on Build GRCh38
rs543122941 Chr.19: 52880524-52880524 on Build GRCh38
rs138342793 Chr.19: 52880856-52880856 on Build GRCh38
rs372571551 Chr.19: 52880840-52880840 on Build GRCh38
rs770249543 Chr.19: 52880562-52880562 on Build GRCh38
rs796626320 Chr.19: 52880612-52880612 on Build GRCh38
rs376563288 Chr.19: 52880632-52880632 on Build GRCh38
rs138817536 Chr.19: 52880915-52880915 on Build GRCh38
rs1009823323 Chr.19: 52880800-52880800 on Build GRCh38
rs758084679 Chr.19: 52880642-52880642 on Build GRCh38
rs954214469 Chr.19: 52880885-52880885 on Build GRCh38
rs185538280 Chr.19: 52880635-52880635 on Build GRCh38
rs746493857 Chr.19: 52880554-52880554 on Build GRCh38
rs1010006791 Chr.19: 52880550-52880550 on Build GRCh38
rs755709205 Chr.19: 52880887-52880887 on Build GRCh38
rs767249705 Chr.19: 52880881-52880881 on Build GRCh38
rs371617114 Chr.19: 52880594-52880594 on Build GRCh38
rs971112204 Chr.19: 52880621-52880621 on Build GRCh38
rs771607827 Chr.19: 52880790-52880790 on Build GRCh38
rs1001453788 Chr.19: 52880530-52880530 on Build GRCh38
rs1009957440 Chr.19: 52880565-52880565 on Build GRCh38
rs369546350 Chr.19: 52880719-52880719 on Build GRCh38
rs772489875 Chr.19: 52880817-52880817 on Build GRCh38
rs780910534 Chr.19: 52880780-52880780 on Build GRCh38
rs765125570 Chr.19: 52880729-52880729 on Build GRCh38
rs772232565 Chr.19: 52880700-52880700 on Build GRCh38
rs765248150 Chr.19: 52880855-52880855 on Build GRCh38
rs763518248 Chr.19: 52880588-52880588 on Build GRCh38
rs1049486545 Chr.19: 52880862-52880862 on Build GRCh38
rs780504603 Chr.19: 52880653-52880653 on Build GRCh38
rs750131215 Chr.19: 52880677-52880677 on Build GRCh38
rs774924865 Chr.19: 52880689-52880689 on Build GRCh38
rs757128093 Chr.19: 52880628-52880628 on Build GRCh38
rs774802100 Chr.19: 52880602-52880602 on Build GRCh38
rs753778414 Chr.19: 52880627-52880627 on Build GRCh38
rs376084625 Chr.19: 52880864-52880864 on Build GRCh38
rs746589025 Chr.19: 52880646-52880646 on Build GRCh38
rs747302514 Chr.19: 52880910-52880910 on Build GRCh38
rs985712839 Chr.19: 52880624-52880624 on Build GRCh38
rs145022961 Chr.19: 52880889-52880889 on Build GRCh38
rs573045488 Chr.19: 52880691-52880691 on Build GRCh38
rs767777453 Chr.19: 52880609-52880609 on Build GRCh38
rs932266422 Chr.19: 52880923-52880923 on Build GRCh38
rs755770413 Chr.19: 52880947-52880947 on Build GRCh38
rs780400335 Chr.19: 52880963-52880963 on Build GRCh38

View Full Product Details