Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 52952586-52953096 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF816
Entrez Gene ID
125893
Gene Name
zinc finger protein 816
Gene Aliases
ZNF816A
UniGene
Hs.655592
Gene Chromosome Location
Chr.19: 52949379-52962911 on Build GRCh38
Transcripts
NM_001031665.2 NM_001202456.1 NM_001202457.1 NM_001202473.1

Gene Symbol
ZNF816-ZNF321P
Entrez Gene ID
100529240
Gene Name
ZNF816-ZNF321P readthrough
Gene Aliases
ZNF816-ZNF321
UniGene
Hs.675715
Gene Chromosome Location
Chr.19: 52927135-52962911 on Build GRCh38
Transcripts
NM_001031665.2 NM_001202456.1 NM_001202457.1 NM_001202473.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6288952 c.96G>A p.E32E Chr.19: 52952845-52952845 on Build GRCh38
7117763 c.72G>C p.L24F Chr.19: 52952869-52952869 on Build GRCh38
5804994 c.180G>A p.L60L Chr.19: 52952761-52952761 on Build GRCh38
3538244 c.160T>C p.L54L Chr.19: 52952781-52952781 on Build GRCh38
1000694 c.99C>A p.F33L Chr.19: 52952842-52952842 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs943445688 Chr.19: 52952782-52952782 on Build GRCh38
rs774089384 Chr.19: 52952752-52952752 on Build GRCh38
rs867971269 Chr.19: 52952657-52952657 on Build GRCh38
rs185038404 Chr.19: 52952926-52952926 on Build GRCh38
rs755243814 Chr.19: 52952628-52952628 on Build GRCh38
rs752991420 Chr.19: 52952812-52952812 on Build GRCh38
rs760280625 Chr.19: 52952704-52952704 on Build GRCh38
rs771711459 Chr.19: 52952761-52952761 on Build GRCh38
rs925221181 Chr.19: 52952641-52952641 on Build GRCh38
rs906007583 Chr.19: 52952916-52952916 on Build GRCh38
rs139679541 Chr.19: 52952874-52952874 on Build GRCh38
rs746714099 Chr.19: 52952894-52952894 on Build GRCh38
rs758880184 Chr.19: 52952623-52952623 on Build GRCh38
rs776386196 Chr.19: 52952915-52952915 on Build GRCh38
rs751005304 Chr.19: 52952712-52952712 on Build GRCh38
rs756229242 Chr.19: 52952728-52952728 on Build GRCh38
rs767743104 Chr.19: 52952855-52952855 on Build GRCh38
rs73584424 Chr.19: 52952911-52952911 on Build GRCh38
rs748295895 Chr.19: 52952727-52952727 on Build GRCh38
rs752985239 Chr.19: 52952857-52952857 on Build GRCh38
rs774769130 Chr.19: 52952935-52952935 on Build GRCh38
rs770780676 Chr.19: 52952665-52952665 on Build GRCh38
rs151124879 Chr.19: 52952842-52952842 on Build GRCh38
rs1039403352 Chr.19: 52952828-52952828 on Build GRCh38
rs774181029 Chr.19: 52952672-52952672 on Build GRCh38
rs532144492 Chr.19: 52953029-52953029 on Build GRCh38
rs763671094 Chr.19: 52952711-52952711 on Build GRCh38
rs765515535 Chr.19: 52952740-52952741 on Build GRCh38
rs911032091 Chr.19: 52952756-52952756 on Build GRCh38
rs564823191 Chr.19: 52952999-52952999 on Build GRCh38
rs936012234 Chr.19: 52952860-52952860 on Build GRCh38
rs369422358 Chr.19: 52952740-52952740 on Build GRCh38
rs756915816 Chr.19: 52952851-52952851 on Build GRCh38
rs898191197 Chr.19: 52952942-52952942 on Build GRCh38
rs781500056 Chr.19: 52952726-52952726 on Build GRCh38
rs768345755 Chr.19: 52952768-52952768 on Build GRCh38
rs781208367 Chr.19: 52952632-52952632 on Build GRCh38
rs7508016 Chr.19: 52952714-52952714 on Build GRCh38
rs925948578 Chr.19: 52952833-52952833 on Build GRCh38
rs749195289 Chr.19: 52952664-52952664 on Build GRCh38
rs775253097 Chr.19: 52952764-52952764 on Build GRCh38
rs779707302 Chr.19: 52952893-52952893 on Build GRCh38
rs202129754 Chr.19: 52952748-52952748 on Build GRCh38
rs151033143 Chr.19: 52952625-52952625 on Build GRCh38
rs770866535 Chr.19: 52952751-52952751 on Build GRCh38
rs187551944 Chr.19: 52952640-52952640 on Build GRCh38
rs776297545 Chr.19: 52952788-52952788 on Build GRCh38
rs756317241 Chr.19: 52952858-52952858 on Build GRCh38
rs751965933 Chr.19: 52952815-52952815 on Build GRCh38
rs2011496 Chr.19: 52952624-52952624 on Build GRCh38
rs35548880 Chr.19: 52953018-52953019 on Build GRCh38
rs992263491 Chr.19: 52952998-52952998 on Build GRCh38
rs761434657 Chr.19: 52952920-52952920 on Build GRCh38
rs550371677 Chr.19: 52953055-52953055 on Build GRCh38
rs201394440 Chr.19: 52952873-52952873 on Build GRCh38
rs761342266 Chr.19: 52952790-52952790 on Build GRCh38
rs563812762 Chr.19: 52952694-52952694 on Build GRCh38
rs745706741 Chr.19: 52952754-52952754 on Build GRCh38
rs375978725 Chr.19: 52952767-52952767 on Build GRCh38
rs750906601 Chr.19: 52952611-52952611 on Build GRCh38
rs974818605 Chr.19: 52952811-52952811 on Build GRCh38
rs774965083 Chr.19: 52952696-52952696 on Build GRCh38
rs773182609 Chr.19: 52952653-52952653 on Build GRCh38
rs766740883 Chr.19: 52952722-52952722 on Build GRCh38
rs769817734 Chr.19: 52952648-52952648 on Build GRCh38
rs755261333 Chr.19: 52952723-52952723 on Build GRCh38
rs771938427 Chr.19: 52952878-52952878 on Build GRCh38
rs899573023 Chr.19: 52952837-52952837 on Build GRCh38
rs979295479 Chr.19: 52953058-52953058 on Build GRCh38
rs758966694 Chr.19: 52952720-52952720 on Build GRCh38
rs767106085 Chr.19: 52952885-52952885 on Build GRCh38
rs766945574 Chr.19: 52952804-52952804 on Build GRCh38
rs745767641 Chr.19: 52952876-52952876 on Build GRCh38
rs777912323 Chr.19: 52952871-52952871 on Build GRCh38
rs759085333 Chr.19: 52953073-52953073 on Build GRCh38
rs759192810 Chr.19: 52952680-52952680 on Build GRCh38
rs200179785 Chr.19: 52952843-52952843 on Build GRCh38

View Full Product Details