Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 53251450-53251963 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF677
Entrez Gene ID
342926
Gene Name
zinc finger protein 677
Gene Aliases
-
UniGene
Hs.20506
Gene Chromosome Location
Chr.19: 53235381-53254903 on Build GRCh38
Transcripts
NM_001317998.1 NM_182609.3 XM_005258862.4 XM_011526912.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4400311 c.13C>T p.Q5* Chr.19: 53251538-53251538 on Build GRCh38
4127851 c.12T>A p.S4S Chr.19: 53251539-53251539 on Build GRCh38
4854490 c.13C>G p.Q5E Chr.19: 53251538-53251538 on Build GRCh38
4422578 c.10T>A p.S4T Chr.19: 53251541-53251541 on Build GRCh38
4653208 c.4G>A p.A2T Chr.19: 53251547-53251547 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs45575234 Chr.19: 53251762-53251762 on Build GRCh38
rs148964887 Chr.19: 53251930-53251930 on Build GRCh38
rs529567835 Chr.19: 53251788-53251788 on Build GRCh38
rs565702943 Chr.19: 53251924-53251924 on Build GRCh38
rs1012895732 Chr.19: 53251667-53251667 on Build GRCh38
rs1050942590 Chr.19: 53251566-53251566 on Build GRCh38
rs973826208 Chr.19: 53251897-53251897 on Build GRCh38
rs376998797 Chr.19: 53251640-53251640 on Build GRCh38
rs775761973 Chr.19: 53251539-53251542 on Build GRCh38
rs369650788 Chr.19: 53251730-53251730 on Build GRCh38
rs1021429601 Chr.19: 53251555-53251568 on Build GRCh38
rs914977288 Chr.19: 53251880-53251880 on Build GRCh38
rs771099483 Chr.19: 53251544-53251544 on Build GRCh38
rs749932800 Chr.19: 53251577-53251580 on Build GRCh38
rs903388197 Chr.19: 53251929-53251929 on Build GRCh38
rs771169707 Chr.19: 53251588-53251588 on Build GRCh38
rs942458598 Chr.19: 53251888-53251888 on Build GRCh38
rs955273959 Chr.19: 53251784-53251784 on Build GRCh38
rs747810418 Chr.19: 53251934-53251934 on Build GRCh38
rs750018583 Chr.19: 53251538-53251538 on Build GRCh38
rs141315010 Chr.19: 53251663-53251664 on Build GRCh38
rs200123735 Chr.19: 53251535-53251535 on Build GRCh38
rs779531765 Chr.19: 53251560-53251560 on Build GRCh38
rs915225885 Chr.19: 53251524-53251524 on Build GRCh38
rs568705024 Chr.19: 53251776-53251776 on Build GRCh38
rs895285847 Chr.19: 53251937-53251937 on Build GRCh38
rs760687227 Chr.19: 53251496-53251496 on Build GRCh38
rs919595749 Chr.19: 53251907-53251907 on Build GRCh38
rs968014997 Chr.19: 53251768-53251768 on Build GRCh38
rs562610543 Chr.19: 53251536-53251536 on Build GRCh38
rs770447279 Chr.19: 53251751-53251754 on Build GRCh38
rs573870872 Chr.19: 53251525-53251525 on Build GRCh38
rs1038473617 Chr.19: 53251612-53251612 on Build GRCh38
rs946452719 Chr.19: 53251889-53251889 on Build GRCh38
rs1023490591 Chr.19: 53251673-53251673 on Build GRCh38
rs758946515 Chr.19: 53251567-53251567 on Build GRCh38
rs114088665 Chr.19: 53251735-53251735 on Build GRCh38
rs376886189 Chr.19: 53251578-53251578 on Build GRCh38
rs752797442 Chr.19: 53251780-53251780 on Build GRCh38
rs757980180 Chr.19: 53251552-53251552 on Build GRCh38
rs560273068 Chr.19: 53251480-53251480 on Build GRCh38
rs550339099 Chr.19: 53251765-53251765 on Build GRCh38
rs764059865 Chr.19: 53251498-53251498 on Build GRCh38
rs71363316 Chr.19: 53251627-53251627 on Build GRCh38
rs547506915 Chr.19: 53251914-53251914 on Build GRCh38
rs778655400 Chr.19: 53251553-53251555 on Build GRCh38
rs777053459 Chr.19: 53251655-53251655 on Build GRCh38
rs780828229 Chr.19: 53251866-53251866 on Build GRCh38
rs1035191294 Chr.19: 53251693-53251693 on Build GRCh38
rs1047927317 Chr.19: 53251661-53251661 on Build GRCh38
rs911045595 Chr.19: 53251859-53251859 on Build GRCh38
rs28676766 Chr.19: 53251754-53251754 on Build GRCh38
rs886617046 Chr.19: 53251676-53251676 on Build GRCh38
rs753683438 Chr.19: 53251522-53251522 on Build GRCh38
rs977971582 Chr.19: 53251550-53251550 on Build GRCh38
rs942447782 Chr.19: 53251603-53251603 on Build GRCh38
rs201268025 Chr.19: 53251570-53251570 on Build GRCh38
rs922510627 Chr.19: 53251851-53251851 on Build GRCh38
rs138863940 Chr.19: 53251618-53251618 on Build GRCh38
rs756625859 Chr.19: 53251565-53251565 on Build GRCh38
rs750922952 Chr.19: 53251499-53251499 on Build GRCh38
rs978013822 Chr.19: 53251770-53251770 on Build GRCh38
rs148734030 Chr.19: 53251825-53251826 on Build GRCh38
rs754511271 Chr.19: 53251769-53251769 on Build GRCh38
rs985066843 Chr.19: 53251872-53251872 on Build GRCh38
rs1004153400 Chr.19: 53251688-53251688 on Build GRCh38
rs377226787 Chr.19: 53251506-53251506 on Build GRCh38
rs767144135 Chr.19: 53251560-53251563 on Build GRCh38
rs533097978 Chr.19: 53251699-53251699 on Build GRCh38
rs995068773 Chr.19: 53251652-53251652 on Build GRCh38
rs751002619 Chr.19: 53251562-53251562 on Build GRCh38
rs374002530 Chr.19: 53251495-53251495 on Build GRCh38
rs760200482 Chr.19: 53251587-53251588 on Build GRCh38
rs2965240 Chr.19: 53251483-53251483 on Build GRCh38

View Full Product Details