Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 53576507-53577011 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF331
Entrez Gene ID
55422
Gene Name
zinc finger protein 331
Gene Aliases
RITA, ZNF361, ZNF463
UniGene
Hs.185674
Gene Chromosome Location
Chr.19: 53503819-53580269 on Build GRCh38
Transcripts
NM_001079906.1 NM_001079907.1 NM_001253798.1 NM_001253799.1 NM_001253800.1 NM_001253801.2 NM_001317113.1 NM_001317114.1 NM_001317115.1 NM_001317116.1 NM_001317117.1 NM_001317118.1 NM_001317119.1 NM_001317120.1 NM_001317121.1 NM_018555.5 XM_011527076.2 XM_011527078.2 XM_017026936.1 XM_017026937.1 XM_017026938.1 XM_017026939.1 XM_017026940.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5650682 c.240C>T p.G80G Chr.19: 53576800-53576800 on Build GRCh38
5515737 c.324C>A p.V108V Chr.19: 53576884-53576884 on Build GRCh38
3893327 c.381G>A p.K127K Chr.19: 53576941-53576941 on Build GRCh38
475221 c.447T>G p.H149Q Chr.19: 53577007-53577007 on Build GRCh38
6829392 c.295T>G p.Y99D Chr.19: 53576855-53576855 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs904039782 Chr.19: 53576565-53576565 on Build GRCh38
rs1020973825 Chr.19: 53576730-53576730 on Build GRCh38
rs778952155 Chr.19: 53576709-53576709 on Build GRCh38
rs747179966 Chr.19: 53576866-53576866 on Build GRCh38
rs374635678 Chr.19: 53576810-53576810 on Build GRCh38
rs1013988836 Chr.19: 53576587-53576587 on Build GRCh38
rs750130989 Chr.19: 53576682-53576682 on Build GRCh38
rs748236758 Chr.19: 53576733-53576733 on Build GRCh38
rs146144933 Chr.19: 53576807-53576807 on Build GRCh38
rs555149394 Chr.19: 53576564-53576564 on Build GRCh38
rs935482287 Chr.19: 53576779-53576779 on Build GRCh38
rs147610030 Chr.19: 53576777-53576777 on Build GRCh38
rs760274215 Chr.19: 53576924-53576924 on Build GRCh38
rs74660347 Chr.19: 53576579-53576579 on Build GRCh38
rs188879831 Chr.19: 53576537-53576537 on Build GRCh38
rs751046137 Chr.19: 53576653-53576653 on Build GRCh38
rs564775772 Chr.19: 53576678-53576678 on Build GRCh38
rs141196058 Chr.19: 53576794-53576794 on Build GRCh38
rs8106447 Chr.19: 53576539-53576539 on Build GRCh38
rs775913323 Chr.19: 53576898-53576898 on Build GRCh38
rs772337491 Chr.19: 53576734-53576734 on Build GRCh38
rs144927498 Chr.19: 53576571-53576571 on Build GRCh38
rs573071724 Chr.19: 53576766-53576766 on Build GRCh38
rs758486389 Chr.19: 53576840-53576840 on Build GRCh38
rs754853112 Chr.19: 53576708-53576708 on Build GRCh38
rs1053617488 Chr.19: 53576919-53576919 on Build GRCh38
rs76587173 Chr.19: 53576620-53576620 on Build GRCh38
rs747239238 Chr.19: 53576748-53576748 on Build GRCh38
rs114674002 Chr.19: 53576533-53576533 on Build GRCh38
rs778083871 Chr.19: 53576744-53576744 on Build GRCh38
rs753763555 Chr.19: 53576701-53576701 on Build GRCh38
rs776403470 Chr.19: 53576937-53576937 on Build GRCh38
rs757396686 Chr.19: 53576877-53576877 on Build GRCh38
rs905084517 Chr.19: 53576621-53576621 on Build GRCh38
rs34867717 Chr.19: 53576953-53576954 on Build GRCh38
rs765946542 Chr.19: 53576925-53576925 on Build GRCh38
rs142196434 Chr.19: 53576802-53576802 on Build GRCh38
rs779748465 Chr.19: 53576694-53576694 on Build GRCh38
rs769561349 Chr.19: 53576688-53576688 on Build GRCh38
rs1006667616 Chr.19: 53576824-53576824 on Build GRCh38
rs8109631 Chr.19: 53576890-53576890 on Build GRCh38
rs1016476280 Chr.19: 53576623-53576623 on Build GRCh38
rs1046764205 Chr.19: 53576581-53576581 on Build GRCh38
rs749796126 Chr.19: 53576911-53576911 on Build GRCh38
rs369494593 Chr.19: 53576841-53576841 on Build GRCh38
rs772690900 Chr.19: 53576922-53576922 on Build GRCh38
rs201885186 Chr.19: 53576772-53576772 on Build GRCh38
rs954574618 Chr.19: 53576896-53576896 on Build GRCh38
rs765129221 Chr.19: 53576960-53576960 on Build GRCh38
rs776880475 Chr.19: 53576759-53576759 on Build GRCh38
rs935533346 Chr.19: 53576570-53576570 on Build GRCh38
rs543993677 Chr.19: 53576801-53576801 on Build GRCh38
rs766379010 Chr.19: 53576939-53576941 on Build GRCh38
rs752695555 Chr.19: 53576822-53576822 on Build GRCh38
rs368241758 Chr.19: 53576913-53576913 on Build GRCh38
rs1052468927 Chr.19: 53576575-53576575 on Build GRCh38
rs112855712 Chr.19: 53576886-53576886 on Build GRCh38
rs752642053 Chr.19: 53576969-53576969 on Build GRCh38
rs755876770 Chr.19: 53576693-53576693 on Build GRCh38
rs758433153 Chr.19: 53576985-53576985 on Build GRCh38
rs766107190 Chr.19: 53576806-53576806 on Build GRCh38
rs770166196 Chr.19: 53576769-53576769 on Build GRCh38
rs376189297 Chr.19: 53576646-53576646 on Build GRCh38
rs1006139528 Chr.19: 53576622-53576622 on Build GRCh38
rs574088222 Chr.19: 53576780-53576782 on Build GRCh38
rs961871832 Chr.19: 53576645-53576645 on Build GRCh38
rs1052577637 Chr.19: 53576785-53576785 on Build GRCh38
rs771286443 Chr.19: 53576753-53576753 on Build GRCh38
rs761350718 Chr.19: 53576660-53576660 on Build GRCh38
rs143490707 Chr.19: 53576795-53576795 on Build GRCh38
rs140399327 Chr.19: 53576823-53576823 on Build GRCh38
rs534408623 Chr.19: 53576576-53576576 on Build GRCh38
rs150285639 Chr.19: 53576825-53576825 on Build GRCh38
rs372185704 Chr.19: 53576873-53576873 on Build GRCh38
rs925575008 Chr.19: 53576778-53576778 on Build GRCh38
rs61744130 Chr.19: 53576813-53576813 on Build GRCh38

View Full Product Details