Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 54148258-54148761 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNOT3
Entrez Gene ID
4849
Gene Name
CCR4-NOT transcription complex subunit 3
Gene Aliases
LENG2, NOT3, NOT3H
UniGene
Hs.343571
Gene Chromosome Location
Chr.19: 54137689-54155708 on Build GRCh38
Transcripts
NM_014516.3 XM_005278279.1 XM_005278280.2 XM_005278281.1 XM_005278282.2 XM_011526992.1 XM_011526993.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5751752 c.863+1G>AG p.? Chr.19: 54148744-54148744 on Build GRCh38
4081191 c.1140C>A p.P380P Chr.19: 54148393-54148393 on Build GRCh38
116731 c.1073G>T p.S358I Chr.19: 54148326-54148326 on Build GRCh38
3423132 c.1189G>A p.G397R Chr.19: 54148442-54148442 on Build GRCh38
6631461 c.731G>T p.G244V Chr.19: 54148527-54148527 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780817596 Chr.19: 54148447-54148447 on Build GRCh38
rs771212010 Chr.19: 54148439-54148440 on Build GRCh38
rs768608878 Chr.19: 54148456-54148456 on Build GRCh38
rs759703178 Chr.19: 54148450-54148451 on Build GRCh38
rs187281282 Chr.19: 54148564-54148564 on Build GRCh38
rs753359008 Chr.19: 54148690-54148690 on Build GRCh38
rs759140502 Chr.19: 54148691-54148691 on Build GRCh38
rs587620376 Chr.19: 54148705-54148705 on Build GRCh38
rs774905572 Chr.19: 54148650-54148650 on Build GRCh38
rs753191686 Chr.19: 54148478-54148478 on Build GRCh38
rs763419217 Chr.19: 54148460-54148465 on Build GRCh38
rs765659643 Chr.19: 54148475-54148475 on Build GRCh38
rs746987499 Chr.19: 54148590-54148590 on Build GRCh38
rs776288486 Chr.19: 54148334-54148334 on Build GRCh38
rs778738177 Chr.19: 54148567-54148567 on Build GRCh38
rs780680395 Chr.19: 54148413-54148413 on Build GRCh38
rs774424359 Chr.19: 54148459-54148459 on Build GRCh38
rs768965964 Chr.19: 54148648-54148648 on Build GRCh38
rs934844672 Chr.19: 54148308-54148308 on Build GRCh38
rs202244707 Chr.19: 54148647-54148647 on Build GRCh38
rs777737001 Chr.19: 54148580-54148580 on Build GRCh38
rs779427467 Chr.19: 54148397-54148397 on Build GRCh38
rs587647820 Chr.19: 54148552-54148552 on Build GRCh38
rs776890135 Chr.19: 54148469-54148469 on Build GRCh38
rs142721836 Chr.19: 54148292-54148292 on Build GRCh38
rs587633280 Chr.19: 54148297-54148297 on Build GRCh38
rs775300624 Chr.19: 54148421-54148421 on Build GRCh38
rs762004272 Chr.19: 54148354-54148354 on Build GRCh38
rs770533336 Chr.19: 54148333-54148333 on Build GRCh38
rs768661906 Chr.19: 54148425-54148425 on Build GRCh38
rs760892457 Chr.19: 54148437-54148437 on Build GRCh38
rs781143246 Chr.19: 54148709-54148709 on Build GRCh38
rs776849334 Chr.19: 54148448-54148468 on Build GRCh38
rs755411891 Chr.19: 54148392-54148392 on Build GRCh38
rs587642594 Chr.19: 54148385-54148385 on Build GRCh38
rs147355537 Chr.19: 54148668-54148668 on Build GRCh38
rs766515811 Chr.19: 54148439-54148439 on Build GRCh38
rs770957095 Chr.19: 54148529-54148529 on Build GRCh38
rs777208855 Chr.19: 54148303-54148303 on Build GRCh38
rs758848913 Chr.19: 54148479-54148479 on Build GRCh38
rs767770883 Chr.19: 54148429-54148429 on Build GRCh38
rs752043894 Chr.19: 54148554-54148554 on Build GRCh38
rs371452117 Chr.19: 54148610-54148610 on Build GRCh38
rs773046365 Chr.19: 54148526-54148526 on Build GRCh38
rs745922399 Chr.19: 54148710-54148710 on Build GRCh38
rs754190780 Chr.19: 54148440-54148440 on Build GRCh38
rs768934869 Chr.19: 54148723-54148723 on Build GRCh38
rs749896898 Chr.19: 54148483-54148483 on Build GRCh38
rs769897893 Chr.19: 54148714-54148714 on Build GRCh38
rs759232707 Chr.19: 54148339-54148339 on Build GRCh38
rs746808824 Chr.19: 54148438-54148439 on Build GRCh38
rs766288492 Chr.19: 54148576-54148576 on Build GRCh38
rs775735242 Chr.19: 54148637-54148637 on Build GRCh38
rs200421099 Chr.19: 54148457-54148457 on Build GRCh38
rs954697492 Chr.19: 54148446-54148446 on Build GRCh38
rs778793002 Chr.19: 54148520-54148520 on Build GRCh38
rs587692629 Chr.19: 54148528-54148528 on Build GRCh38
rs745387907 Chr.19: 54148448-54148448 on Build GRCh38
rs386810848 Chr.19: 54148663-54148665 on Build GRCh38
rs759815446 Chr.19: 54148534-54148534 on Build GRCh38
rs767822171 Chr.19: 54148369-54148369 on Build GRCh38
rs771557222 Chr.19: 54148300-54148300 on Build GRCh38
rs199960898 Chr.19: 54148286-54148286 on Build GRCh38
rs898936978 Chr.19: 54148435-54148435 on Build GRCh38
rs760701265 Chr.19: 54148467-54148467 on Build GRCh38
rs201138446 Chr.19: 54148348-54148348 on Build GRCh38
rs775604995 Chr.19: 54148458-54148478 on Build GRCh38
rs587663518 Chr.19: 54148460-54148460 on Build GRCh38
rs367695100 Chr.19: 54148570-54148570 on Build GRCh38
rs753246658 Chr.19: 54148443-54148443 on Build GRCh38
rs587594795 Chr.19: 54148502-54148502 on Build GRCh38
rs781193024 Chr.19: 54148603-54148603 on Build GRCh38
rs765712541 Chr.19: 54148442-54148442 on Build GRCh38
rs771033124 Chr.19: 54148594-54148594 on Build GRCh38
rs779723240 Chr.19: 54148501-54148501 on Build GRCh38
rs370903869 Chr.19: 54148441-54148441 on Build GRCh38
rs769672981 Chr.19: 54148344-54148344 on Build GRCh38
rs587767936 Chr.19: 54148347-54148347 on Build GRCh38
rs764516938 Chr.19: 54148480-54148480 on Build GRCh38
rs373169471 Chr.19: 54148453-54148453 on Build GRCh38
rs745602918 Chr.19: 54148418-54148418 on Build GRCh38
rs756758337 Chr.19: 54148444-54148444 on Build GRCh38
rs587724455 Chr.19: 54148424-54148424 on Build GRCh38
rs587617892 Chr.19: 54148565-54148565 on Build GRCh38
rs773275096 Chr.19: 54148463-54148463 on Build GRCh38
rs113583507 Chr.19: 54148619-54148619 on Build GRCh38
rs751821201 Chr.19: 54148285-54148285 on Build GRCh38
rs773292484 Chr.19: 54148430-54148430 on Build GRCh38
rs866658475 Chr.19: 54148686-54148686 on Build GRCh38
rs35550825 Chr.19: 54148320-54148321 on Build GRCh38
rs769466994 Chr.19: 54148419-54148419 on Build GRCh38
rs43211 Chr.19: 54148468-54148468 on Build GRCh38
rs749528991 Chr.19: 54148721-54148721 on Build GRCh38
rs774477113 Chr.19: 54148426-54148426 on Build GRCh38
rs751645336 Chr.19: 54148349-54148349 on Build GRCh38
rs750701546 Chr.19: 54148291-54148291 on Build GRCh38
rs755624856 Chr.19: 54148498-54148498 on Build GRCh38
rs746475797 Chr.19: 54148407-54148407 on Build GRCh38
rs747960849 Chr.19: 54148522-54148522 on Build GRCh38
rs772006294 Chr.19: 54148525-54148525 on Build GRCh38
rs898817026 Chr.19: 54148568-54148568 on Build GRCh38
rs181910878 Chr.19: 54148530-54148530 on Build GRCh38
rs759864730 Chr.19: 54148472-54148472 on Build GRCh38
rs769966922 Chr.19: 54148615-54148615 on Build GRCh38
rs587696346 Chr.19: 54148317-54148317 on Build GRCh38
rs917464339 Chr.19: 54148492-54148492 on Build GRCh38
rs771917482 Chr.19: 54148462-54148462 on Build GRCh38
rs774511528 Chr.19: 54148725-54148725 on Build GRCh38
rs760224954 Chr.19: 54148559-54148559 on Build GRCh38
rs201311298 Chr.19: 54148708-54148708 on Build GRCh38
rs368619014 Chr.19: 54148408-54148408 on Build GRCh38
rs587662644 Chr.19: 54148553-54148553 on Build GRCh38
rs780247874 Chr.19: 54148718-54148718 on Build GRCh38
rs769889646 Chr.19: 54148458-54148460 on Build GRCh38
rs781593644 Chr.19: 54148288-54148288 on Build GRCh38
rs756457769 Chr.19: 54148572-54148572 on Build GRCh38
rs1017552124 Chr.19: 54148681-54148681 on Build GRCh38
rs370457959 Chr.19: 54148293-54148293 on Build GRCh38
rs765862398 Chr.19: 54148560-54148560 on Build GRCh38
rs1053315022 Chr.19: 54148314-54148314 on Build GRCh38
rs748576550 Chr.19: 54148737-54148737 on Build GRCh38
rs375381880 Chr.19: 54148654-54148654 on Build GRCh38

View Full Product Details