Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 54180742-54181234 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MBOAT7
Entrez Gene ID
79143
Gene Name
membrane bound O-acyltransferase domain containing 7
Gene Aliases
BB1, LENG4, LPIAT, LRC4, MBOA7, OACT7, hMBOA-7
UniGene
Hs.467279
Gene Chromosome Location
Chr.19: 54173412-54190050 on Build GRCh38
Transcripts
NM_001146056.2 NM_001146082.2 NM_001146083.2 NM_024298.4 XM_011527299.2 XM_011527300.2 XM_017027296.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1001028 c.653G>A p.R218H Chr.19: 54180974-54180974 on Build GRCh38
5804434 c.700A>T p.M234L Chr.19: 54180927-54180927 on Build GRCh38
1001029 c.649G>A p.V217M Chr.19: 54180978-54180978 on Build GRCh38
5637084 c.565C>T p.R189W Chr.19: 54181062-54181062 on Build GRCh38
6709199 c.678G>A p.P226P Chr.19: 54180949-54180949 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs199544070 Chr.19: 54180896-54180896 on Build GRCh38
rs780699317 Chr.19: 54180826-54180826 on Build GRCh38
rs368650520 Chr.19: 54180766-54180766 on Build GRCh38
rs1002066814 Chr.19: 54181115-54181115 on Build GRCh38
rs963764696 Chr.19: 54180875-54180875 on Build GRCh38
rs777804291 Chr.19: 54181006-54181006 on Build GRCh38
rs762838702 Chr.19: 54180762-54180762 on Build GRCh38
rs770883180 Chr.19: 54180950-54180950 on Build GRCh38
rs759699339 Chr.19: 54180957-54180957 on Build GRCh38
rs371452356 Chr.19: 54180909-54180909 on Build GRCh38
rs772114260 Chr.19: 54180940-54180940 on Build GRCh38
rs1051882954 Chr.19: 54180989-54180989 on Build GRCh38
rs776628093 Chr.19: 54181024-54181024 on Build GRCh38
rs774228066 Chr.19: 54180936-54180936 on Build GRCh38
rs766679693 Chr.19: 54180873-54180873 on Build GRCh38
rs544377469 Chr.19: 54180929-54180929 on Build GRCh38
rs200188828 Chr.19: 54181094-54181094 on Build GRCh38
rs868498080 Chr.19: 54180870-54180870 on Build GRCh38
rs751978674 Chr.19: 54180972-54180972 on Build GRCh38
rs370479065 Chr.19: 54180850-54180850 on Build GRCh38
rs746878926 Chr.19: 54181012-54181012 on Build GRCh38
rs17855385 Chr.19: 54180846-54180846 on Build GRCh38
rs775813487 Chr.19: 54181158-54181158 on Build GRCh38
rs371233526 Chr.19: 54180988-54180988 on Build GRCh38
rs751520193 Chr.19: 54180765-54180765 on Build GRCh38
rs769560316 Chr.19: 54180828-54180828 on Build GRCh38
rs778703267 Chr.19: 54180978-54180978 on Build GRCh38
rs755683540 Chr.19: 54180973-54180973 on Build GRCh38
rs756299945 Chr.19: 54180804-54180804 on Build GRCh38
rs750632839 Chr.19: 54180783-54180783 on Build GRCh38
rs765592230 Chr.19: 54180958-54180958 on Build GRCh38
rs1002682177 Chr.19: 54181090-54181090 on Build GRCh38
rs773274817 Chr.19: 54180866-54180866 on Build GRCh38
rs1008624546 Chr.19: 54180853-54180853 on Build GRCh38
rs1055465093 Chr.19: 54180780-54180780 on Build GRCh38
rs866462902 Chr.19: 54181159-54181159 on Build GRCh38
rs761791932 Chr.19: 54180864-54180864 on Build GRCh38
rs1024290876 Chr.19: 54180922-54180922 on Build GRCh38
rs770063247 Chr.19: 54181036-54181036 on Build GRCh38
rs569292312 Chr.19: 54181143-54181143 on Build GRCh38
rs554078016 Chr.19: 54180832-54180832 on Build GRCh38
rs147943964 Chr.19: 54180942-54180942 on Build GRCh38
rs745441950 Chr.19: 54180911-54180911 on Build GRCh38
rs748620020 Chr.19: 54180816-54180816 on Build GRCh38
rs766656768 Chr.19: 54180811-54180811 on Build GRCh38
rs868738531 Chr.19: 54180854-54180854 on Build GRCh38
rs769080101 Chr.19: 54181172-54181172 on Build GRCh38
rs781098311 Chr.19: 54181129-54181129 on Build GRCh38
rs759611488 Chr.19: 54181027-54181027 on Build GRCh38
rs546898988 Chr.19: 54180953-54180953 on Build GRCh38
rs1025895867 Chr.19: 54180889-54180889 on Build GRCh38
rs112064263 Chr.19: 54180967-54180967 on Build GRCh38
rs890215020 Chr.19: 54180840-54180840 on Build GRCh38
rs755372797 Chr.19: 54180885-54180885 on Build GRCh38
rs748032878 Chr.19: 54180979-54180979 on Build GRCh38
rs891735961 Chr.19: 54181050-54181050 on Build GRCh38
rs753220219 Chr.19: 54180899-54180899 on Build GRCh38
rs142100180 Chr.19: 54181169-54181169 on Build GRCh38
rs779894517 Chr.19: 54180919-54180919 on Build GRCh38
rs767593470 Chr.19: 54180778-54180778 on Build GRCh38
rs544702941 Chr.19: 54180867-54180867 on Build GRCh38
rs775668002 Chr.19: 54181037-54181037 on Build GRCh38
rs753465461 Chr.19: 54181080-54181080 on Build GRCh38
rs552304142 Chr.19: 54181049-54181049 on Build GRCh38
rs568301361 Chr.19: 54181176-54181176 on Build GRCh38
rs756743624 Chr.19: 54180825-54180825 on Build GRCh38
rs762167405 Chr.19: 54181075-54181075 on Build GRCh38
rs944385075 Chr.19: 54181192-54181192 on Build GRCh38
rs755557904 Chr.19: 54180918-54180918 on Build GRCh38
rs200372229 Chr.19: 54180761-54180761 on Build GRCh38
rs372916774 Chr.19: 54180871-54180871 on Build GRCh38
rs774511523 Chr.19: 54181174-54181174 on Build GRCh38
rs886041060 Chr.19: 54180772-54180772 on Build GRCh38
rs778754252 Chr.19: 54181092-54181092 on Build GRCh38
rs768541323 Chr.19: 54180852-54180852 on Build GRCh38
rs754522503 Chr.19: 54181081-54181081 on Build GRCh38
rs373603478 Chr.19: 54180865-54180865 on Build GRCh38
rs991531137 Chr.19: 54180948-54180948 on Build GRCh38
rs185682596 Chr.19: 54180974-54180974 on Build GRCh38
rs746001095 Chr.19: 54181136-54181136 on Build GRCh38
rs756563045 Chr.19: 54181156-54181156 on Build GRCh38
rs758241718 Chr.19: 54181108-54181108 on Build GRCh38
rs763416489 Chr.19: 54181044-54181044 on Build GRCh38
rs529954463 Chr.19: 54180949-54180949 on Build GRCh38
rs971416882 Chr.19: 54181083-54181083 on Build GRCh38
rs972203785 Chr.19: 54180888-54180888 on Build GRCh38
rs754032811 Chr.19: 54180874-54180874 on Build GRCh38
rs530531478 Chr.19: 54180902-54180902 on Build GRCh38
rs753130476 Chr.19: 54180962-54180962 on Build GRCh38
rs763217778 Chr.19: 54180966-54180966 on Build GRCh38
rs770923938 Chr.19: 54181116-54181116 on Build GRCh38
rs574739585 Chr.19: 54180763-54180763 on Build GRCh38
rs892852610 Chr.19: 54180810-54180810 on Build GRCh38
rs764328645 Chr.19: 54181046-54181046 on Build GRCh38
rs934061597 Chr.19: 54181086-54181086 on Build GRCh38
rs774462839 Chr.19: 54180862-54180862 on Build GRCh38
rs748911292 Chr.19: 54180924-54180924 on Build GRCh38
rs201515587 Chr.19: 54180934-54180934 on Build GRCh38
rs754265522 Chr.19: 54180812-54180812 on Build GRCh38
rs140431223 Chr.19: 54181157-54181157 on Build GRCh38
rs12460873 Chr.19: 54180834-54180834 on Build GRCh38
rs1054262233 Chr.19: 54181038-54181038 on Build GRCh38
rs368727348 Chr.19: 54181023-54181023 on Build GRCh38
rs1042378789 Chr.19: 54180836-54180836 on Build GRCh38
rs752132040 Chr.19: 54181062-54181062 on Build GRCh38
rs1026364627 Chr.19: 54181061-54181061 on Build GRCh38
rs886041061 Chr.19: 54180801-54180807 on Build GRCh38
rs765990528 Chr.19: 54181079-54181079 on Build GRCh38
rs535699644 Chr.19: 54180938-54180938 on Build GRCh38
rs759040034 Chr.19: 54180805-54180805 on Build GRCh38
rs777379271 Chr.19: 54181111-54181111 on Build GRCh38
rs755319900 Chr.19: 54180815-54180815 on Build GRCh38

View Full Product Details