Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 54189640-54190148 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MBOAT7
Entrez Gene ID
79143
Gene Name
membrane bound O-acyltransferase domain containing 7
Gene Aliases
BB1, LENG4, LPIAT, LRC4, MBOA7, OACT7, hMBOA-7
UniGene
Hs.467279
Gene Chromosome Location
Chr.19: 54173412-54190050 on Build GRCh38
Transcripts
NM_001146056.2 NM_001146082.2 NM_001146083.2 NM_024298.4 XM_011527299.2 XM_017027296.1 XM_011527294.2 XM_011527295.1

Gene Symbol
TSEN34
Entrez Gene ID
79042
Gene Name
tRNA splicing endonuclease subunit 34
Gene Aliases
LENG5, PCH2C, SEN34, SEN34L
UniGene
Hs.15580
Gene Chromosome Location
Chr.19: 54189517-54194536 on Build GRCh38
Transcripts
NM_001146056.2 NM_001146082.2 NM_001146083.2 NM_024298.4 XM_011527299.2 XM_017027296.1 XM_011527294.2 XM_011527295.1

SNP Detail

SNP ID Chromosome Location
rs1046262542 Chr.19: 54190076-54190076 on Build GRCh38
rs954735376 Chr.19: 54189984-54189984 on Build GRCh38
rs1053853581 Chr.19: 54189741-54189741 on Build GRCh38
rs775845743 Chr.19: 54189762-54189762 on Build GRCh38
rs1024907760 Chr.19: 54189692-54189692 on Build GRCh38
rs769214511 Chr.19: 54189910-54189910 on Build GRCh38
rs1002213273 Chr.19: 54189721-54189721 on Build GRCh38
rs3810233 Chr.19: 54190114-54190114 on Build GRCh38
rs957715680 Chr.19: 54190020-54190020 on Build GRCh38
rs535733073 Chr.19: 54189828-54189828 on Build GRCh38
rs685063 Chr.19: 54189908-54189908 on Build GRCh38
rs775319415 Chr.19: 54189899-54189899 on Build GRCh38
rs949240275 Chr.19: 54189888-54189888 on Build GRCh38
rs933764372 Chr.19: 54190099-54190099 on Build GRCh38
rs936117134 Chr.19: 54189905-54189932 on Build GRCh38
rs566045786 Chr.19: 54189929-54189929 on Build GRCh38
rs34011696 Chr.19: 54189885-54189885 on Build GRCh38
rs936450492 Chr.19: 54189832-54189832 on Build GRCh38
rs868376122 Chr.19: 54189899-54189899 on Build GRCh38
rs1013487621 Chr.19: 54189690-54189690 on Build GRCh38
rs899151587 Chr.19: 54189949-54189949 on Build GRCh38
rs569559377 Chr.19: 54189817-54189817 on Build GRCh38
rs910805089 Chr.19: 54189822-54189822 on Build GRCh38
rs39714 Chr.19: 54189831-54189831 on Build GRCh38
rs949122459 Chr.19: 54190027-54190027 on Build GRCh38
rs1049952915 Chr.19: 54189896-54189896 on Build GRCh38
rs1026512109 Chr.19: 54189779-54189779 on Build GRCh38
rs972122876 Chr.19: 54189765-54189765 on Build GRCh38
rs943582608 Chr.19: 54189823-54189823 on Build GRCh38
rs557585422 Chr.19: 54190022-54190022 on Build GRCh38
rs937958522 Chr.19: 54189728-54189728 on Build GRCh38
rs183652776 Chr.19: 54189977-54189977 on Build GRCh38
rs778699237 Chr.19: 54189919-54189919 on Build GRCh38
rs534732344 Chr.19: 54189932-54189932 on Build GRCh38
rs373472898 Chr.19: 54190101-54190101 on Build GRCh38
rs981897636 Chr.19: 54190012-54190012 on Build GRCh38
rs971478377 Chr.19: 54190011-54190011 on Build GRCh38
rs183716 Chr.19: 54189703-54189703 on Build GRCh38
rs979407311 Chr.19: 54190018-54190018 on Build GRCh38
rs36624 Chr.19: 54190017-54190017 on Build GRCh38
rs913656241 Chr.19: 54190024-54190024 on Build GRCh38
rs574317698 Chr.19: 54190085-54190085 on Build GRCh38
rs549566920 Chr.19: 54189768-54189768 on Build GRCh38
rs963491123 Chr.19: 54189756-54189756 on Build GRCh38
rs40357 Chr.19: 54189676-54189676 on Build GRCh38
rs1016646281 Chr.19: 54189740-54189740 on Build GRCh38
rs527245323 Chr.19: 54190124-54190124 on Build GRCh38
rs989371618 Chr.19: 54190019-54190019 on Build GRCh38
rs543283433 Chr.19: 54190102-54190102 on Build GRCh38
rs988022014 Chr.19: 54189807-54189807 on Build GRCh38
rs1016289339 Chr.19: 54189947-54189947 on Build GRCh38
rs1038035351 Chr.19: 54190106-54190106 on Build GRCh38
rs578152364 Chr.19: 54189985-54189985 on Build GRCh38
rs959547189 Chr.19: 54190018-54190019 on Build GRCh38
rs926523063 Chr.19: 54189720-54189720 on Build GRCh38
rs1002524857 Chr.19: 54189921-54189921 on Build GRCh38
rs78435143 Chr.19: 54189671-54189671 on Build GRCh38
rs975139104 Chr.19: 54189824-54189824 on Build GRCh38
rs747782903 Chr.19: 54190001-54190001 on Build GRCh38
rs954871103 Chr.19: 54189781-54189781 on Build GRCh38
rs922336579 Chr.19: 54190029-54190030 on Build GRCh38
rs914949712 Chr.19: 54189754-54189754 on Build GRCh38
rs1052124042 Chr.19: 54189855-54189855 on Build GRCh38
rs867199065 Chr.19: 54189989-54189989 on Build GRCh38
rs543106809 Chr.19: 54190019-54190019 on Build GRCh38
rs892283179 Chr.19: 54189689-54189689 on Build GRCh38
rs1046617159 Chr.19: 54189891-54189891 on Build GRCh38
rs1026418805 Chr.19: 54189973-54189973 on Build GRCh38
rs761432874 Chr.19: 54189993-54189993 on Build GRCh38
rs776968354 Chr.19: 54189969-54189969 on Build GRCh38
rs892189731 Chr.19: 54189881-54189881 on Build GRCh38

View Full Product Details