Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 54338772-54339277 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LILRA4
Entrez Gene ID
23547
Gene Name
leukocyte immunoglobulin like receptor A4
Gene Aliases
CD85g, ILT7
UniGene
Hs.406708
Gene Chromosome Location
Chr.19: 54333185-54339169 on Build GRCh38
Transcripts
NM_012276.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6152415 c.58C>A p.R20R Chr.19: 54338878-54338878 on Build GRCh38
4942241 c.59G>T p.R20L Chr.19: 54338877-54338877 on Build GRCh38
1396285 c.26T>A p.L9H Chr.19: 54339068-54339068 on Build GRCh38
3404593 c.13C>G p.L5V Chr.19: 54339081-54339081 on Build GRCh38
4697981 c.57C>T p.T19T Chr.19: 54338879-54338879 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747598796 Chr.19: 54338830-54338830 on Build GRCh38
rs374849494 Chr.19: 54338934-54338934 on Build GRCh38
rs746792534 Chr.19: 54339034-54339034 on Build GRCh38
rs35649442 Chr.19: 54338914-54338915 on Build GRCh38
rs772428712 Chr.19: 54339063-54339063 on Build GRCh38
rs750918627 Chr.19: 54338821-54338821 on Build GRCh38
rs756760305 Chr.19: 54338864-54338864 on Build GRCh38
rs373330112 Chr.19: 54339017-54339017 on Build GRCh38
rs779193913 Chr.19: 54339055-54339055 on Build GRCh38
rs117653309 Chr.19: 54339150-54339150 on Build GRCh38
rs774540488 Chr.19: 54339011-54339011 on Build GRCh38
rs750797847 Chr.19: 54338880-54338880 on Build GRCh38
rs374179725 Chr.19: 54339044-54339044 on Build GRCh38
rs11880590 Chr.19: 54338917-54338917 on Build GRCh38
rs748253120 Chr.19: 54339130-54339130 on Build GRCh38
rs368643601 Chr.19: 54339140-54339140 on Build GRCh38
rs779732030 Chr.19: 54339032-54339032 on Build GRCh38
rs767221445 Chr.19: 54339099-54339099 on Build GRCh38
rs771165902 Chr.19: 54339141-54339141 on Build GRCh38
rs182804716 Chr.19: 54339159-54339159 on Build GRCh38
rs776583344 Chr.19: 54338848-54338848 on Build GRCh38
rs200178658 Chr.19: 54339077-54339077 on Build GRCh38
rs761302358 Chr.19: 54338888-54338888 on Build GRCh38
rs766792062 Chr.19: 54338909-54338909 on Build GRCh38
rs1042290371 Chr.19: 54339222-54339222 on Build GRCh38
rs11880616 Chr.19: 54338833-54338833 on Build GRCh38
rs372227072 Chr.19: 54338829-54338829 on Build GRCh38
rs779067300 Chr.19: 54339174-54339174 on Build GRCh38
rs779598634 Chr.19: 54338950-54338952 on Build GRCh38
rs112690983 Chr.19: 54339188-54339188 on Build GRCh38
rs745641233 Chr.19: 54338936-54338936 on Build GRCh38
rs377681130 Chr.19: 54339045-54339045 on Build GRCh38
rs761128310 Chr.19: 54339071-54339071 on Build GRCh38
rs753386421 Chr.19: 54338824-54338824 on Build GRCh38
rs149986255 Chr.19: 54338878-54338878 on Build GRCh38
rs768815174 Chr.19: 54338951-54338951 on Build GRCh38
rs748234797 Chr.19: 54339061-54339061 on Build GRCh38
rs201646503 Chr.19: 54339146-54339146 on Build GRCh38
rs749408063 Chr.19: 54338941-54338941 on Build GRCh38
rs547017809 Chr.19: 54339227-54339227 on Build GRCh38
rs35407007 Chr.19: 54339147-54339147 on Build GRCh38
rs762870467 Chr.19: 54338868-54338868 on Build GRCh38
rs927868018 Chr.19: 54338884-54338884 on Build GRCh38
rs766927081 Chr.19: 54339022-54339022 on Build GRCh38
rs1040352390 Chr.19: 54339167-54339167 on Build GRCh38
rs35633572 Chr.19: 54338973-54338974 on Build GRCh38
rs575498078 Chr.19: 54339204-54339204 on Build GRCh38
rs751176985 Chr.19: 54338923-54338923 on Build GRCh38
rs752260829 Chr.19: 54338915-54338915 on Build GRCh38
rs990025809 Chr.19: 54339182-54339182 on Build GRCh38
rs760091456 Chr.19: 54339085-54339085 on Build GRCh38
rs773253105 Chr.19: 54339068-54339068 on Build GRCh38
rs765726825 Chr.19: 54339086-54339086 on Build GRCh38
rs1035115095 Chr.19: 54339101-54339101 on Build GRCh38
rs756549797 Chr.19: 54338823-54338823 on Build GRCh38
rs931695309 Chr.19: 54339206-54339206 on Build GRCh38
rs140673590 Chr.19: 54338972-54338972 on Build GRCh38
rs753655161 Chr.19: 54339111-54339111 on Build GRCh38
rs1040786374 Chr.19: 54338993-54338993 on Build GRCh38
rs370502743 Chr.19: 54339053-54339053 on Build GRCh38
rs749221810 Chr.19: 54339049-54339049 on Build GRCh38
rs375274738 Chr.19: 54339181-54339181 on Build GRCh38
rs367656176 Chr.19: 54339088-54339088 on Build GRCh38
rs774665185 Chr.19: 54338873-54338873 on Build GRCh38
rs369255909 Chr.19: 54338940-54338940 on Build GRCh38
rs901610127 Chr.19: 54339205-54339205 on Build GRCh38
rs117045067 Chr.19: 54339191-54339191 on Build GRCh38
rs1025367746 Chr.19: 54339247-54339247 on Build GRCh38
rs762176155 Chr.19: 54339013-54339013 on Build GRCh38
rs536601643 Chr.19: 54339226-54339226 on Build GRCh38
rs181991337 Chr.19: 54338877-54338877 on Build GRCh38
rs758507087 Chr.19: 54339135-54339135 on Build GRCh38
rs751052618 Chr.19: 54339041-54339041 on Build GRCh38
rs768694629 Chr.19: 54339054-54339054 on Build GRCh38
rs549776836 Chr.19: 54339118-54339118 on Build GRCh38
rs780830862 Chr.19: 54338831-54338832 on Build GRCh38
rs761252428 Chr.19: 54339095-54339095 on Build GRCh38
rs139382275 Chr.19: 54338962-54338962 on Build GRCh38
rs370837184 Chr.19: 54339043-54339043 on Build GRCh38
rs548572203 Chr.19: 54338806-54338806 on Build GRCh38
rs960647708 Chr.19: 54339243-54339243 on Build GRCh38
rs761071056 Chr.19: 54339257-54339257 on Build GRCh38
rs769602390 Chr.19: 54338857-54338857 on Build GRCh38
rs777344182 Chr.19: 54338839-54338839 on Build GRCh38
rs764423816 Chr.19: 54338869-54338869 on Build GRCh38
rs940664091 Chr.19: 54338989-54338989 on Build GRCh38
rs962149099 Chr.19: 54338978-54338978 on Build GRCh38
rs764932059 Chr.19: 54338926-54338926 on Build GRCh38
rs778928034 Chr.19: 54339129-54339129 on Build GRCh38
rs749943298 Chr.19: 54338866-54338867 on Build GRCh38
rs1011960930 Chr.19: 54339246-54339246 on Build GRCh38
rs201143889 Chr.19: 54338935-54338935 on Build GRCh38
rs878945022 Chr.19: 54339144-54339144 on Build GRCh38
rs567583311 Chr.19: 54339225-54339225 on Build GRCh38
rs370442169 Chr.19: 54339102-54339102 on Build GRCh38
rs374866885 Chr.19: 54338961-54338961 on Build GRCh38
rs755755461 Chr.19: 54338921-54338922 on Build GRCh38
rs373461046 Chr.19: 54338949-54338949 on Build GRCh38
rs745864452 Chr.19: 54338854-54338854 on Build GRCh38
rs778091689 Chr.19: 54339139-54339139 on Build GRCh38
rs969855196 Chr.19: 54339162-54339163 on Build GRCh38
rs1009929552 Chr.19: 54338889-54338889 on Build GRCh38
rs1040402421 Chr.19: 54339197-54339197 on Build GRCh38
rs777428278 Chr.19: 54338918-54338918 on Build GRCh38
rs11880626 Chr.19: 54339019-54339019 on Build GRCh38
rs1022034483 Chr.19: 54338837-54338837 on Build GRCh38
rs966398509 Chr.19: 54338847-54338847 on Build GRCh38
rs374472041 Chr.19: 54339108-54339108 on Build GRCh38

View Full Product Details