Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 54338970-54339369 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LILRA4
Entrez Gene ID
23547
Gene Name
leukocyte immunoglobulin like receptor A4
Gene Aliases
CD85g, ILT7
UniGene
Hs.406708
Gene Chromosome Location
Chr.19: 54333185-54339169 on Build GRCh38
Transcripts
NM_012276.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1396285 c.26T>A p.L9H Chr.19: 54339068-54339068 on Build GRCh38
3404593 c.13C>G p.L5V Chr.19: 54339081-54339081 on Build GRCh38
4755549 c.8T>C p.L3P Chr.19: 54339086-54339086 on Build GRCh38
7097249 c.30C>A p.F10L Chr.19: 54339064-54339064 on Build GRCh38
6086180 c.9C>A p.L3L Chr.19: 54339085-54339085 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs575498078 Chr.19: 54339204-54339204 on Build GRCh38
rs539157496 Chr.19: 54339268-54339268 on Build GRCh38
rs990025809 Chr.19: 54339182-54339182 on Build GRCh38
rs746792534 Chr.19: 54339034-54339034 on Build GRCh38
rs760091456 Chr.19: 54339085-54339085 on Build GRCh38
rs575402843 Chr.19: 54339295-54339295 on Build GRCh38
rs984595198 Chr.19: 54339267-54339267 on Build GRCh38
rs773253105 Chr.19: 54339068-54339068 on Build GRCh38
rs772428712 Chr.19: 54339063-54339063 on Build GRCh38
rs373330112 Chr.19: 54339017-54339017 on Build GRCh38
rs765726825 Chr.19: 54339086-54339086 on Build GRCh38
rs779193913 Chr.19: 54339055-54339055 on Build GRCh38
rs1035115095 Chr.19: 54339101-54339101 on Build GRCh38
rs538063825 Chr.19: 54339305-54339305 on Build GRCh38
rs117653309 Chr.19: 54339150-54339150 on Build GRCh38
rs774540488 Chr.19: 54339011-54339011 on Build GRCh38
rs769113398 Chr.19: 54339285-54339285 on Build GRCh38
rs931695309 Chr.19: 54339206-54339206 on Build GRCh38
rs374179725 Chr.19: 54339044-54339044 on Build GRCh38
rs753655161 Chr.19: 54339111-54339111 on Build GRCh38
rs1040786374 Chr.19: 54338993-54338993 on Build GRCh38
rs749221810 Chr.19: 54339049-54339049 on Build GRCh38
rs370502743 Chr.19: 54339053-54339053 on Build GRCh38
rs951951771 Chr.19: 54339258-54339258 on Build GRCh38
rs375274738 Chr.19: 54339181-54339181 on Build GRCh38
rs367656176 Chr.19: 54339088-54339088 on Build GRCh38
rs748253120 Chr.19: 54339130-54339130 on Build GRCh38
rs183081280 Chr.19: 54339329-54339329 on Build GRCh38
rs57986240 Chr.19: 54339311-54339311 on Build GRCh38
rs901610127 Chr.19: 54339205-54339205 on Build GRCh38
rs117045067 Chr.19: 54339191-54339191 on Build GRCh38
rs1025367746 Chr.19: 54339247-54339247 on Build GRCh38
rs779732030 Chr.19: 54339032-54339032 on Build GRCh38
rs368643601 Chr.19: 54339140-54339140 on Build GRCh38
rs762176155 Chr.19: 54339013-54339013 on Build GRCh38
rs767221445 Chr.19: 54339099-54339099 on Build GRCh38
rs536601643 Chr.19: 54339226-54339226 on Build GRCh38
rs182804716 Chr.19: 54339159-54339159 on Build GRCh38
rs758507087 Chr.19: 54339135-54339135 on Build GRCh38
rs771165902 Chr.19: 54339141-54339141 on Build GRCh38
rs751052618 Chr.19: 54339041-54339041 on Build GRCh38
rs914795352 Chr.19: 54339340-54339340 on Build GRCh38
rs768694629 Chr.19: 54339054-54339054 on Build GRCh38
rs200178658 Chr.19: 54339077-54339077 on Build GRCh38
rs549776836 Chr.19: 54339118-54339118 on Build GRCh38
rs761252428 Chr.19: 54339095-54339095 on Build GRCh38
rs370837184 Chr.19: 54339043-54339043 on Build GRCh38
rs566843411 Chr.19: 54339259-54339259 on Build GRCh38
rs960647708 Chr.19: 54339243-54339243 on Build GRCh38
rs761071056 Chr.19: 54339257-54339257 on Build GRCh38
rs558860618 Chr.19: 54339290-54339290 on Build GRCh38
rs1042290371 Chr.19: 54339222-54339222 on Build GRCh38
rs779067300 Chr.19: 54339174-54339174 on Build GRCh38
rs778928034 Chr.19: 54339129-54339129 on Build GRCh38
rs1011960930 Chr.19: 54339246-54339246 on Build GRCh38
rs112690983 Chr.19: 54339188-54339188 on Build GRCh38
rs878945022 Chr.19: 54339144-54339144 on Build GRCh38
rs1050392330 Chr.19: 54339335-54339335 on Build GRCh38
rs567583311 Chr.19: 54339225-54339225 on Build GRCh38
rs370442169 Chr.19: 54339102-54339102 on Build GRCh38
rs377681130 Chr.19: 54339045-54339045 on Build GRCh38
rs761128310 Chr.19: 54339071-54339071 on Build GRCh38
rs201646503 Chr.19: 54339146-54339146 on Build GRCh38
rs778091689 Chr.19: 54339139-54339139 on Build GRCh38
rs748234797 Chr.19: 54339061-54339061 on Build GRCh38
rs547017809 Chr.19: 54339227-54339227 on Build GRCh38
rs969855196 Chr.19: 54339162-54339163 on Build GRCh38
rs35407007 Chr.19: 54339147-54339147 on Build GRCh38
rs1040402421 Chr.19: 54339197-54339197 on Build GRCh38
rs923420020 Chr.19: 54339321-54339321 on Build GRCh38
rs11880626 Chr.19: 54339019-54339019 on Build GRCh38
rs1040352390 Chr.19: 54339167-54339167 on Build GRCh38
rs766927081 Chr.19: 54339022-54339022 on Build GRCh38
rs374472041 Chr.19: 54339108-54339108 on Build GRCh38

View Full Product Details