Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 54842776-54843272 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIR2DS4
Entrez Gene ID
3809
Gene Name
killer cell immunoglobulin like receptor, two Ig domains and short cytoplasmic tail 4
Gene Aliases
CD158I, KIR-2DS4, KIR1D, KIR2DS1, KIR412, KKA3, NKAT-8, NKAT8
UniGene
Hs.654608
Gene Chromosome Location
Chr.19: 54832676-54848569 on Build GRCh38
Transcripts
NM_001281971.1 NM_001281972.1 NM_012314.5

SNP Detail

SNP ID Chromosome Location
rs752692599 Chr.19: 54842857-54842857 on Build GRCh38
rs906161014 Chr.19: 54843135-54843135 on Build GRCh38
rs899351271 Chr.19: 54843111-54843111 on Build GRCh38
rs568952774 Chr.19: 54842887-54842887 on Build GRCh38
rs372022426 Chr.19: 54842985-54842985 on Build GRCh38
rs201291354 Chr.19: 54842931-54842931 on Build GRCh38
rs2365219 Chr.19: 54843055-54843055 on Build GRCh38
rs781526559 Chr.19: 54842884-54842884 on Build GRCh38
rs753334689 Chr.19: 54842939-54842940 on Build GRCh38
rs765922040 Chr.19: 54842910-54842910 on Build GRCh38
rs11878537 Chr.19: 54843181-54843181 on Build GRCh38
rs372272121 Chr.19: 54843149-54843149 on Build GRCh38
rs775844936 Chr.19: 54842892-54842892 on Build GRCh38
rs559283457 Chr.19: 54843091-54843091 on Build GRCh38
rs2365220 Chr.19: 54843075-54843075 on Build GRCh38
rs372861851 Chr.19: 54843229-54843229 on Build GRCh38
rs2364459 Chr.19: 54843107-54843107 on Build GRCh38
rs758960159 Chr.19: 54842916-54842916 on Build GRCh38
rs747194755 Chr.19: 54842881-54842881 on Build GRCh38
rs113046409 Chr.19: 54842833-54842833 on Build GRCh38
rs763403989 Chr.19: 54842822-54842822 on Build GRCh38
rs764230740 Chr.19: 54843061-54843061 on Build GRCh38
rs576096468 Chr.19: 54843039-54843039 on Build GRCh38
rs548275024 Chr.19: 54843224-54843224 on Build GRCh38
rs752274641 Chr.19: 54842942-54842943 on Build GRCh38
rs183048057 Chr.19: 54843211-54843211 on Build GRCh38
rs750035746 Chr.19: 54842831-54842831 on Build GRCh38
rs766434300 Chr.19: 54842937-54842938 on Build GRCh38
rs771081558 Chr.19: 54842883-54842883 on Build GRCh38
rs2262065 Chr.19: 54842912-54842912 on Build GRCh38
rs760350272 Chr.19: 54842908-54842908 on Build GRCh38
rs2886308 Chr.19: 54843129-54843129 on Build GRCh38
rs764902112 Chr.19: 54842941-54842942 on Build GRCh38
rs4020208 Chr.19: 54842859-54842859 on Build GRCh38
rs745904552 Chr.19: 54842886-54842886 on Build GRCh38
rs4969501 Chr.19: 54842861-54842861 on Build GRCh38
rs763567195 Chr.19: 54842898-54842898 on Build GRCh38
rs752565990 Chr.19: 54842919-54842919 on Build GRCh38
rs554023235 Chr.19: 54843004-54843004 on Build GRCh38
rs113681030 Chr.19: 54842812-54842812 on Build GRCh38
rs943538039 Chr.19: 54843086-54843086 on Build GRCh38
rs2364458 Chr.19: 54843019-54843019 on Build GRCh38
rs760481684 Chr.19: 54842912-54842912 on Build GRCh38
rs749770065 Chr.19: 54842958-54842958 on Build GRCh38
rs754534787 Chr.19: 54842940-54842941 on Build GRCh38
rs377242929 Chr.19: 54843231-54843231 on Build GRCh38
rs545569973 Chr.19: 54843017-54843017 on Build GRCh38
rs769305759 Chr.19: 54842959-54842959 on Build GRCh38
rs774884194 Chr.19: 54842907-54842907 on Build GRCh38
rs555971251 Chr.19: 54843033-54843033 on Build GRCh38
rs933325743 Chr.19: 54843001-54843001 on Build GRCh38
rs758105054 Chr.19: 54842944-54842945 on Build GRCh38
rs138633828 Chr.19: 54843252-54843252 on Build GRCh38
rs764819372 Chr.19: 54842917-54842917 on Build GRCh38
rs754560089 Chr.19: 54842839-54842839 on Build GRCh38
rs118177188 Chr.19: 54842797-54842797 on Build GRCh38
rs777955863 Chr.19: 54842958-54842958 on Build GRCh38
rs3952464 Chr.19: 54842828-54842828 on Build GRCh38
rs994129191 Chr.19: 54843117-54843117 on Build GRCh38
rs781265626 Chr.19: 54842939-54842939 on Build GRCh38
rs776092178 Chr.19: 54842968-54842968 on Build GRCh38
rs755829821 Chr.19: 54842832-54842832 on Build GRCh38
rs2365218 Chr.19: 54842873-54842873 on Build GRCh38
rs1051795716 Chr.19: 54843052-54843052 on Build GRCh38
rs776103842 Chr.19: 54842819-54842819 on Build GRCh38
rs189977103 Chr.19: 54843103-54843103 on Build GRCh38
rs775043581 Chr.19: 54842961-54842961 on Build GRCh38
rs563894436 Chr.19: 54843191-54843191 on Build GRCh38
rs758365048 Chr.19: 54842926-54842926 on Build GRCh38
rs576771880 Chr.19: 54843011-54843011 on Build GRCh38
rs769704296 Chr.19: 54842944-54842944 on Build GRCh38
rs542333157 Chr.19: 54843065-54843065 on Build GRCh38
rs368334586 Chr.19: 54842954-54842954 on Build GRCh38
rs879209945 Chr.19: 54843126-54843126 on Build GRCh38
rs185165419 Chr.19: 54842929-54842929 on Build GRCh38
rs533695718 Chr.19: 54843151-54843151 on Build GRCh38
rs769072782 Chr.19: 54842904-54842904 on Build GRCh38
rs777549996 Chr.19: 54842864-54842864 on Build GRCh38
rs113482692 Chr.19: 54842943-54842943 on Build GRCh38
rs555807413 Chr.19: 54843226-54843226 on Build GRCh38
rs117546449 Chr.19: 54842796-54842796 on Build GRCh38
rs369165762 Chr.19: 54843198-54843198 on Build GRCh38
rs4020682 Chr.19: 54842818-54842818 on Build GRCh38
rs778716399 Chr.19: 54842852-54842852 on Build GRCh38
rs200249551 Chr.19: 54843029-54843029 on Build GRCh38
rs561890233 Chr.19: 54843070-54843070 on Build GRCh38
rs764609706 Chr.19: 54842825-54842825 on Build GRCh38

View Full Product Details