Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 55091209-55091714 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PPP1R12C
Entrez Gene ID
54776
Gene Name
protein phosphatase 1 regulatory subunit 12C
Gene Aliases
LENG3, MBS85, p84, p85
UniGene
Hs.631579
Gene Chromosome Location
Chr.19: 55090913-55117600 on Build GRCh38
Transcripts
NM_001271618.1 NM_017607.3 XM_005259013.3 XM_011527045.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6753880 c.2215C>T p.R739C Chr.19: 55091697-55091697 on Build GRCh38
4717048 c.2239G>A p.A747T Chr.19: 55091673-55091673 on Build GRCh38
2886506 c.2316G>A p.A772A Chr.19: 55091505-55091505 on Build GRCh38
2886507 c.2279G>T p.R760L Chr.19: 55091542-55091542 on Build GRCh38
2886508 c.2279G>A p.R760H Chr.19: 55091542-55091542 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1027379738 Chr.19: 55091391-55091391 on Build GRCh38
rs367675451 Chr.19: 55091421-55091421 on Build GRCh38
rs374836820 Chr.19: 55091465-55091465 on Build GRCh38
rs747600217 Chr.19: 55091555-55091555 on Build GRCh38
rs746996507 Chr.19: 55091563-55091563 on Build GRCh38
rs138437435 Chr.19: 55091535-55091535 on Build GRCh38
rs950442014 Chr.19: 55091415-55091415 on Build GRCh38
rs761860223 Chr.19: 55091430-55091430 on Build GRCh38
rs996726524 Chr.19: 55091566-55091566 on Build GRCh38
rs935955913 Chr.19: 55091335-55091335 on Build GRCh38
rs757141388 Chr.19: 55091587-55091599 on Build GRCh38
rs553427519 Chr.19: 55091342-55091342 on Build GRCh38
rs771931355 Chr.19: 55091497-55091497 on Build GRCh38
rs570991987 Chr.19: 55091234-55091234 on Build GRCh38
rs994474731 Chr.19: 55091536-55091536 on Build GRCh38
rs886115069 Chr.19: 55091683-55091683 on Build GRCh38
rs766702126 Chr.19: 55091681-55091681 on Build GRCh38
rs777656205 Chr.19: 55091562-55091562 on Build GRCh38
rs763124099 Chr.19: 55091582-55091582 on Build GRCh38
rs983560279 Chr.19: 55091271-55091271 on Build GRCh38
rs756449579 Chr.19: 55091596-55091596 on Build GRCh38
rs376268294 Chr.19: 55091605-55091605 on Build GRCh38
rs1048934703 Chr.19: 55091688-55091688 on Build GRCh38
rs201023615 Chr.19: 55091448-55091448 on Build GRCh38
rs906463965 Chr.19: 55091357-55091357 on Build GRCh38
rs747643608 Chr.19: 55091495-55091495 on Build GRCh38
rs373537158 Chr.19: 55091475-55091475 on Build GRCh38
rs751315139 Chr.19: 55091560-55091561 on Build GRCh38
rs913350269 Chr.19: 55091496-55091496 on Build GRCh38
rs548846993 Chr.19: 55091499-55091499 on Build GRCh38
rs879084232 Chr.19: 55091432-55091432 on Build GRCh38
rs544413423 Chr.19: 55091457-55091457 on Build GRCh38
rs990171255 Chr.19: 55091576-55091576 on Build GRCh38
rs140838315 Chr.19: 55091543-55091543 on Build GRCh38
rs754979203 Chr.19: 55091455-55091455 on Build GRCh38
rs367827817 Chr.19: 55091594-55091594 on Build GRCh38
rs370873222 Chr.19: 55091541-55091541 on Build GRCh38
rs745868384 Chr.19: 55091626-55091627 on Build GRCh38
rs1009009957 Chr.19: 55091318-55091318 on Build GRCh38
rs1017491991 Chr.19: 55091332-55091332 on Build GRCh38
rs530315073 Chr.19: 55091483-55091483 on Build GRCh38
rs750733490 Chr.19: 55091595-55091595 on Build GRCh38
rs757758468 Chr.19: 55091469-55091469 on Build GRCh38
rs950887249 Chr.19: 55091262-55091262 on Build GRCh38
rs776694011 Chr.19: 55091517-55091517 on Build GRCh38
rs771965841 Chr.19: 55091674-55091674 on Build GRCh38
rs993255181 Chr.19: 55091407-55091407 on Build GRCh38
rs752182487 Chr.19: 55091540-55091540 on Build GRCh38
rs368523851 Chr.19: 55091606-55091606 on Build GRCh38
rs1003457961 Chr.19: 55091361-55091361 on Build GRCh38
rs769547446 Chr.19: 55091610-55091610 on Build GRCh38
rs757830171 Chr.19: 55091330-55091330 on Build GRCh38
rs377460629 Chr.19: 55091505-55091505 on Build GRCh38
rs752751585 Chr.19: 55091527-55091527 on Build GRCh38
rs772951489 Chr.19: 55091447-55091447 on Build GRCh38
rs767466051 Chr.19: 55091443-55091443 on Build GRCh38
rs1033795545 Chr.19: 55091376-55091376 on Build GRCh38
rs770832811 Chr.19: 55091608-55091608 on Build GRCh38
rs372522834 Chr.19: 55091454-55091454 on Build GRCh38
rs142871287 Chr.19: 55091528-55091528 on Build GRCh38
rs144853372 Chr.19: 55091547-55091547 on Build GRCh38
rs567512980 Chr.19: 55091603-55091603 on Build GRCh38
rs778425358 Chr.19: 55091466-55091466 on Build GRCh38
rs574191132 Chr.19: 55091255-55091255 on Build GRCh38
rs148545697 Chr.19: 55091523-55091523 on Build GRCh38
rs374197470 Chr.19: 55091297-55091297 on Build GRCh38
rs752094214 Chr.19: 55091593-55091593 on Build GRCh38
rs373087491 Chr.19: 55091682-55091682 on Build GRCh38
rs775828742 Chr.19: 55091612-55091612 on Build GRCh38
rs1023377949 Chr.19: 55091579-55091579 on Build GRCh38
rs894821017 Chr.19: 55091383-55091383 on Build GRCh38
rs975188721 Chr.19: 55091310-55091310 on Build GRCh38
rs774534486 Chr.19: 55091637-55091637 on Build GRCh38
rs202085356 Chr.19: 55091609-55091609 on Build GRCh38
rs376699437 Chr.19: 55091627-55091635 on Build GRCh38
rs540030322 Chr.19: 55091573-55091573 on Build GRCh38
rs769025857 Chr.19: 55091635-55091635 on Build GRCh38
rs545369271 Chr.19: 55091353-55091353 on Build GRCh38
rs536300767 Chr.19: 55091282-55091282 on Build GRCh38
rs775636221 Chr.19: 55091580-55091580 on Build GRCh38
rs1015657256 Chr.19: 55091377-55091377 on Build GRCh38
rs575277411 Chr.19: 55091397-55091397 on Build GRCh38
rs973117690 Chr.19: 55091254-55091254 on Build GRCh38
rs761990197 Chr.19: 55091644-55091644 on Build GRCh38
rs766572853 Chr.19: 55091449-55091449 on Build GRCh38
rs376539716 Chr.19: 55091567-55091567 on Build GRCh38
rs774128150 Chr.19: 55091422-55091422 on Build GRCh38
rs779851380 Chr.19: 55091470-55091470 on Build GRCh38
rs965005429 Chr.19: 55091334-55091334 on Build GRCh38
rs773190646 Chr.19: 55091498-55091498 on Build GRCh38
rs994422228 Chr.19: 55091387-55091387 on Build GRCh38
rs944365518 Chr.19: 55091471-55091471 on Build GRCh38
rs765406163 Chr.19: 55091525-55091525 on Build GRCh38
rs911607459 Chr.19: 55091619-55091625 on Build GRCh38
rs752740644 Chr.19: 55091458-55091458 on Build GRCh38
rs566932830 Chr.19: 55091680-55091680 on Build GRCh38
rs766200167 Chr.19: 55091508-55091508 on Build GRCh38
rs771636892 Chr.19: 55091560-55091560 on Build GRCh38
rs1054548875 Chr.19: 55091341-55091341 on Build GRCh38
rs921949322 Chr.19: 55091327-55091327 on Build GRCh38
rs763319371 Chr.19: 55091618-55091618 on Build GRCh38
rs768010986 Chr.19: 55091673-55091673 on Build GRCh38
rs200677433 Chr.19: 55091542-55091542 on Build GRCh38
rs565262132 Chr.19: 55091418-55091418 on Build GRCh38
rs375269938 Chr.19: 55091592-55091592 on Build GRCh38
rs368912034 Chr.19: 55091468-55091468 on Build GRCh38
rs370484633 Chr.19: 55091568-55091568 on Build GRCh38
rs768030104 Chr.19: 55091479-55091479 on Build GRCh38
rs573194810 Chr.19: 55091345-55091345 on Build GRCh38

View Full Product Details